Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
CuTATB-60 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare CuTATB-60. 
Applications
  • Gas adsorption:  Carbon dioxide
Properties
  • Maximum Surface Area: 3778 m2 / g BET surface area