Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
In-NDC Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare In-NDC. 
Applications
  • Gas adsorption:  Hydrogen gas
Properties
  • Maximum Surface Area: 1233.9 m2 / g Langmuir Surface Area