Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
MIL-101 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MIL-101. 
Applications
  • Gas adsorption: Carbon dioxide and methane gas
  • Liquid adsorption
  • Catalysis
Properties
  • Maximum Surface Area: 5900 m2 / g Langmuir surface area