Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
MOF-200 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MOF-200. 
Applications
  • Gas adsorption:  Hydrogen, methane, and carbon dioxide gas
Properties
  • Maximum Surface Area: 10400 m2 / g Langmuir Surface Area
  • Density: 0.22 g/cm3
Materials
References
  1. Furukawa, H.; Ko, N.; Go, Y. B.; Aratani, N.; Choi, S. B.; Choi, E.; Yazaydin, A. O.; Snurr, R. Q.; O'Keeffe, M.; Kim, J.; Yaghi, O. M. Science, 2010, 329, 5990.
Related Links