Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
MOF-74-Co Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MOF-74-Co. 
Applications
  • Gas Adsorption:  Hydrogen, ammonia, cyanogen chloride, sulfur dioxide gas
Properties
  • Maximum Surface Area: 1360 m2 / g BET surface area
  • One Dimensional Pores, Open Metal Sites
Materials
References
  1. Bloch, E.; Murray, L.; Queen, W.; Chavan, S.; Maximoff, S.; Bigi, J.; Krishna, R.; Peterson, V.; Grandjean, F.; Long, G.; Smit, B.; Bordiga, S.; Brown, C.; Long, J. J. Am. Chem. Soc. 2011, 133, 14814-14822.
  2. Bhattacharjee, S.; Choi, J.; Yang, S.; Choi, S.; Kim, J.; Ahn, W. J. Nanoscience and Nanotechnology 2010, 10(1), 135-141."
Related Links