Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
PCN-17(Yb) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare PCN-17(Yb). 
  • Linker: TATB (4,4',4"-s-triazine-2,4,6-triyl-tribenzoic acid)
  • Metal Source: Ytterbium Salts (Yb)
Applications
  • Gas adsorption:  Hydrogen gas
Properties
  • Maximum Surface Area: 820 m2 / g BET surface area, carbon dioxide gas
  • Pore volume = 0.34 cm3 / g
Materials
References
  1. Ma, S.; Yuan, D.; Wang, X.; Zhou, H. Inorg. Chem. 2009, 48(5), 2072-2077.
Related Links