Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
PCN-5 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare PCN-5. 
  • Linker: TATB (4,4',4"-s-triazine-2,4,6-triyl-tribenzoic acid)
  • Metal Source: Nickel Salts (Ni)
Applications
  • Gas adsorption:  Carbon dioxide, methane gas
Properties
  • Maximum Surface Area: 225 m2 / g Langmuir Surface Area
  • Pore volume = 0.13 cm3 / g
Materials
References
  1. Ma, S.; Wang, X.; Manis, E.; Collier, C.; Zhou, H. Inorg. Chem. 2007, 46(9), 3432-3434.
Related Links