Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
TIF-2 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare TIF-2. 
Applications
  • Gas adsorption
Properties
  • Maximum Surface Area: 618.2 m2 / g Langmuir Surface Area
  • Pore opening = 22.4 Å x 20.8 Å
  • Density = 1.004 g / cm3
  • Pore Volume = 0.208 cm3 / g