Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UiO-66-X Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare UiO-66-X. 
Applications
  • Separations
  • Catalysis
Properties
  • Maximum Surface Area: 1200 m2 / g Langmuir Surface Area
Materials
References
  1. Garibay, S.; Cohen, S. Chem. Commun. 2010, 46, 7700-7702.
Related Links