Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UMCM-2 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare UMCM-2. 
Applications
  • Gas adsorption:  Hydrogen, nitrogen gas
  • Catalysis
Properties
  • Maximum Surface Area: 5200 m2 / g BET surface area
Materials
References
  1. Koh, K.; Wong-Foy, A.; Matzger, A. J. Am. Chem. Soc. 2009, 131(12), 4184-4185.
Related Links