[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/US/en) Products Cart0 USEN Products ProductsApplicationsServicesResourcesSupport [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/US/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/US/en)[Microelectronics & Nanoelectronics](https://www.sigmaaldrich.com/US/en/applications/materials-science-and-engineering/microelectronics-and-nanoelectronics)Deposition Grade Silanes for Sol-Gel Processes # Deposition Grade Silanes for Sol-Gel Processes __Sol-gel processing__ is beneficial in the formation of ceramic and glass films for a number of reasons.1-4 It is a simple process that does not require exotic materials, catalysts or expensive deposition equipment. Sol-gel reactions do not employ extreme reaction conditions and the properties of the materials prepared using sol-gel approaches are easy to modify by utilizing an organically modified precursors.2,3 __Deposition Grade Silanes__ comprise alkoxysilanes and their predecessors chlorosilanes typically used in a Sol-Gel process. Transformations of alkoxysilanes during the processing proceed through a series of subsiquent hydrolysis, condensation and thermal treatment steps briefly outlined in __Figure 1__. ![alkoxysilanes](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/materials-science-and-engineering/microelectronics-and-nanoelectronics/alkoxysilanes.jpg "alkoxysilanes") __Figure 1.__Transformations of alkoxysilanes during the Sol-Gel process through a series of subsequent hydrolysis, condensation and thermal treatment steps. __Applications__ of *Deposition Grade Silanes* include but are not limited to - film coatings on glass and other appropriate surfaces1,4 - reinforcement of polymeric structures[1](https://www.sigmaaldrich.com/technical-documents/articles/materials-science/microelectronics-nanoelectronics/silane-sol-gel.html#ref), such as polyesters,16 Nafion®,17 Surlyn® - hermetic barriers for electronics and electrochemical devices5 - and others applications where Si-O-based chain assemblies are desirable.3,4 __Extensive analytical data__ available for Deposition Grade Silanes allow for a better understanding of chemical composition and properties of the materials created using these precursors as compared to those prepared form conventional silanes available commercially. __Purity__ of our Deposition Grade Silanes exceed 98%. They are fully characterized by chemical analysis and nuclear magnetic resonance (NMR). The impurities present in these materials are detected using a gas-chromatography (GC) and/or a GC-MS technique and reported on the Certificate of Analysis (CA). In addition, trace metals impurities are also reported on the CA. ## Trademarks Nafion is a registered trademark of The Chemours Company FC, LLC Surlyn is a registered trademark of E.I. du Pont de Nemours and Company ## Materials Sorry, an unexpected error has occurred Response not successful: Received status code 500 [](https://www.sigmaaldrich.com) [](https://www.sigmaaldrich.com) ### References 1\. Young SK. 2006. Material Matters, . 1(3):8. 2\. Caruso F. 2003. Colloids and Colloid Assemblies. [https://doi.org/10.1002/3527602100](https://doi.org/10.1002/3527602100) 3\. Shapes LC. 1988. Sol-Gel Technology for Thin Films, Fibers, Preforms, Electronics, and Specialty.. Klein, Ed.. Park Ridge, NJ: Noyes Publ. 4\. Pierre AC. 2002. Introduction to Sol-Gel Processing. Boston, Dodrecht, London, Kulwer: Academic Publishers . 5\. Wojcik AB, Klein LC. 1995. Transparent inorganic/organic copolymers by the Sol-Gel process: Copolymers of tetraethyl orthosilicate (TEOS), vinyl triethoxysilane (VTES) and (meth)acrylate monomers. J Sol-Gel Sci Technol. 4(1):57-66. [https://doi.org/10.1007/bf00486703](https://doi.org/10.1007/bf00486703) __Related Articles__ - [AISF: A Solid Alternative to Sulfuryl Fluoride Gas](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/fluorination/stable-alternative-aisf) - [Atomic Layer Deposition of Nanomaterials for Li-Ion Batteries, Fuel Cells, and Solar Cells](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/atomic-layer-deposition-of-nanomaterials-for-li-ion-batteries-fu) - [Carbon Nanofiber Applications & Properties](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/carbon-nanofibers) - [Graphene in Biotechnology](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/bioelectronics/graphene-in-biotechnology) - [Electronic and Biomedical Applications of High-Purity Single-Walled Nanotubes (SWNTs)](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/bioelectronics/nanointegris) - [Nature as the Source of Materials for More Sustainable Organic Electronics](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/bioelectronics/nature-as-source-of-materials-for-more-sustainable-organic-electronics) - [Substrate and Conductor Materials Towards More Sustainable Electronics](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/bioelectronics/substrate-and-conductor-materials-towards-more-sustainable-electronics) - [Graphene Quantum Dots: Properties, Synthesis & Applications](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/biosensors-and-imaging/graphene-quantum-dots) - [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search) __Related Product Categories__ - [Organosilicon Reagents](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/chemical-synthesis/organosilicon-reagents) - [Protein degrader building blocks](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/chemical-biology/protein-degrader-building-blocks) - [Environmental Accounting Supplier Toolkit](https://www.sigmaaldrich.com/US/en/life-science/ssbi/sustainability-toolkits/environmental-accounting-supplier-toolkit) - [Renewable Electricity Toolkit](https://www.sigmaaldrich.com/US/en/life-science/ssbi/sustainability-toolkits/renewable-electricity-toolkit) - [Crown Ethers and Cryptands for Advanced Applications](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/crown-ethers-cryptands-advanced-applications) - [Purification of Extracellular Vesicles with Eshmuno® Q Resin and Natrix® Q Membrane](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/membrane-chromatography/purification-extracellular-vesicles) - [Sterilizing filters for final sterile filtration](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/pharma-and-biopharma-manufacturing/final-sterile-filtration-and-filling/sterilizing-grade-membrane-filters) - [Single-use assemblies for final sterile filtration](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/pharma-and-biopharma-manufacturing/mobius-single-use-assemblies/single-use-filtration-assemblies) - [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search) Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars) Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions) Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices) Social Media [![LinkedIn icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/linkedin.svg)](https://www.linkedin.com/company/merck-life-science) [![X icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/x-icon.svg)](https://x.com/Merck_lifesci) [![Facebook Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/facebook.svg)](https://www.facebook.com/Merck.lifescience) [![Instagram Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/instagram.svg)](https://www.instagram.com/mercklifescience/) [![YouTube Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/youtube.svg)](https://www.youtube.com/@MerckLifeScience/featured) MilliporeSigma __Research. Development. Production.__ We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production. [![Vibrant M string](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/vibrant-m-string-1-rrvcrp.svg)](https://www.sigmaaldrich.com/US/en) [Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco) © 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies. Reproduction of any materials from the site is strictly forbidden without permission. [Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map) An unknown error has occured.