[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)Campylobacter Detection: Rapid Testing Solutions for the Detection of Campylobacter
# *Campylobacter* Detection: Rapid Testing Solutions for the Detection of *Campylobacter*
*Campylobacter spp.* are regarded as the leading cause of bacterial gastroenteritis in humans worldwide. Infections are usually caused by consuming cross-contaminated or insufficiently processed food (typically red meat, poultry, shellfish and unpasteurized milk). There is an increasing demand for *Campylobacter* testing in food to meet new regulations regarding contamination levels in poultry.
__Our portfolio includes:__
Dehydrated Culture Media: Unique and convenient granulated media meeting the highest industry performance standards as laid out in ISO 1113 and our AOAC approved immunoassays: The Singlepath® *Campylobacter* rapid test for the detection of *Campylobacter* in foods: Easy-to-use and one-step immunoassays in the format of “pregnancy tests” for detection of *Campylobacter* with ultimate convenience in as little as 20 minutes after enrichment.
### Reference method for *Campylobacter*
MethodISO 10272-1
ISO 11133FDA BAM Chapter 7FSIS MLG 41.00Catalog No.
__Pre-enrichment__Bolton Broth, 37 ± 1 °C;
4-6 h, microaerophilicBolton Broth, 37 ± 1 °C;
4 h, Microaerophilic
or 0 °C, 3 h + 37 °C,
2 h, microaerophilicN/A[40568](https://www.sigmaaldrich.com/US/en/product/sial/40568)
__Selective enrichment__Bolton Broth 41.5 °C;
44 ± 4 h microaerophilicBolton Broth 42° C,
20 and 44 ± 4 h
microaerophilicBPW + 2 x Blood-Free
Bolton Broth,
42 ± 1 °C; 48 ± 2 h,
microaerophilic[40568](https://www.sigmaaldrich.com/US/en/product/sial/40568)
__Isolation__mCCD agar + additional, 41.5 °C;
44 ± 4 h, microaerophilicmCCD agar or AHB Agar,
37 - 42 °C; 24 - 48 h,
microaerophilicCampy-Cefex Agar,
42 ± 1 °C; 48 ± 2 h,
microaerophilic
[1.00070](https://www.sigmaaldrich.com/US/en/product/mm/100070)
[1.00071](https://www.sigmaaldrich.com/US/en/product/mm/100071)
### Read our article by: Georgina Manning, Microbiology Focus Edition 2.1
Emerging Pathogen Group. School of Science and Technology, Nottingham Trent University, Nottingham, NG11 8NS, UK.
*georgina.manning@ntu.ac.uk*
#### *Campylobacter* is a major human pathogen worldwide
The two species *Campylobacter jejuni* and *C. coli* are responsible for the majority of human food-borne infections in the UK. In 2008 there were 49,880 reported cases of Campylobacteriosis in England and Wales (*www.hpa.org.uk*). Currently *C. jejuni* is known to account for approximately 90% of all reported cases. *C. coli*, accounting for 7-10% of the Campylobacteriosis cases, is still the fourth most common cause of gastrointestinal illness in England and Wales and therefore remains a significant health burden.
Human food poisoning is acquired through the consumption of contaminated food. Poultry is a major food source of infection and infection can occur through the consumption of undercooked poultry meat or through cross-contamination of other food products in the kitchen. In a recent report from the Food Standards Agency the prevalence of *Campylobacter* in chicken at retail was 65.2% (*www.food.gov.uk*). Other sources of infection include contaminated raw milk and water.
Symptoms of the disease can occur up to two weeks after consuming the contaminated food. They usually consist of a fever and general feeling of unwellness which is accompanied or followed by abdominal cramps and then watery, sometimes bloody, diarrhoea (Blaser and Engberg 2008). The disease is usually self-limiting after 2-7 days; however, in some cases, particularly in the very young, elderly or immuno-compromised, antibiotic treatment may be required. In more severe cases, hospitalization may result.
The mechanisms utilised by *Campylobacter* to cause disease are not yet fully defi ned. However, it is thought that the organism first colonises the small intestine before travelling to the large intestine, where it causes a local inflammatory response (Black et al. 1988). The organism is motile by way of polar *fl agella* and it can propel itself through the mucous layer on top of the epithelial cells. Adhesion to the surface of the epithelial cells followed by invasion into the cells is also thought to occur before the onset of diarrhoea. *Campylobacter* does produce a cytolethal distending toxin, however the role of this toxin in disease is not fully characterized (Ketley 1997).
*C. jejuni* and *C. coli* are Gram negative, spiral, rod shaped cells (__Figure 1__). They are microaerophilic, requiring oxygen levels between 2 and 10% for growth. These two species are classed as thermophilic *campylobacters* as they have an optimum temperature for growth of 42 °C. *Campylobacter* can often be a diffi cult organism to isolate and grow in the laboratory. The current method used for detection of *campylobacters* from foodstuff s is the EN/ISO 10272-1 method. This method relies on homogenization of a known amount of material in Bolton Broth (BB) with selective supplement and 5% lysed horse blood either microaerobically or in a Schott bottle fi lled with Bolton enrichment broth (leaving a gap at the top of no more that 2 cm) aerobically at 41.5 °C for 48 hours. However in a recent publication (*Moran et al*. 2009) a change to the ISO method was recommended, as it was found that aerobic incubation during enrichment was not very successful in isolation of the organism. Therefore, microaerobic enrichment was suggested as the preferred method, with incubation for 24 hours being suffi cient. Following incubation, a sample of the enrichment broth is then plated onto modifi ed charcoal cefoperazone deoxycholate agar (mCCDA, see __Figure 2__) with the addition of a selective supplement. Identification is usually done by colony morphology and speciation by PCR-based methods.
")
__Figure 1.__Electron micrograph of Campylobacter jejuni (Gaynor et al. 2004)
__Figure 2.__Campylobacter colonies on mCCDA
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
### References
1\.
Black RE, Levine MM, Clements ML, Hughes TP, Blaser MJ. 1988. Experimental Campylobacter jejuni Infection in Humans. Journal of Infectious Diseases. 157(3):472-479. [https://doi.org/10.1093/infdis/157.3.472](https://doi.org/10.1093/infdis/157.3.472)
2\.
2008\. Clinical Aspects of Campylobacter jejuni and Campylobacter coli Infections.99-121. [https://doi.org/10.1128/9781555815554.ch6](https://doi.org/10.1128/9781555815554.ch6)
3\.
Gaynor EC, Cawthraw S, Manning G, MacKichan JK, Falkow S, Newell DG. 2004. The Genome-Sequenced Variant of Campylobacter jejuni NCTC 11168 and the Original Clonal Clinical Isolate Differ Markedly in Colonization, Gene Expression, and Virulence-Associated Phenotypes. JB. 186(2):503-517. [https://doi.org/10.1128/jb.186.2.503-517.2004](https://doi.org/10.1128/jb.186.2.503-517.2004)
4\.
Ketley JM. 1997. Pathogenesis of Enteric Infection by Campylobacter. Microbiology. 143(1):5-21. [https://doi.org/10.1099/00221287-143-1-5](https://doi.org/10.1099/00221287-143-1-5)
5\.
Moran L, Kelly C, Madden R. 2009. Factors affecting the recovery ofCampylobacterspp. from retail packs of raw, fresh chicken using ISO 10272-1:2006. 48(5):628-632. [https://doi.org/10.1111/j.1472-765x.2009.02581.x](https://doi.org/10.1111/j.1472-765x.2009.02581.x)
__Related Articles__
- [*Lactobacilli* Basics, Testing & Identification](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/lactobacilli)
- [Microbial Detection Using Chromogenic and Fluorogenic Culture Media](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/overview-chromogenic-media)
- [Identification of Microorganisms Based on Color](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/cytology-and-microscopy/color-identification-microorganisms)
- [Culture Method for Beer Quality Control](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/culture-method-beer-quality)
- [Detection and Differentiation of Staphylococcus Aureus on Baird Parker Agar](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/detection-and-differentiation)
- [Differentiation of Escherichia coli from coliforms](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/differentiation-of-e-coli-coliforms)
- [DiluCult™ Gravimetric Dilutor & Sample Homogenizer](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/dilucult-gravimetric-dilutor-homogenizer)
- [EN ISO 11133 Microbiological Culture Media](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/en-iso-11133-water-for-the-preparation-and-performance-testing)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Molecular cloning & protein expression](https://www.sigmaaldrich.com/US/en/products/molecular-biology-and-functional-genomics/cloning-and-expression/molecular-cloning-and-protein-expression)
- [Microbial Culture Media](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/microbial-culture-media)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.