[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Pathogen & Spoilage Testing](https://www.sigmaaldrich.com/US/en/applications/microbiological-testing/pathogen-and-spoilage-testing)Vibrio Detection
# *Vibrio* Detection
__Jvo Siegrist__
*Microbiology Focus Edition 1.3*
## *Vibrio* cholerae causes cholera in humans, *Vibrio parahaemolyticus* and *Vibrio* *vulnificus* are the leading cause of seafood-associated gastroenteritis.
*Vibrio*s are motile, curved or comma-shaped bacilli and have a single polar flagella with sheet proteins. They are often found in open water, freshwater and saltwater. *Vibrio*s are facultative aerobe and Gram-negative bacterium and do not form spores. The metabolism can be oxidative and fermentative. Most species are oxidase-positive expect *V. metschnikovii*. Some *vibrio*s such as *Vibrio* fischeri exhibit bioluminescence (Quorum sensing) under certain conditions. In most ways *vibrio*s are close to Enterobacteriaceae, but share also some properties with pseudomonads. They can be differentiated from enteric bacteria by oxidase-positive reaction and motility. Differentiation from Pseudomonas can be made based on the ability of *vibrio*s to undergo oxidative and fermentative metabolism.
Most *vibrio*s are not fastidious and a simple C-source like glucose serves as an energy source. As it is a typically marine organism, most species require 2-4% NaCl or other salts and trace elements present in sea water for optimal growth. Some species are like Pseudomonas and can use diverse energy sources and show great versatility in their metabolism.
The widely used media for *Vibrio* isolation are TCBS Agar and Alkaline Peptone Water. However, accompanying sucrose-fermenting bacteria may pose a problem in the identification of *Vibrio* species on TCBS Agar\[3]. The TCBS Agar contains a mixed indicator of bromothymol blue and thymol. This system reacts upon acid production from sucrose fermentation. On a chromogenic medium like HiCrome *Vibrio* Agar (__Table 2__), the color development by *Vibrio* species is not affected by the presence of colonies of other bacteria. This is because, the amount of color developed depends on the reaction of the bacterial ß-galactosidase with the substrate contained in the media. The TCBS Agar also contains a sodium thiosulfate and ferric citrate indicator system which detects the production of hydrogen sulphide.
Pepton from animal origin provides carbonaceous, nitrogeneous and essential nutrients to the *Vibrio* species to promote growth. High concentrations of sodium chloride in the medium are used to get an inhibitory effect on the accompanying microflora. Sodium thiosulphate, sodium citrate and sodium cholate are used as well to inhibit the growth of gram-positive and some gram-negative bacteria, but not members of Enterobacteriaceae. The strongly alkaline pH of the medium is also an important tool to get selectivity for *Vibrio* species.
__Figure 1.__Vibrio vulnificus is the cause of seafood-related mortality. Scientists from Northwest Fisheries Science Center have isolated and characterized a key surface protein involved in the ability of Vibrio vulnificus to attach to shellfish, such as oysters (microscopic image from Northwest Fisheries Science Center, Seattle, USA)
*V. cholerae* is a non-invasive bacteria, affecting the small intestine by producing the cholera enterotoxins. The result is a life-threatening watery diarrhea because of activation of the adenylate cyclase in the intestinal cells. This reaction causes water and electrolytes from blood and tissues to be pumped into the intestinal tract. The rapid loss of fluids leads to dehydration, anuria, acidosis and shock. An additional loss of potassium ions may result in cardiac complications and circulatory failure. The mortality rate is very high (50-60%) if the disease is not treated. Infection source is the water or food contaminated with human feces.
__Figure 2.__HiCrome Vibrio Agar, a selective Agar with chromogenic system as differential system.
*V. parahaemolyticus* causes gastroenteritis. It is an invasive organism affecting primarily the colon tissue and excretes a presently unidentified toxin. The origin of an infection leads in most cases back to contaminated raw and improper refrigerated seafood or a fecal contamination of water and food.
*V. vulnificus* lives in warm seawater and is halophile, meaning they require salt for growth. Contaminated seafood which is eaten raw or is undercooked is in most case the source of infections and cause gastroenteritis, or a syndrome known as “primary septicemia.” Also open wounds that are exposed to seawater can lead to a wound infection.
__Table 1.__ Typical biochemical reactions
| | | | | |
|-----------------------------------------------------------|---------------|----------------------------------------------------------------|-----------------------------------|------|
| [](https://www.sigmaaldrich.com/US/en/product/SIAL/43856) | Brand | Cat. No. | Name | ISO |
| __Nonselective media__ | Sigma-Aldrich | [43856](https://www.sigmaaldrich.com/US/en/product/SIAL/43856) | Alkaline Peptone Water(ISO) | 8914 |
| | Sigma-Aldrich | [77185](https://www.sigmaaldrich.com/US/en/product/SIAL/77185) | Peptone Water | |
| | Sigma-Aldrich | [T2117](https://www.sigmaaldrich.com/US/en/product/SIAL/T2117) | Thiol broth | |
| __Nonselective media__ | Sigma-Aldrich | [62895](https://www.sigmaaldrich.com/US/en/product/SIAL/62895) | Lysine Decarboxylase Salt Broth | |
| __w/differential system__ | Sigma-Aldrich | [75315](https://www.sigmaaldrich.com/US/en/product/SIAL/75315) | OF Test Nutrient Agar | |
| | Sigma-Aldrich | [22091](https://www.sigmaaldrich.com/US/en/product/SIAL/22091) | Tryptic Soy Agar with supplement: | |
| | Sigma-Aldrich | [43856](https://www.sigmaaldrich.com/US/en/product/SIAL/43856) | TTC solution | 8914 |
| __Selective media__ | Sigma-Aldrich | [49281](https://www.sigmaaldrich.com/US/en/product/SIAL/49281) | Glucose Salt Teepol Broth | |
| __Selective media__ | Sigma-Aldrich | [43856](https://www.sigmaaldrich.com/US/en/product/SIAL/43856) | 17134 CPC-Agar (Base) | |
| __w/differential system__ | Sigma-Aldrich | [70135](https://www.sigmaaldrich.com/US/en/product/SIAL/70135) | DCLS Agar | |
| | Sigma-Aldrich | [90035](https://www.sigmaaldrich.com/US/en/product/SIAL/90035) | DCLS Agar No. 2 | |
| | Sigma-Aldrich | [92323](https://www.sigmaaldrich.com/US/en/product/SIAL/92323) | HiCrome™ Vibrio Agar | |
| | Sigma-Aldrich | [86348](https://www.sigmaaldrich.com/US/en/product/SIAL/86348) | TCBS Agar | 8914 |
Table 2.Media for enrichment, detection and differentiation of Vibrio species
| | | |
|---------------|----------------------------------------------------------------|----------------------------------|
| Brand | Cat. No. | Name |
| Sigma-Aldrich | [P9602](https://www.sigmaaldrich.com/US/en/product/SIAL/P9602) | Polymyxin B Selective Supplement |
| Sigma-Aldrich | [17779](https://www.sigmaaldrich.com/US/en/product/SIAL/17779) | TTC Solution |
Table 3.Supplements for Vibrio media
| | | |
|---------------|----------------------------------------------------------------|------------------------------------|
| Brand | Cat. No. | Name |
| Sigma-Aldrich | [18502](https://www.sigmaaldrich.com/US/en/product/SIAL/18502) | Oxidase Reagent acc. Gordon-McLeod |
| Sigma-Aldrich | [40560](https://www.sigmaaldrich.com/US/en/product/SIAL/40560) | Oxidase Strips |
| Sigma-Aldrich | [70439](https://www.sigmaaldrich.com/US/en/product/SIAL/70439) | Oxidase Test |
| Sigma-Aldrich | [07345](https://www.sigmaaldrich.com/US/en/product/SIAL/07345) | Oxidase Reagent acc. Gaby-Hadley A |
| Sigma-Aldrich | [07817](https://www.sigmaaldrich.com/US/en/product/SIAL/07817) | Oxidase Reagent acc. Gaby-Hadley B |
| Sigma-Aldrich | [49940](https://www.sigmaaldrich.com/US/en/product/SIAL/49940) | ONPG Disks |
| Sigma-Aldrich | [51138](https://www.sigmaaldrich.com/US/en/product/SIAL/51138) | Nitrate Reagent Disks Kit |
| Sigma-Aldrich | [38497](https://www.sigmaaldrich.com/US/en/product/SIAL/38497) | Nitrate Reagent A |
| Sigma-Aldrich | [39441](https://www.sigmaaldrich.com/US/en/product/SIAL/39441) | Nitrate Reagent B |
| Sigma-Aldrich | [77730](https://www.sigmaaldrich.com/US/en/product/SIAL/77730) | Gram Staining Kit |
Table 4.Biochemical products and kits for Vibrio identification and differentiation
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
### References
1\.
Thompson FL, Austin B, Swings J. 2006. The Biology of Vibrios. [https://doi.org/10.1128/9781555815714](https://doi.org/10.1128/9781555815714)
2\.
Alcamo E. (2001). Fundamentals of Microbiology, Inc.. 6th ed, Jones and Bartlett Publishers,. pg 254, 244 .
3\.
(1998). Clesceri, Greenberg and Eaton (ed.), Standard Method for the Examination of Water and Waste Water, 20th ed. American Public Health Association, Washington, D. C. .
4\.
Hara-Kudo Y, Nishina T, Nakagawa H, Konuma H, Hasegawa J, Kumagai S. 2001. Improved Method for Detection of Vibrio parahaemolyticus in Seafood. Appl. Environ. Microbiol.. 67(12):5819-5823. [https://doi.org/10.1128/aem.67.12.5819-5823.2001](https://doi.org/10.1128/aem.67.12.5819-5823.2001)
5\.
ISO 8914:1990, Microbiology -- General Guidance for the Detection of Vibrio parahaemolyticus.
6\.
(1997). Color Atlas and Textbook of Diagnostic Microbiology, . 5th edition, Lippincott Williams &Wilkins .
__Related Articles__
- [*Lactobacilli* Basics, Testing & Identification](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/lactobacilli)
- [Microbial Detection Using Chromogenic and Fluorogenic Culture Media](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/bacteriology/overview-chromogenic-media)
- [Identification of Microorganisms Based on Color](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/clinical-testing-and-diagnostics-manufacturing/cytology-and-microscopy/color-identification-microorganisms)
- [Culture Method for Beer Quality Control](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/culture-method-beer-quality)
- [Detection and Differentiation of Staphylococcus Aureus on Baird Parker Agar](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/detection-and-differentiation)
- [Differentiation of Escherichia coli from coliforms](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/differentiation-of-e-coli-coliforms)
- [DiluCult™ Gravimetric Dilutor & Sample Homogenizer](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/dilucult-gravimetric-dilutor-homogenizer)
- [EN ISO 11133 Microbiological Culture Media](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/microbiological-analysis-for-food-and-beverage/en-iso-11133-water-for-the-preparation-and-performance-testing)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Microbial Culture Media](https://www.sigmaaldrich.com/US/en/products/industrial-microbiology/microbial-culture-media)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
An unknown error has occured.