Contact Plates for Personnel and Surface Monitoring
Our ready-to-use contact plates can be used for environmental monitoring of surfaces in critical clean rooms and isolators (ICR series) as well as in non-critical areas (RT series). To minimize the risk of contamination, our contact plates are produced in clean rooms. They can be stored at room temperature and the ICRplus/RTplus versions are lockable and can be safely transported for incubation under aerobic, microaerophilic or anaerobic conditions.
Our contact plates contain either Tryptic Soy Agar (TSA), Sabouraud Dextrose Agar (SDA) or a non-animal, vegetable peptone media for minimal risk of BSE/TSE contamination. The plates are available supplemented with neutralizers of a wide range of disinfectants or β-lactam antibiotics for monitoring sanitized, dry surfaces. Neutralizers such as lecithin (L), Tween 80 (T), histidine (H) and sodium thiosulfate (Th/Thio) can inactivate residues of disinfectants, a wide range of which are used worldwide. Plates supplemented with cephalosporinase or penicillinase are used to inactivate a wide spectrum of β-lactam antibiotics.
Our ready-to-use contact plates can be used for environmental monitoring of surfaces in critical clean rooms and isolators (ICR series) as well as in non-critical areas (RT series). To minimize the risk of contamination, our contact plates are produced in clean rooms. They can be stored at room temperature and the ICRplus/RTplus versions are lockable and can be safely transported for incubation under aerobic, microaerophilic or anaerobic conditions.
Our contact plates contain either Tryptic Soy Agar (TSA), Sabouraud Dextrose Agar (SDA) or a non-animal, vegetable peptone media for minimal risk of BSE/TSE contamination. The plates are available supplemented with neutralizers of a wide range of disinfectants or β-lactam antibiotics for monitoring sanitized, dry surfaces. Neutralizers such as lecithin (L), Tween 80 (T), histidine (H) and sodium thiosulfate (Th/Thio) can inactivate residues of disinfectants, a wide range of which are used worldwide. Plates supplemented with cephalosporinase or penicillinase are used to inactivate a wide spectrum of β-lactam antibiotics.
Gamma-irradiated contact plates – ICR and ICR plus plates for isolators and critical cleanrooms
ICR/ICRplus Contact PlatesGamma-irradiated and triple bagged contact plates are suitable for use in critical cleanrooms, RABS and isolators. They can be used for surface and personnel monitoring, but also for active air monitoring with MAS® air samplers. We offer two versions in a non-lockable (ICR) and lockable (ICR plus) format. The ICR plus version is furthermore suitable for anaerobic monitoring.
Secure and reliable
  • CLOSED- or VENT-Closure: safe transport and flexible incubation conditions
  • 2D Data matrix barcode on each plate for secure identification of each plate
  • Transparent, VHP-impermeable triple-sleeve packaging for safe transportation in cleanrooms (ISO 5) and isolators
  • Produced in cleanrooms and gamma-irradiated in final packaging to minimize risk of contamination
Convenient
  • Storage at room temperature
  • Long shelf life: allows fewer incoming goods controls
  • SDA in pink plates: easily distinguish between TSA and SDA
Flexible
  • Neutralizer supplements for use in isolators after VHP decontamination
  • Broad range of formulations: total bacterial count, yeast and mold even in the presence of antimicrobial residues such as sanitizers or disinfectants or beta-lactam-antibiotics
  • Media with non-animal origin available: minimizing the risk of BSE contamination
NEW - TSA LTHThio contact ICR/ICR+
The new TSA (Tryptic soy agar) media including the four neutralizers lecithin (L), Tween 80 (T), Histidine (H) and sodium Thiosulfate (Thio) provides additional unique features:
  • 12 months shelf-life – decrease the frequencies of incoming controls – cost reduction
  • Add-ons to the certificate of analyses:
    • verification of anaerobic growth for lockable plates
    • functionality check of individual 2-D matrix code
    • disinfectant test part of the release testing
Non-irradiated contact plates - RT and RT plus plates for monitoring in controlled areas
Non-irradiated Contact PlatesOur RT and RTplus contact plates are designed for microbial monitoring of dry surfaces such as floors, walls, textiles and working garments of personnel in controlled areas.
Formulations are available for determining total aerobic microbial count (Tryptic Soy Agar = Casein Soya Bean Digest Agar = TSA or Plate Count Agar= PCA), yeast and mold count and total count for specified microorganisms. When monitoring sanitized surfaces plates neutralizers such as lecithin (L), tween 80 (T); histidine (H) and sodium thiosulfate (Th/Thio) can inactivate a wide range of disinfectants.
  • Secure & Reliable: 2D Data matrix barcode on each plate: Safe and paperless identification of individual plates
  • Convenient: Room temperature stability: Storable at site of use
Lid in CLOSED Position
  • Safe transportation from sampling area to the lab
  • Recommended for prolonged aerobic incubation period
Lid in VENT Position
  • Mandatory for incubation under microaerophilic or anaerobic conditions
  • Suitable also for short-term incubation under aerobic conditions
Materials
Product # Description
1.46783 Environmental monitoring for isolators & clean rooms (surface and air monitoring), Detection of fungi and bacteria
Packaging: 55 mm contact plate with safety lock (20 or 200 plates per box)
Synonym: TSA, CASO Agar, Casein Soybean Digest Agar with Lecithin, Tween, Histidin and Thiosulfate, Tryptone Soya Agar, Tryptic Soy Agar with neutralizer
1.46798 Surface monitoring for isolators and clean rooms in presence of ß-Lactam antibiotic (Penase)
Packaging: 55 mm contact plate with safety lock (20 or 200 plates per box)
Synonym: CASO Agar, Casein Soybean Digest Agar, Tryptic Soy Agar with Lecithin, Tween, Histidin, Thiosulfate and Penicillinase (Penase), ß-Lactamase I
1.46552 Environmental monitoring for isolators & clean rooms (surface and air monitoring), Detection of fungi and bacteria
Packaging: 55 mm contact plate with safety lock (20 or 200 plates per box)
Synonym: TSA, CASO Agar, Casein Soybean Digest Agar with neutralizer, Tryptic Soy Agar with Lecithin and Tween
1.46555 Chocolate Cont. A. +LTH-ICR+ Application: Environmental monitoring for isolators & clean rooms, Active or passive air monitoring, Detection of fastidious microorganisms
Packaging: 55 mm contact plate with safety lock (20 plates per box)
Synonym: Chocolate Agar with neutralizer, Chocolate Agar with Lecithin, Tween and Histidin
1.46697 Neutralizer A - Contact-ICR+ Application: Environmental monitoring for surfaces & personnel in critical area, Neuralization of disinfectants
Packaging: 55 mm contact plate with safety lock (20 or 200 plates per box) including neutralizer A to inactivate a wide range of disinfectants
1.46502   Rack for contact plates Size: 7 x 25 cm for 10 Contact Plates (∅ 55 mm)
Material: High-Quality Stainless Steel (1.4301 el. polished)
Suitability for: Isolators & Clean Rooms (persistent against H2O2 and disinfectants), Autoclaving, Safe and Convenient Handling due to the Locking Device
1.46501 Sab-Gluc-Cont. A +LTHTh-ICR+ Application: Environmental monitoring for isolators & clean rooms, Surface monitoring
Packaging: 55 mm contact plate with safety lock (20 or 200 plates per box)
Synonym: Sabouraud Dextrose Agar with Lecithin, Tween, Histidin and Thiosulfate, SDA with neutralizer
1.46549 SDA Contact +Cm - RT+ Application: Environmental monitoring for surfaces & personnel in non-critical area, Selective detection of yeast and molds, Room Temperature storage
Packaging: 55 mm contact plate with safety lock (20 plates per box)
Synonym: Sabouraud Dextrose Agar (SDA) with Chloramphenicol
1.46538 SDA Contact +LTHTh sel-ICR+ Application: Environmental monitoring for isolators & clean rooms, Selective detection of yeast & molds
Packaging: 55 mm contact plate with safety lock (20 or 200 plates per box)
Synonym: Selective Sabouraud Dextrose Agar with Lecithin, Tween, Histidin and Thiosulfate, SDA with neutralizer
1.46201 SDA Contact w. LTHTh - ICR Application: Environmental monitoring for isolators & clean rooms, Detection of yeast & molds
Packaging: 55 mm contact plate (20 or 200 plates per box)
Synonym: Sabouraud Dextrose Agar with Lecithin, Tween, Histidin and Thiosulfate, SDA with neutralizer
1.46195 Tr. Soy Cont. A. w. LT-ICR Application: Environmental monitoring for isolators & clean rooms (surface and air monitoring), Detection of fungi and bacteria
Packaging: 55 mm contact plate (20 or 200 plates per box)
Synonym: TSA, CASO Agar, Casein Soybean Digest Agar with Lecithin and Tween, Tryptic Soy Agar with neutralizer
1.46231 Tr. Soy Cont. A w. LTHTh-ICR Application: Environmental monitoring for isolators & clean rooms (surface and air monitoring), Detection of fungi and bacteria
Packaging: 55 mm contact plate (20 or 200 plates per box)
Synonym: TSA, CASO Agar, Casein Soybean Digest Agar with Lecithin, Tween, Histidin and Thiosulfate, Tryptone Soya Agar, Tryptic Soy Agar with neutralizer
146529 ICR-Swab Application: Presence/absence test of contamination on surfaces in cleanrooms and isolators with minimized cross-contamination risk, Determination for environmental monitoring of dry surfaces, Filling needles
Packaging: 100 pre-moistened swabs per box; triple bagged, gamma-sterilized
Synonym: Soybean-Casein Digest Medium with Neutralizers, Tryptic Soy Broth with Neutralizers [GS2] 
1.46554 TSA Contact +LTH - RT+ Application: Environmental monitoring for surfaces & personnel in non-critical area, Room Temperature storage
Packaging: 55 mm contact plate with safety lock (20 plates per pack)
Synonym: CASO Agar, Casein Soybean Digest Agar, Tryptic Soy Agar with Lecithin, Tween and Histidin
1.46539 TS-Cont. +LT +Cephase-ICR+ Application: Environmental monitoring, Surface monitoring in presence of ß-Lactam antibiotic
Packaging: 55 mm contact plate with safety lock (20 or 200 plates per box)
Synonym: TSA, CASO Agar, Casein Soybean Digest Agar, Tryptic Soy Agar with Lecithin, Tween and Cephase, ß-Lactamase II
1.46154 Contact Agar Plate Count-RT Application: Environmental monitoring determination of total count of aerobic bacteria, Room temperature storage
Packaging: 55 mm contact plate (20 plates/box)
Synonym: Tryptone Glucose Yeast Agar, Casein-Peptone Dextrose Yeast Agar, Standard Methods Agar, PCA
1.46191 Contact A. Malt Extract - RT Application: Environmental monitoring for the identification of yeasts and moulds, Room temperature storage
Packaging: 55 mm contact plate (20 plates per box)
Synonym: Malt Extract Agar
1.46200 Contact A. Tr. Soy w. LTH-RT Application: Environmental monitoring for surfaces & personnel in non-critical area, Room Temperature storage
Packaging: 55 mm contact plate (20 plates per box)
Synonym: TSA, CASO Agar, Casein Soybean Digest Agar with neutralizer, Tryptone Soya Agar, Tryptic Soy Agar with Lecithin, Tween and Histidin
1.46240 Cont. A. Tr. Soy-RT EP+USP Application: Environmental monitoring for surfaces & personnel in non-critical area, Room Temperature storage
Packaging: 55 mm contact plate (20 plates per box)
Synonym: TSA, CASO Agar, Casein Soybean Digest Agar, Tryptone Soya Agar, Tryptic Soy Agar
1.46427 MacConkey Cont. A.-RT acc EP Application: Environmental monitoring isolation of Enterobacteriaceae, Room temperature storage
Packaging: 55 mm contact plate (20 plates per box)
1.46189 ReadyPlate CT Baird-Parker ISO6888 Application: Detection of Staphylococcus aureus
Packaging: 55 mm contact plate (20 plates per box)
Synonym: Staphylococcus Selective Agar
1.46266 ReadyPlate CT DG 18 Agar ISO 21527 Application: Environmental monitoring enumeration of yeast and moulds
Packaging: 55 mm contact plate (20 plates per box)
Synonym: Dichloran 18% Glycerol Agar
1.46253 >ReadyPlate CT VRBG Agar ISO21528 Application: Environmental monitoring, detection of Enterobacteriaceae
Packaging: 55 mm contact plate (20 plates per box)
Synonym: Violet Red Bile Glucose Agar, Violet Red Bile Dextrose Agar
146530 wab Rack Size: 19,6 cm x 9,9 cm x 10,4 cm for 40 ICR Swabs
Material: Stainless steel