Simplify Your Workflow with the New DiluCult™ Gravimetric Dilutor and Sample Homogenizer Product Range
Accurate microbiology results are highly reliant upon the quality of your sample preparation.
DiluCultsOur new automatic gravimetric systems, DiluCult™ and DiluCult™ 2, can help you to streamline your enrichment sample dilution workflow efficiency. The automated gravimetric dilutor instruments with two integrated pumps, are designed to facilitate work in a microbiological safety cabinet. They are low in height and have a removable drip tray. They operate in three modes of dispensing: a fast flow with lower accuracy, a standard flow with standard accuracy, and a slow flow with optimal accuracy. DiluCult™ and DiluCult™ 2 can be extremely easy calibrated with a certified weight. Choose your specific dilution factor on the user interface or switch to a dispensing mode for other applications.
  • The DiluCult™ is a simple and flexible instrument for precise dilution that can be tailored to your needs. The authorized weight for this instrument is up to 3 kg
  • The DiluCult™ 2 has a robotic dispensing arm for higher safety and can be used for dilution of large pooling samples of up to 5 kg
ESHFor efficient solid food sample preparation, we would also like to introduce the enrichment sample homogenizer (ESH). Introduce automatic homogenization into your workflow, and with 2 removable and autoclavable massaging paddles, the ESH can be cleaned extremely easily. The instrument has a multi-function digital display which serves as the control panel, where variable speed and time can be set. The maximum volume for this instrument is 400 mL. The gap between the window door and the paddles can be adjusted manually to better accommodate your specific sample types.
  • The ESH for the automatic homogenization of solid food enrichment samples with a security dip tray
From sample preparation to analysis, we can support your entire QC workflow.
Materials