Safety at High Speed: Single-use EZ-Fit® Filtration Units
Improve the safety of your membrane filtration workflow
You don’t have to be exposed to obvious risks every time you execute your microbiological analyses. Discover a safe and fast way of performing your membrane filtration with single-use EZ-Fit® Filtration Units.
Filtration set-up with reusable funnels
Flaming for disinfection can lead to burns or blistering.
Be careful when installing the funnel on the manifold, you might clip your fingers.
Flaming manifold head Flaming funnel base and inside Cooling time with sterile water Placing filter on manifold head Installing filter funnel on manifold Membrane transfer
Flaming manifold head Flaming funnel base and inside Cooling time with sterile water Placing filter on manifold head Installing filter funnel on manifold  
5 sec 4 sec 3 to 5 sec 5 sec 3 sec  
Filtration set-up with sterile single-use EZ-Fit® Filtration Units
EZ-Fit® Filtration Units save you precious time while ensuring accurate results. The sterilized, disposable filtration devices for bioburden testing streamline your workflow.
EZ-Fit® Filtration Unit set-up Membrane transfer
EZ-Fit® Filtration
Unit set-up
1 sec  
For optimal microbial recovery, use EZ-Fit® Filtration Units with our Readyplate™ 55 plates. These 55 mm agar plates are small but mighty. The high filling volume ensures reproducible membrane transfer without bubbles and folds.
EZ-Fit® Filtration Unit, pink base
Product Number Product Description Packaging Size
EFHAW100B EZ-Fit® Filtration Unit, white membrane, 0.45µm pore size, 100 mL funnel, bulk packaging 48
EFHAW100I EZ-Fit® Filtration Unit, white membrane, 0.45µm pore size, 100 mL funnel, individual packaging 48
EFHAW250B EZ-Fit® Filtration Unit, white membrane, 0.45µm pore size, 250 mL funnel, bulk packaging 48
EFHAW250I EZ-Fit® Filtration Unit, white membrane, 0.45µm pore size, 250 mL funnel, individual packaging 48
EFHAB100B EZ-Fit® Filtration Unit, black membrane, 0.45µm pore size, 100 mL funnel, bulk packaging 48
EFHAB100I EZ-Fit® Filtration Unit, black membrane, 0.45µm pore size, 100 mL funnel, individual packaging 48
EFHAB250B EZ-Fit® Filtration Unit, black membrane, 0.45µm pore size, 100mL funnel, bulk packaging 48
EFHAB250I EZ-Fit® Filtration Unit, black membrane, 0.45µm pore size, 250 mL funnel, individual packaging 48
EZ-Fit® Filtration Unit, pink base
Product Number Product Description Packaging Size
EFHVW10IS EZ-Fit® Filtration Unit, white membrane, 0.45µm pore size, 100 mL funnel, individual packaging, no pad 48
EFHAW10MS EZ-Fit® Filtration Unit, white membrane, 0.45µm pore size, 100 mL funnel, multi packaging, no pad 48
EFHAW25BS EZ-Fit® Filtration Unit, white membrane, 0.45µm pore size, 250 mL funnel, bulk packaging, no pad 48
EFHAB10MS EZ-Fit® Filtration Unit, black membrane, 0.45µm pore size, 100 mL funnel, multi packaging, no pad 48
EFHAB25BS EZ-Fit® Filtration Unit, black membrane, 0.45µm pore size, 250 mL funnel, bulk packaging, no pad 48
EFGSW10MS EZ-Fit® Filtration Unit, white membrane, 0.22µm pore size, 100 mL funnel, multi packaging, no pad 48
EFGSW10IS EZ-Fit® Filtration Unit, white membrane, 0.22µm pore size, 100 mL funnel, individual packaging, no pad 48
EFAAW10BS EZ-Fit® Filtration Unit, white membrane, 0.8µm pore size, 100 mL funnel, bulk packaging, no pad 48
EFAAW25BS EZ-Fit® Filtration Unit, white membrane, 0.8µm pore size, 250 mL funnel, bulk packaging, no pad 48
EFAAB10BS EZ-Fit® Filtration Unit, black membrane, 0.8µm pore size, 100 mL funnel, bulk packaging, no pad 48
EFAAB25BS EZ-Fit® Filtration Unit, black membrane, 0.8µm pore size, 250 mL funnel, bulk packaging, no pad 48