EZ-Product Family for Bioburden Testing
The EZ family has been designed to optimize and streamline your bioburden workflow. Ideal for microbiological water, beverage, and medical device testing using the membrane filtration method. The family includes:
EZ-Product Family for Bioburden Testing
EZ-Fit® Filtration Units
The EZ-Fit® Filtration Unit is a disposable filtration device for microbiological testing of liquid samples, including water, in-process samples, and final products.
EZ-Fit® Filtration Units offer:
  • Drain support and optimized funnel-to-base assembly ensure optimal microbial recovery—including the new pink base for viscous or hard-to-filter samples.
  • Unique funnel design prevents any leaks and ensures filtration of the entire sample volume
  • Protective rim reduces contamination risk
  • 100 mL and 250 mL funnels accommodate a range of sample volumes
  • Compliance with water testing regulations, and conformance with international standards including EP, USP and ISO.
The blue EZ-Fit® Filtration Units come with pad and plugs
The blue EZ-Fit® Filtration Units come with pad and plugs.
Product No. Filter Color Pore Size Funnel Volume Packaging
EFHAB100B Black 0.45µm 100mL Bulk
EFHAB100I Black 0.45µm 100mL Individual
EFHAB250B Black 0.45µm 250mL Bulk
EFHAB250I Black 0.45µm 250mL Individual
EFHAW100B White 0.45µm 100mL Bulk
EFHAW100I White 0.45µm 100mL Individual
EFHAW250B White 0.45µm 250mL Bulk
EFHAW250I White 0.45µm 250mL Individual
The pink EZ-Fit® Filtration Units come without pad
The pink EZ-Fit® Filtration Units come without pad.
Product No. Membrane Color Pore Size Funnel Volume Packaging / Format
EFHVW10IS White 0.45 µm 100 mL Single
EFHAW10MS White 0.45 µm 100 mL Multipack of 4 units
EFHAW25BS White 0.45 µm 250 mL Bulk with protective bag
EFHAB10MS Black 0.45 µm 100 mL Multipack of 4 units
EFHAB25BS White 0.45 µm 250 mL Bulk with protective bag
EFGSW10MS White 0.22 μm 100 mL Multipack of 4 units
EFGSW10IS White 0.22 μm 100 mL Single
EFAAW10BS White 0.8 μm 100 mL Bulk with protective bag
EFAAW25BS White 0.8 μm 250 mL Bulk with protective bag
EFAAB10BS Black 0.8 μm 100 mL Bulk with protective bag
EFAAB25BS Black 0.8 μm 250 mL Bulk with protective bag
EZ-Stream® Vaccum Pump
EZ-Stream® Vaccum Pump
Eliminate the need for intermediate waste containers and increase efficiency by completing your filtration set-up with the EZ-Stream® Vacuum Pump. This pump can be used on the bench, or in a laminar flow hood to quietly direct waste fluid to its final destination.
EZ-Pak® Membrane Dispenser Curve
EZ-Pak® Membrane Dispenser Curve
This dispenser provides high-speed sterile membrane dispensing with no-touch operation for quick and easy membrane loading.
Product No. Description
EZSTREAM1 EZ-Stream® vacuum pump for liquid transfer
EZCURVE01 EZ-Pak® Membrane Dispenser Curve
EZCURVEAC EZ-Pak® Membrane Dispenser Curve (AC only), without battery
EZCURVCHA External power supply / charger for battery powered EZ-Pak® Membrane Dispenser Curve
EZCURVBAT Lithium-ion battery for EZ-Pak® Membrane Dispenser Curve
EZ-Fit® Manifold
EZ-Fit® ManifoldThe membrane filtration method is commonly used for in-process microbial testing. The EZ-Fit® Manifold is a filtration manifold for universal laboratory filtrations and is designed for ease of maintenance, reducing contamination risk, and workflow optimization. All internal areas of the manifold are accessible for efficient cleaning, and it is autoclavable for the prevention of biofilm formation. An integrated check valve prevents liquid backflow and the 2-way valve prevents the user from accidentally touching the filtration head during use.
The EZ-Fit® Manifold has a low height for use in laminar flow hoods. Tubing can be connected to either side of the manifold and each component can be easily removed without additional tools. 
Characteristics Specifications
Handles, valve (trigger and knob): Aluminum
Connectors, pipe, valve body: 316L stainless steel
Connectors, seals and valve seals: Ethylene propylene diene monomer (EPDM)
Filtration o-ring: Silicone
Dimensions (W x L x H) in mm 168 mm x 433 mm x 117 mm for a 3P manifold – Product No. EZFITMIC03
Weight 2.9 kg kg for a 3-place manifold – Product No. EZFITMIC03
Tubing connection Hose barb for 10mm tubing (NPS 3/8 or DN10)
Autoclaving conditions 121°C for 30 min
EZ-Fit® Manifold Components
Product No. Description
EZFITBASE1 EZ-Fit® Manifold base, 1-place, stainless steel
EZFITBASE3 EZ-Fit® Manifold base, 3-place, stainless steel
EZFITBASE6 EZ-Fit® Manifold base, 6-place, stainless steel
EZFITEXTE1 Height extension for EZ-Fit® Manifold
EZFITEXTE3 Height extension for EZ-Fit® Manifold
EZFITHIHE1 Filtration head for rubber stopper tulip for EZ-Fit® Manifold (1 unit)
EZFITHIHE3 Three filtration heads for rubber stopper tulip for EZ-Fit® Manifold (3 units)
EZFITL0HE1 Microfil® V or Microfil® S filtration head for EZ-Fit® Manifold (1 unit)
EZFITL0HE3 Microfil® V or Microfil® S filtration head for EZ-Fit® Manifold (3 units)
EZFITMIHE1 Microfil® Filtration Head for EZ-Fit® Manifold (1 unit)
EZFITMIHE3 Microfil® Filtration Head for EZ-Fit® Manifold (3 units)
EZFITMVHE1
EZ-Fit® Filtration head for EZ-Fit® Filtration Units
EZ-Fit® Manifold Replacement Parts
Product No. Description
EZFITBACKF EZ-Fit® Manifold check valve
EZFITFRIT3 EZ-Fit® Manifold removable porous filter support, stainless steel
EZFITGASK3 EZ-Fit® Manifold o-ring kit
EZFITMAKIT EZ-Fit® Manifold maintenance kit including lubricant and brush
EZFITQUICKC EZ-Fit® Manifold quick connection, 1 plug and 1 connector
EZFITVALV1 EZ-Fit® Manifold complete valve
XX2004718 No. 8 stopper, 9.5 mm (3/8 in.) hole, silicone
EZ-Pak® Filters
EZ-Pak® Filters
Product No. Filter Diameter Membrane Color Pore Size Packaging
EZAABG474
47 mm Black 0.8 µm 4 bands of 150 filters
EZAAWG474
47 mm White 0.8 µm 4 bands of 150 filters
EZGSWG474
47 mm White 0.22 µm 4 bands of 150 filters
EZHAGG474
47 mm Green 0.45 µm 4 bands of 150 filters
EZHCWG474
47 mm White 0.7 µm 4 bands of 150 filters
Microfil® Funnels and EZ-Pak® Filters
Microfil® Funnels and EZ-Pak® Filters
Product No. Funnel Volume Filter Diameter Pore Size Membrane Color
MZAABG101 100 mL 47 mm 0.8 µm Black
MZAAWG101 100 mL 47 mm 0.8 µm White
MZGSWG101 100 mL 47 mm 0.22 µm White
MZHABG101 100 mL 47 mm 0.45 µm Black
MZHAWG101 100 mL 47 mm 0.45 µm White
MZAAWG251 250 mL 47 mm 0,8 µm White
MZHABG251 250 mL 47 mm 0,45 µm Black
MZHAWG251 250 mL 47 mm 0,45 µm White