EZ-Fit® Vacuum Manifold – Product Sustainability
The EZ-Fit® Manifold was developed using design for sustainability principles to reduce environmental and health impacts for the end-user, including energy consumption and waste.
Commitment to product sustainability
The products we create help our customers improve people’s lives every day, but we recognize that every product we make also has an environmental impact, both during manufacturing and in its use. That’s why we are committed to continually improving the sustainability performance of our products.
Our products are designed to offer the highest in innovation, quality, safety and effectiveness, while at the same time helping minimize environmental impacts associated with their use. We aim to develop future-forward products and solutions that meet performance needs, result in reduced life cycle impacts and help solve global sustainability challenges.
EZ-Fit® ManifoldEZ-Fit® Manifold: universal laboratory filtration
  • Different filtration heads fit to both reusable and disposable filtration devices
  • Easy to prevent biofilms
    • easy access to all inner parts for efficient cleaning
    • each component can be removed by hand and autoclaved
  • Quick-fit connections for the vacuum tubing
  • Low height for ease of use in laminar flow hoods
Product improvement highlights
Product improvement highlights
Raw material processing
The EZ-Fit® Manifold has been developed to reduce overall material usage and especially stainless steel which is the main constituent material. Thanks to its innovative design, the amount of stainless steel used was reduced which corresponds to a weight reduction of 47% in comparison with the current Hydrosol manifold.
Product manufacturing
The EZ-Fit® Manifold has been designed to generate minimal waste during production. The final design was selected for its efficient use of materials for the base supports. The aluminum used for the supports and the valve buttons (see Figure 1) comes in a circular disc shape. The design team revised initial plans in order to maximize usage of the disc with the simple semi-circle supports.
Product components
Number Material
1 Stainless Steel
2 Aluminium
3 Ethylene propylene diene monomer (EPDM)
4 Silicone
* The valve button is composed of several parts. The main parts are made of aluminium. Some internal parts are made of stainless steel. These parts can be separated by disassembling the valve button.
Packaging
The packaging of the EZ-Fit® Manifold was also developed in order to minimize its environmental impacts. The protection of the unit was achieved with a 100% corrugated cardboard concept, instead of foam which is more common but also more difficult to recycle.
Distribution
Because the EZ-Fit® Manifold is lightweight, it requires less fuel for shipping and therefore generates less emissions. Shipping one EZ-Fit® Manifold (6-place product) by air a typical distance to reach our markets produces approximately 38% less emissions than those from the prior Hydrosol Manifold product.
Use
Autoclaving is a resource-intensive process, requiring both water and energy in order to produce highly pressurized steam to ensure proper cleaning and sterility. A 10 year lifetime has been validated for the EZ-Fit® Manifold.
Bayonet FittingDuring this period, the main environmental impacts are generated during autoclaving. Thanks to the bayonet fitting (see Figure 2 below), the filtration heads can be easily removed from the EZ-Fit® Manifold and autoclaved separately between periodic full product autoclave cleanings whereas the entire Hydrosol™ Manifold required autoclaving. This allows users to reduce the volume for autoclave by 92% and reduces the water and energy consumption accordingly during the use of the product for every cleaning. Autoclaving only the filtration heads of the EZ-Fit® Manifold results in approximately 91% less emissions than autoclaving the complete Hyrdrosol Manifold, primarily from reduced electrical consumption.
End of life
The EZ-Fit® Manifold is built from highly recyclable materials like steel and aluminum and is about 99.9% recyclable by weight. After the durable product has been used for approximately ten years it can be easily disassembled in just a few minutes into its constituent materials (see Figure 1), thanks to quick-fit connections and recycled according to your local practices and regulations.
Design for sustainability
Product Live CircleDesign for Sustainability is an approach to product development that looks to minimize the environmental and health impacts at each stage of the product life cycle from manufacturing through use to disposal. At the same time, we look to maximize the product features that improve its performance and ease of use. We incorporate sustainability considerations early in the design process before impacts are locked in and we use a set of criteria for major impact areas like energy and waste to measure improvements.
These approaches help reduce energy and water consumption, create more productive processes that minimize waste, streamline packaging and reduce associated costs. The benefits come both during the manufacturing process as well as during product usage.
EZ-Fit® Manifold: reduced impacts from design for sustainability
Reduced impacts from design for sustainability
Future improvements and opportunities
Our commitment to product sustainability is a never-ending journey. We are just beginning to identify the possibilities and potential within the life science industry. We welcome your partnership and feedback as we continue to further improve the EZ-Fit® Manifold and other products.
The information and statements in this document should not be used for comparison with other filtration manifolds’ environmental and health impacts or improvements.
Materials