EZ-Fluo Rapid Microbial Detection System
EZ-Fluo™ Rapid Detection SystemDetect contamination earlier
The EZ-Fluo system uses a non-destructive method to speed up detection of microbial contamination. Unlike many other rapid test systems, this method allows identification of microorganisms with the same sample, after they have been stained, using any standard ID technology.
  • Rapid results in one third of the time in comparison to traditional techniques
  • Flexible approach based on standard membrane filtration with reduced incubation time on media plate
  • Non-destructive method—compatible with any ID technique
  • Compact hardware, fits easily on any laboratory bench
  • Cost effective solution for routine use as well as investigation
Release your products faster
Today, more and more QC laboratories are switching to rapid methods for the detection of microbial contamination to reduce the time to result. Traditional methods require several days to provide results and are therefore of limited value when it comes to releasing products to the market faster, or for conducting investigations quickly when necessary.
Applications
The EZ-Fluo system can rapidly detect and quantify microbial contamination in any filterable sample. Applications are numerous and include testing of the following sample types:
  • Raw materials
  • In-process samples
  • Final products
Support and Services
We provide a comprehensive range of support and services to help you save your QC resources and facilitate the implementation of your EZ-Fluo system into your daily testing routine. Our highly experienced service teams can assist you with the following activities:
  • Feasibility study and on-site evaluation
  • Method development and method development consultancy
  • Equipment installation
  • Operator training
  • On-site system qualification (IQ/OQ)
  • Yearly preventive maintenance
  • Service plan
For further information on our support and services offering please contact your local sales representative.
EZ-Fluo™ Spot Counter Software
The EZ-Fluo™ camera combined with the EZ-Fluo™ Spot Counter software makes the sample reading more convenient and allows you to track records for optimized microbiology QC labs. Download the software here.  
Materials
Product No. Description Qty./Pk
EZ-Fluo System Kits
EZFKIT001WW* EZ-Fluo System standard kit, including 1 reader, 1 camera and installation CD 1
EZ-Fluo Hardware
EZFREAD01WW* EZ-Fluo reader 1
EZFCAM001 EZ-Fluo camera 1
Consumables
EZFREAG57 EZ-Fluo Reagent Kit 57 tests
MSP000814 EZ-Pak® Filters 0.45 µm 47 mm black gridded 600
EZHAWG474 EZ-Pak® Filters 0.45 µm 47 mm white gridded 600
HABG047S6 S-Pak® Filters 0.45 µm 47 mm black gridded 600
HAWG047S6 S-Pak® Filters 0.45 µm 47 mm white gridded 600
PD20047S0 Petri-Pad Petri dish, sterile, 47 mm 150
PD20047S5 Petri-Pad Petri dish, sterile, 47 mm 600
EZ-Fluo Accessories
EZDISP001 EZ-Pak® Membrane Dispenser 1
EZCURVE01 EZ-Pak® Dispenser Curve 1
EZCURVEAC EZ-Pak® Dispenser Curve (AC only) 1
EZSTREAM1 EZ-Stream® Pump 1
STREAMTUB Silicone Hose for EZ-Stream® Pump: Autoclavable; Internal diameter: 9.5 mm (3/8 in.); Length: 5.0 m (196.8 in.) 1
XX6700034 PVC Hose for EZ-Stream® Pump: Not autoclavable; Internal diameter: 9.5 mm (3/8 in.); Length: 3.0 m (118.1 in.) 1
XX6200006P Filter forceps: Traceability labels 3
TRLABEL01 Label Printing Software: available at EMDMilipore.com 1000
*Country code to be defined at ordering step.