Liquid Media & ReadyCult®
Ready-to-use Liquid Media AmpoulesLiquid Media
Designed specifically for use with the Milliflex® system, our ready-to-use culture media come in single-test 2ml plastic ampoules.
  • Nutrient media developed specifically for culturing microorganisms on membrane filters.
  • Selective media formulations to match organism type and analytical test needs.
  • Rigorous quality control procedures during manufacturing and testing ensures optimal growth and maximum shelf life.
ReadyCult® Chromogenic Media for Rapid Culture TestingReadyCult
Rapid and economical E.coli/coliform and Enterococci tests for drinking water. The EPA approved ReadyCult® test gives fast, easy-to-read color change results, and is supplied in a snap pack for minimal handling.
Related Links