Milliflex® and Milliflex Oasis® Solid Culture Media and Cassettes
Our complete range of ISO certified long-life media
Complete range of ISO certified long-life culture mediaWe offer a full range of ready-to-use agar media cassettes manufactured according to ISO certified processes with high purity ingredients for long life to use with the Milliflex Oasis® system for bioburden testing.
QC tested for pH, sterility assurance, and recovery of ATCC strains typically found in bioburden applications, each product comes with a certificate of quality and are compliant with European, US and Japanese pharmacopeia requirements.
New Milliflex Oasis® media plates
Ready-to-use solid culture media cassettes for use with the Milliflex Oasis® system for bioburden testingThe new media plates have been designed to ease your workflow steps and provide accurate results. The media material is color-coded for control of critical handling steps all along the workflow, no more media mismatch. Thanks to the Milliflex Rack & Stack® feature, the lids are locked and secured when closed to avoid any cross-contamination or drying out of the media. The incubated plates can then be assembled and locked to ensure a safe transportation.
Download the Media Selection Guide to find the right culture medium for your application.
Materials
Description Qty. Article Nos.
Milliflex Oasis® R2A low nutrient media plates 48 MMSMCRA48
Milliflex Oasis® tryptic soy media plates for detection of aerobic and anaerobic flora 48 MMSMCTS48
Milliflex Oasis® sabouraud dextrose media plates for detection of yeast & molds 48 MMSMCSD48
Milliflex® cassette sabouraud dextrose agar (w/Chloramphenicol) 48 MXSMCSP48
Milliflex® cassette m-Endo agar for the detection of coliforms 48 MXSMEND48
Milliflex® cassette PCA agar for the detection of bacteria in water 48 MXSMPCA48
Milliflex® cassette cetrimide agar with naladixic acid for detection of P. aeruginosa 48 MXSMCET48
Milliflex® cassette TSA and lecithin agar for the detection of heterotrophic microorganisms 48 MXSMTLP48
Milliflex® cassette HPC agar media, for detection of stressed heterotrophic plate count bacteria 48 MXSMHPC48
Milliflex® cassette KF strep agar for the detection of fecal streptococci 48 MXSMKFS48
Milliflex® cassette PIA agar for the detection and recovery of P aeruginosa 48 MXSMPIA48
Mac Conkey Milliflex® agar for detection of lactose fermenting bacteria and coliforms 24 MXSMCMC24
Milliflex® empty cassette for agar media 120 MXSMC0120
Milliflex® empty cassette for liquid media 120 MXLMC0120