MVP ICON® System for ATP Hygiene Monitoring & Management
The MVP ICON® is an instrument and software platform that combines HACCP and hygiene monitoring with powerful program management capabilities.
The MVP ICON® system allows you to monitor key HACCP parameters, reducing the need for multiple instruments. Available measurements include: 
  • ATP (Adenosine Triphosphate)
  • Chemical Concentration (ppm)
  • Conductivity (µS)
  • pH
  • Temperature
Assured ATP testing and sanitation management
This unique and powerful software also provides direct visibility into how well the Quality Assurance staff is implementing and maintaining the HACCP monitoring program. By tracking quality indicators, such as number of swabs used, calibration and verification records for each instrument, and re-test compliance for failed test points, QA managers can ensure their sanitation program is being properly and safely managed.
Advanced ATP testing and HACCP management technology
The MVP ICON® is the first system to provide true HACCP management capabilities. Featuring advanced photon counting sensor (PCS) technology, the MVP ICON® provides superior accuracy, sensitivity, and reproducibility for ATP, pH, temperature, conductivity, and concentration. The innovative design is sleek, lightweight and features an easy to use touch-screen interface.
HACCP data management for quality assurance
The ICON Dashboard software displays key performance metrics at-a-glance, allowing Quality Assurance managers to instantly determine if sanitation standard operating procedures (SSOPs) are being followed and are effective. Dashboard data is updated in real-time after results are sync’d. And a wide selection of print-ready reports, including Action, Trending, HACCP, and Raw Data, allow you to generate necessary documentation in seconds
Reduce time preparing for HACCP and hygiene audits
The MVP ICON® Dashboard software also features innovative print-and-present HACCP reports. These reports are customized to each company’s HACCP program requirements to help demonstrate the program is adequately and effectively followed. Print-and-present reports are generated with a click of a button and reduce the amount of time spent preparing for audits.
MVP ICON® System for ATP Testing
Materials