Rapid Surface Monitoring
Rapid Surface Monitoring In order to meet the food and beverage industry’s need for faster results, we have developed rapid microbiological solutions for surface and personnel monitoring.
Our methods are based on ATP, NAD or protein detection; ATP and NAD are known as indicators of overall surface hygiene and cleaning efficacy.
In combination with our traditional methods, our rapid testing solutions match perfectly and conveniently to fulfill your surface and personnel monitoring needs—for rapid and reliable environmental monitoring routines.
MVP ICON®
MVP ICON® hygiene monitoring system
Hygiene monitoring system (ATP test)
HY-RiSE®
HY-RiSE® hygiene monitoring test strips
Hygiene monitoring test strips (NAD Test)          
FLASH®
Flash® rapid allergen indicator
Rapid allergen-indicator protein surface test