Ready-to-Use Culture Media for Microbial Food Testing
Our prepared and ready-to-use culture media products can save you time and resources, while ensuring that your media is fully compliant with the regulations relevant to your industry.
Whether you need carry out pathogen testing or indicator organism testing in food samples, we have a full range of liquid media and pre-filled agar plates suitable for your needs.
Prepared Media for Guaranteed Compliance: ReadyPlate™ Agar Plates
For isolation and enumeration of pathogens and indicator organisms, our ReadyPlate™ pre-prepared agar plates are certified to be fully compliant with EN ISO 11133:2014, as well as with individual standards, and are quality controlled by ISO/IEC 17025:2005 accredited laboratories. Each plate is provided with a label including a data matrix code for paperless plate identification.
  • Easy visibility of compliance on product label and Certificate of Analysis
  • No time-consuming preparation
  • High reproducibility with low error rate
  • Long shelf life: 4-6 months
  • Released through ISO 17025 accredited QC lab for reduced incoming quality control tests
Ready-to-use Liquid Media: ReadyTube® Media
Discover ready-to-use media in tubes and bottles for isolation, enumeration or enrichment of microorganisms in food. Our ReadyTube® bottles and tubes are fully compliant with EN ISO 11133:2014 and as well as with individual standards, and are quality controlled by ISO/IEC 17025:2005 accredited laboratories to ensure compliance.
Our clear naming convention where the number indicates the volume in mL of media contained in each bottle or tube (e.g. ReadyTube® 200 media for each bottle in the pack contains 200 mL of media) makes it simple.
  • Easy visibility of compliance on product label and certificate of analysis
  • Safe testing with maximum convenience
  • No time-consuming preparation
  • High reproducibility with a low error rate
  • Long shelf life of 6-12 months
  • Released through ISO 17025 accredited QC lab for reduced incoming quality control tests
Ready-to-use Liquid Media Ampoules
Ready-to-use media solve numerous problems in microbiology laboratories where time, equipment and trained personnel are often in short supply. The media ampoules are available in convenient, single test 2 ml plastic ampoules, eliminating the manipulation of glass ampoules.
  • Nutrient media developed specifically for culturing microorganisms on membrane filters.
  • Selective media formulations to match organism type and analytical test needs.
  • Rigorous quality control procedures during manufacturing and testing ensures optimal growth and maximum shelf life
Materials
Ordering Information Product Group Product Description Application Sample Organism packaging Storage
MHA00MRS2 2 mL ampoule MRS Media Membrane Filtration Food and other samples Lactic Acid Bacteria 50x2 mL 2-8 °C
1462080020 ReadyTube® ReadyTube®10 Fraser Broth ISO 11290 Plating Method Food and other samples Listeria spp 20x10ml 4-12 °C
1463640006 ReadyTube® ReadyTube® 200 MRS Agar  ISO 15214  Plating Method Food and other samples Lactobacillus spp 6x200ml 2-25 °C
1464030006 ReadyTube® ReadyTube®1000 BPW ISO 6579,6887,21528 Plating Method Food and other samples Salmonella 6x1000ml 2-25 °C
1464040006 ReadyTube® ReadyTube®225 BPW ISO 6579,6887,21528 Plating Method Food and other samples Salmonella 6x225ml 2-25 °C
1464230006 ReadyTube® ReadyTube® 200 VRBL Agar  ISO 4832  Plating Method Food and other samples coliforms 6x200ml 2-25 °C
1464350006 ReadyTube® ReadyTube® 200 VRBG Agar  ISO 21528  Plating Method Food and other samples Enterobacteriacea 6x200ml 2-25 °C
1464760006 ReadyTube® ReadyTube®225 Half Fraser ISO 11290 Plating Method Food and other samples Listeria spp 6x225ml 4-12 °C
1466220100 ReadyTube® ReadyTube®12 MSRV Medium  ISO 6579 Plating Method Food and other samples Salmonella 100x12 mL 4-12 °C
1466940020 ReadyTube® ReadyTube®10 RVS Broth ISO 6579 Plating Method Food and other samples Salmonella 20x10ml 2-25 °C
1463650006 Tubes/Bottles Plate Count Agar 200ml Plating Method Food and other samples Total Count 6x200ml 2-25 °C
1464650006 Tubes/Bottles Yeast Glucose Chloramphenicol (YGC) Agar Plating Method Food and other samples Yeast and Mould 6x200ml 2-25 °C
1464050006 Tubes/Bottles Milk Plate Count Agar  Plating Method Food and other samples Total Count 6x200ml 2-25 °C
1461370020 ReadyPlate™ ReadyPlate™   Baird-Parker ISO 6888  Plating Method Food and other samples Staphylococcus  20x90 mm 15-25 °C
1461370120 ReadyPlate™ ReadyPlate™   Baird-Parker ISO 6888  Plating Method Food and other samples Staphylococcus  120x90 mm 15-25 °C
1464310020 ReadyPlate™ ReadyPlate™   TSA ISO, FDA-BAM, EP+USP Plating Method Food and other samples Total Count 20x90 mm 4-12 °C
1464310100 ReadyPlate™ ReadyPlate™   TSA ISO, FDA-BAM, EP+USP Plating Method Food and other samples Total Count 100x90 mm 4-12 °C
1461600020 ReadyPlate™ ReadyPlate™   MYP  ISO 7932, 21871  Plating Method Food and other samples Bacillus spp 20x90 mm 4-12 °C
1461610020 ReadyPlate™ ReadyPlate™   DG 18 agar  ISO 21527  Plating Method Food and other samples Yeast and Mould 20x90 mm 15-25 °C
1461610120 ReadyPlate™ ReadyPlate™  DG 18 agar  ISO 21527  Plating Method Food and other samples Yeast and Mould 120x90 mm 15-25 °C
1461860020 ReadyPlate™ ReadyPlate™  CHROM Listeria Agar ISO 11290 Plating Method Food and other samples Listeria spp 20x90 mm 4-12 °C
1462360020 ReadyPlate™ ReadyPlate™   SDA ISO, FDA-BAM, EP + USP Plating Method Food and other samples Yeast and Mould 20x90 mm 15-25 °C
1462360120 ReadyPlate™ ReadyPlate™   SDA ISO, FDA-BAM, EP + USP Plating Method Food and other samples Yeast and Mould 120x90 mm 15-25 °C
1463260020 ReadyPlate™ ReadyPlate™  CHROM TBX Agar ISO 16649 Plating Method Food and other samples E.coli 20x90 mm 4-12 °C
1467110100 ReadyPlate™ ReadyPlate™   PEMBA  ISO 21871 Plating Method Food and other samples Bacillus spp 100x90 mm 4-12 °C
1467170020 ReadyPlate™ ReadyPlate™   MRS Medium ISO 15214 Plating Method Food and other samples Lactobacillus spp 20x90 mm 4-12 °C
1467180020 ReadyPlate™ ReadyPlate™   TCBS  Agar ISO 21872 Plating Method Food and other samples Vibrio 20x90 mm 4-12 °C
1467510020 ReadyPlate™ ReadyPlate™   XLD  Agar  ISO 6579 Plating Method Food and other samples Salmonella 20x90 mm 15-25 °C
1460010020 90 mm non-branded VRBG Agar LI acc EP, ISO Plating Method Food and other samples Enterobacteriacea 20x90 mm 15-25 °C
1460010120 90 mm non-branded VRBG Agar LI acc EP, ISO Plating Method Food and other samples Enterobacteriacea 120x90 mm 15-25 °C
1461510020 90 mm non-branded Malt Extract Agar Plating Method Food and other samples Yeast and Mould 20x90 mm 4-12 °C
1462690020 90 mm non-branded Plate Count Agar Plating Method Food and other samples Total Count 20x90 mm 4-12 °C
1463210020 90 mm non-branded Bile Esculin Azide Agar Plating Method Food and other samples Enterococcus 20x90 mm 4-12 °C
1463480020 90 mm non-branded Yeast Glucose Chloramphenicol (YGC) Agar Plating Method Food and other samples Yeast and Mould 20x90 mm 4-12 °C
1467190020 90 mm non-branded Rambach Agar Plating Method Food and other samples Salmonella 20x90 mm 4-12 °C
1467190100 90 mm non-branded Rambach Agar Plating Method Food and other samples Salmonella 20x90 mm 4-12 °C