Ready-to-use Culture Media for Beverages and Water Testing
55 mm Media Plates for Microbiological Testing of Beverages and Water Samples: ReadyPlate™ 55 Plates
Microbiological testing of beverages and water samples is an important part of the QC workflow; it only takes a single microorganism to cause contamination in production that could lead to product recall and brand damage.
These 55 mm plates are filled to the top for convenient handling, allowing easy manipulation of the membrane filter to reduce the risk of handling mistakes and false positive results. In addition, the lockable and stackable design.
    ReadyPlate 55-mm Agar Plate
  • Reproducible membrane filter placement thanks to the high-level filling of the agar plate.
  • Intelligent design for better stacking and safer transport.
  • Enhanced flexibility through storage at room temperature long shelf life (6-9 months).
  • Compliant with EN ISO 11133:2014 and individual media standards.
  • Released through ISO 17025 accredited QC lab for reduced incoming quality control tests.
  • Streamline your bioburden workflow by using ReadyPlate™ 55 in combination with EZ Family.
The new compact ready-to-use ReadyPlate™ 55 frees up valuable incubator and storage space while also reducing waste.
Compliant with EN ISO 11133:2014 as well as with individual standards, and released by an ISO 17025 accredited QC lab, these plates make it easy to stay up-to-date with regulatory requirements.
Learn more about ReadyPlate™ 55 agar plates and kits and watch the video.
Ready-to-use Liquid Media Ampoules for beverages and water testing
Ready-to-use Liquid Media AmpoulesReady-to-use media solve numerous problems in microbiology laboratories where time, equipment and trained personnel are often in short supply. The media ampoules are available in convenient, single test 2 mL plastic ampoules, eliminating the manipulation of glass ampoules.
  • Nutrient media developed specifically for culturing microorganisms on membrane filters.
  • Selective media formulations to match organism type and analytical test needs.
  • Rigorous quality control procedures during manufacturing and testing ensures optimal growth and maximum shelf life
ReadyCult® media: E. coli, Coliforms and Enterococci Tests for Drinking Water
ReadyCult ResultOur ReadyCult® media provides a better way to test for the presence or absence of E. coli and Coliforms as well as for enterococci. EPA approved, the ReadyCult® test gives a fast, easy-to-read color change from light yellow to green for definitive positive results. In contrast to competitor products, ReadyCult® Coliforms enables to an optional 30-second Indole reaction for additional confirmation of E. coli.
  • Supplied in irradiated, ready-to-use snap-pack format
  • No media preparation and no autoclaving required
  • Simply add medium, incubate and read results
  • Results within 18 to 24 hours
  • Results 2-3 days earlier than with traditional methods
  • Hands-on time is less than 1 minute
  • Optional additional confirmation of E. coli within 30 seconds possible
Materials
Product No. Description Application Sample Organism
M00PMCB24 m-ColiBlue24 Broth Membrane Filtration Water Coliform, E.coli
MHA000P20 Orange Serum Media Membrane Filtration Beverage Yeast and Mould
MHA000P2D Wallerstein Differential Broth (WLD) Membrane Filtration Brewery Bacteria, Y&M
MHA000P2E m-endo Total Coliform Broth Membrane Filtration Water Coliform, E.coli
MHA000P2M M-Green /Schaffus Pottinger Membrane Filtration Water, Beverage Yeast and Mould
MHA000P2N Wallerstein Nutrient Broth Membrane Filtration Brewery Yeast and Mould
MHA000P2P Pseudomonas Selective Broth Membrane Filtration Water Pseudomonas spp
MHA000P2S Heterotrophic Plate Count (HPC) Broth Membrane Filtration Water Heterotrophic Bacteria
MHA000T2B Tryptic Soybean Broth Membrane Filtration Water and other liquids Heterotrophic Bacteria
MHA00BSM2 Brettanomyces Broth Membrane Filtration Wine and Beer Brettanomyces
MHA00P2SM Yeast and Mold Selective Broth with chloramphenicol Membrane Filtration Beverage and other liquids Yeast and Mould
MHA00P2TT Tryptone glucose extract (TGE) broth Membrane Filtration Water and other liquids Heterotrophic Bacteria
MHA00PRY2 Preservative Resistant Yeast (PRY) broth Membrane Filtration Beverage and other liquids Prspoilage organism
1461850020 Lactose TTC Agar Plating Method Water coliform, E.coli
1012950001 Readycult Coliforms 50 Presence/Absence test Water (50 ml) Coliforms, E. coli
1012980001 Readycult Coliforms 100 Presence/Absence test Water (100 ml) Coliforms, E. coli
1012990001 Readycult Enterococci 100 Presence/Absence test Water (100 ml) Coliforms, E. coli
1466890020 ReadyPlate™ CHROM CCA ISO 9308 Membrane Filtration Water Coliform, E.coli
1467570020 ReadyPlate™ 55 CCA ISO 9308 Membrane Filtration Water, Beverage Coliform, E.coli
1467580150 ReadyPlate™ 55 KIT CCA ISO 9308 Membrane Filtration Water, Beverage Coliform, E.coli
1467610020 ReadyPlate™ 55 TGE Agar Membrane Filtration Water, Beverage Total Count
1467630020 ReadyPlate™ 55 Plate Count Agar Membrane Filtration Water, Beverage Total Count
1467640150 ReadyPlate™ 55 KIT Plate Count Agar Membrane Filtration Water, Beverage Total Count
1467650020 ReadyPlate™ 55 Slanetz & Bartley ISO 7899 Membrane Filtration Water Enterococcus
1467660150 ReadyPlate™ 55 KIT Slanetz & Bartley ISO Membrane Filtration Water Enterococcus
1467670020 ReadyPlate™ 55 CN Agar ISO 16266 Membrane Filtration Water Pseudomonas spp
1467680150 ReadyPlate™ 55 KIT CN Agar ISO 16266 Membrane Filtration Water Pseudomonas spp
1467690020 ReadyPlate™ 55 m-Green Agar Membrane Filtration Water, Beverage Yeast and Mould
1467700150 ReadyPlate™ 55 KIT m-Green Agar Membrane Filtration Water, Beverage Yeast and Mould
1461210020 ReadyTube®18 Yeast Extract ISO 6222 Plating Method Water Total Count
1461210100 ReadyTube®18 Yeast Extract ISO 6222 Plating Method Water Total Count
1463520010 ReadyTube®100 Yeast Extract ISO 6222 Plating Method Water Total Count
1464250006 Orange Serum Agar Plating Method Beverage Yeast and Mould