S-Pak® Membrane Filters for Microbial Testing
Easy-to-use membrane filtersS-Pak® membrane advantages
  • Individually packed
  • Sterile
  • Ready to use
  • Easy to use
  • No hydrophobic spots
  • No toxicity
  • Good bioburden recovery
Filter and packaging: easy to use and manipulate with gloved hands
Remove the membrane with the forceps
Using sterile forceps, peel back the film and separate the
blue paper from the membrane. Remove the membrane
with the forceps.
Place the membrane and blue paper on a wet filtration support
Place the membrane and blue paper on a wet filtration support. When wet, the blue paper curls enabling you to easily remove it using forceps.
We offer a range of membranes to meet your specific filtration requirements.
Please contact us for additional information on S-Pak® Membrane filtration requirements.
For your high-volume applications, try our EZ-Pak® filters.
Materials