Culture Media and Tests for Microbial Water Testing
Routine testing of water for human consumption is essential to protect the health of consumers. We provide a variety of dehydrated and ready-to-use culture media and tests matching your testing needs.
Media
Catalog no. Product Testing organism Medium format Pack size
12398 CP ChromoSelect Agar Clostridium perfringens Powder 500 g
75605 m-CP Agar Base Clostridium perfringens Powder 500 g
93745 TSC Agar Clostridium perfringens Powder 500 g
111972 TSC Agar Clostridium perfringens Granules 500 g
110426 ChromoCult® Coliform agar acc ISO 9308-1 E. coli / Coliforms Granules 500 g
101298 ReadyCult® Coliforms 100 E. coli / Coliforms Granules 1 x 20 tests
101295 ReadyCult® Coliforms 50 E. coli / Coliforms Granules 1 x 20 tests
44655 ECD Agar E. coli / Coliforms Powder 500 g
09142 ECD ChromoSelect Agar with MUG E. coli / Coliforms Powder 500 g
104044 ENDO Agar E. coli / Coliforms Granules 500 g
E5399 Endo Agar E. coli / Coliforms Powder 500 g
70137 ENDO Agar (Base) E. coli / Coliforms Powder 500 g
107680 Lactose TTC Agar mit Tergitol® 7 E. coli / Coliforms Granules 500 g
54232 Lactose TTC Agar with Tergitol®-7 E. coli / Coliforms Powder 500 g
39734 Membrane Lactose Glucuronide Agar (MLGA) E. coli / Coliforms Powder 500 g
96961 m-FC Agar E. coli / Coliforms Powder 500 g
111278 m-FC Agar E. coli / Coliforms Granules 500 g
86455 Tergitol®-7 Agar E. coli / Coliforms Powder 500 g
70143 Mac Conkey Agar No 1 Enterobacteriaceae Powder 500 g
100072 Bile Esculin Azide Agar Enterococci (intestinal) Granules 500 g
06105 Bile Esculin Azide Agar Enterococci (intestinal) Powder 500 g
72678 Bile Esculin Azide Agar ISO 7899-2:2000 Enterococci (intestinal) Powder 500 g
101299 ReadyCult® Enterococci 100 Enterococci (intestinal) Granules 1 x 20 tests
110707 KF-Streptococcus Agar Enterococci (intestinal) Powder 500 g
60641 KF-Streptococcus Agar Enterococci (intestinal) Powder 500 g
63647 Membrane filter Enterococcus Selective Agar Enterococci (intestinal) Powder 500 g
105289 GranuCult® Slanetz-Bartley Agar (Base), acc. to ISO 7899 Enterococci (intestinal) Granules 500 g
105262 GranuCult® SLANETZ-BARTLEY agar incl. TTC acc. ISO 7899 Enterococci (intestinal) Granules 500 g
01497 Yeast Extract Agar General spoilage organisms Powder 500 g
113116 GranuCult® Yeast Extract Agar acc. ISO 6222 General spoilage organisms Granules 500 g
22470 Cetrimide Agar Pseudomonas aeruginosa Powder 500 g
107620 GranuCult® Pseudomonas CFC/CN Agar (Base) Pseudomonas aeruginosa Granules 500 g
105418 Bismuth sulfite Agar Pseudomonas aeruginosa Powder 500 g
95388 Bismuth sulfite Agar Pseudomonas aeruginosa Powder 500 g
110128 Tryptone Glucose Extract Agar (TGE Agar) Total Count/Spoiling organisms Granules 500 g
70159 Tryptone Glucose Extract Agar (TGE Agar) Total Count/Spoiling organisms Powder 500 g
107315 Sabouraud 2% Glucose Agar Yeast & Molds Granules 500 g
84086 Sabouraud 2% Glucose Agar Yeast & Molds Powder 500 g
105438 Sabouraud 4% Glucose Agar Yeast & Molds Granules 500 g
84088 Sabouraud 4% Glucose Agar Yeast & Molds Powder 500 g
Supplements
Catalog no. Supplement Function Pack size
100888 Clostridium perfringens selective supplement Inhibits yeasts & moulds, chromogenic substrate 10 vials
51962 M-CP selective Supplement I Inhibit gram-positive and most gram-negative bactreia 5 vials
82265 m-CP selective Supplement II Indicator 5 vials
P9352 Perfringens T.S.C. Supplement Inhibit gram positive bactreia 1 vials
107624 Pseudomonas CN Selective Supplement Inhibit the gram-positive and gram-negative bacteria 10 vials
861324 Rosolic acid Indicator 25 g
861324 Rosolic acid Indicator 100 g
17779 TTC Solution Chromogenic indicator 10x10 mL