AOCS Reference Mixes Available from Sigma-Aldrich
Reporter US Volume 26.4
The American Oil Chemists’ Society (AOCS) found certain Fatty Acid Methyl Ester (FAME) mixes to be useful as reference standards when performing quantitative analyses of animal and vegetable oils. The compositions of these reference mixes are cited in AOCS Method Ce 1-62.
Through its Supelco brand, Sigma-Aldrich is able to offer quantitative mixes conforming to the AOCS method requirements. The composition of each mix is similar to the fatty acid distribution of certain oils (see Table 1) and is carefully prepared by weight percent. The composition is then verified by gas chromatography using an independently prepared reference standard. A certificate of analysis is provided with each mix.
Table 1. AOCS Animal and Vegetable Reference Mixes
As your source for analytical products, we also offer a full line of specialty GC capillary columns, esterification reagents and sample preparation products used to analyze the oils as FAMEs. To learn more about these products, please visit