QuEChERS Cleanup and Analysis of Veterinary Drugs in Milk by LC/MS/MS
By: Olga I. Shimelis, and Jennifer Claus, Reporter US Volume 34.1
Introduction
The use of mass spectrometry (MS) as an analytical technique allows quick screening, identification, and confirmation of contaminants. In spite of advances that MS instrumentation brings to achieve ever more sensitivity and specificity, sample preparation and cleanup still remain an important part of the analytical workflow. In recent years, adaptation of QuEChERS methods for sample preparation has been successful as it reduces time without sacrificing sample cleanup, and is very compatible with LC/MS analysis. QuEChERS methodology, while originally used for pesticide residue testing of fruits and vegetables, has been applied to other foods such as breads, milk, and oils. In addition, the range of analyzed compounds has been broadened from pesticides to other types of contaminants (e.g., veterinary drugs).
The SPE phases that were originally used in standard QuEChERS methods included primary secondary amine (PSA) for the removal of acids, polar pigments, and sugars; graphitized carbon black (GCB) for the removal of color pigments such as chlorophyll; and silica-bonded C18 for the removal of lipid and non-polar components. Silica-bonded C18 was, until recently, the only sorbent available for the removal of fats and non-polar compounds from samples. As an addition to C18 for fat removal, zirconia-bonded sorbents (Z-Sep and Z-Sep+) have recently been successfully used for cleanup during QuEChERS in the analysis of pesticides, and have also proved to work well for analysis of environmental contaminants. This work evaluated the use of the C18 and Z-Sep+ sorbents for cleanup of an animal-derived food sample (cow’s milk) prior to the LC/MS/MS analysis of veterinary drugs
Experimental
Sample Preparation
Milk sample (2 mL) was placed into a 50 mL centrifuge tube (Product No. 55248-U). The analytes were spiked and the sample was left to sit for 30 minutes. Acetonitrile (8 mL) was added, mixed for 1 min and centrifuged. The supernatant was separated into a 15 mL centrifuge tube. Concentrated formic acid (0.1 mL) and 500 mg Z-Sep+ (Product No. 55296-U) were added or, alternatively, 500 mg of DSC-18 sorbent (Product No. 52600-U). Samples were shaken for 1 min and centrifuged. The supernatant was evaporated at 50 °C to 0.75 mL. Acetonitrile (0.15 mL) was added along with water to total volume of 1.0 mL. The samples were filtered prior to LC/MS analysis using 0.45 μm filter.
HPLC Analysis
An Ascentis® Express column with Fused-Core® particle architecture was used to achieve faster separations while using a standard Agilent 1200 HPLC system. Two different column stationary phases, C18 and Reversed Phase Amide (RPA), were tested for this separation. The RPA HPLC column was chosen because it provided better retention for more polar analytes, such as salbutamol and sulfanilamide (Figure 1). Matrix effects were evaluated by spiking the blank extracts with analytes and comparing the obtained recovery values to the calibration curves in solvent. Solvent standards were used for all quantitation.
Veterinary Drugs Using Ascentis Express RP-Amide HPLC Column
Figure 1. Veterinary Drugs Using Ascentis Express RP-Amide HPLC Column in + ESI Mode
Table 1. Veterinary Drugs and Classes Evaluated with MS/MS Parameters
Compound Class Q1 Q3 ESI
Abamectin B1a
Avermectins (anthelmintics) 890.9 567.7 +
Amoxicillin Antibiotics (beta-lactams) 366.22 208.06 +
Chloramphenicol Antibiotics (phenicols) 321.1 152.1 -
Ciprofloxacin Antibiotics (fluoquinolones) 332.2 288.2 +
Furazolidone Nitrofuranes 226.2 122.1 +
Levamisole Anthelmintic 205.2 178.2 +
Lincomycin Antibiotics (macrolides) 407.2 126.2 +
Salbutamol Beta-blockers 240.3 148.1 +
Sulfanilamide Sulfonamides 173.1 148.1 +
Results and Discussion
Development of the sample extraction procedure was based on available published literature for veterinary drug analysis.1-8 First, the “true” QuEChERS extraction into 100% acetonitrile and the salt-out protocol was tested. It was quickly determined that polar drugs, such as salbutamol and levamisol, did not extract well into pure acetonitrile. Therefore, as a compromise, 20% water was included in the final extraction solvent, and no extraction salts were used for the final procedure.
Addition of 1% formic acid to the extract prior to Z-Sep+ cleanup was necessary to improve the recoveries for compounds that had chelating properties. This additive was not necessary when C18 cleanup was used.
The removal of phospholipids, in particular, phosphatidylcholine, (PC) from milk samples was evaluated and was found to be more complete using Z-Sep+ in comparison to that using C18. The PC chromatogram in cleaned milk samples is shown in Figure 2.
Evaluation of ionization effects (Table 2) indicated that ion suppression was higher in the case of the more polar and less retained analytes: salbutamol, sulfanilamide, and lyncomycin. This ion suppression effect was more pronounced for Z-Sep+ cleaned-samples. The ion suppression effect was higher for later eluting abamectin when C18 cleanup was used. In fact, the abamectin signal was extremely low and almost non-detectable in C18-cleaned samples.
Phospholipids Present in Milk Sample After Cleanup Comparison of phosphotidylcholine in milk samples cleaned using Z-Sep+ (blue) or C18 (pink). Ion of m/z 184 was monitored
Figure 2. Phospholipids Present in Milk Sample After Cleanup Comparison of phosphotidylcholine in milk samples cleaned using Z-Sep+ (blue) or C18 (pink). Ion of m/z 184 was monitored
Table 2. Evaluation of Ionization Effects*
                                                                                                                 Blank Milk Extracts
Compounds Z-Sep+ C18
Abamectin B1a 85% 10%
Amoxicillin 102% 116%
Chloramphenicol 106% 94%
Ciprofloxacine 102% 119%
Furazolidone 113% 97%
Levamisol 101% 93%
Lincomycin 62% 86%
Salbutamol 39% 58%
Sulfanilamide 77% 89%
*Deviation from 100% indicates the presence of either ion suppression or ion enhancement.
The C18 sorbent performed well for spiked milk samples. The cleanup using C18 gave good recoveries for most tested compounds, except for abamectin. As mentioned earlier, abamectin is a late-eluting compound and its signal is subject to ion suppression under the C18 cleanup conditions. This is probably due to the presence of phospholipids that were incompletely removed during sample cleanup. The detection of abamectin was significantly better using Z-Sep+ sorbent, as in this case, cleaner samples were obtained. The use of Z-Sep+, however, was not favorable to the detection of ciprofloxacine, due to retention of that compound through chelation and to the detection of lincomycin and salbutamol, whose signals were lower due to ion suppression in the samples. Geis-Asteggiante et al. also compared multiple sorbents for use in multi-class veterinary drug analysis. They observed that Z-Sep provided the best sample cleanup, yet, C18 was their choice sorbent for analysis of animal tissue samples. It was observed that some of the analytes (floxacines and tetracyclines) chelated to the Z-Sep sorbents, and were, thus, retained, causing low recovery.1
Table 3. Recoveries of Spiked Veterinary Drugs from Milk (n=3)
Spiking Level (ng/mL)      Recoveries (%RSD)
Compounds Milk Z-Sep+ C18
Abamectin B1a 25 61%(10) 3%(100)
Amoxicillin 15 31%(7) 40%(16)
Chloramphenicol 1.25 106%(4) 100%(6)
Ciprofloxacine 5 22%(26) 73%(5)
Furazolidone 12.5 98%(13) 90%(3)
Levamisol 5 98%(7) 95%(9)
Lincomycin 5 63%(11) 75%(9)
Salbutamol 5 37%(17) 80%(13)
Sulfanilamide 20 62%(6) 77%(8)
Conclusion
Analysis of milk for veterinary drugs was performed, and two sorbents were evaluated for cleanup. For multi-class veterinary drug methods, the C18 sorbent provided better recoveries for the majority of compound classes in comparison to Z-Sep+ sorbent. On the contrary, Z-Sep+ sorbent afforded better sample cleanup and more effective removal of phospholipids. Cleanup with Z-Sep+ required the addition of formic acid to the extract to prevent retention of compounds containing acidic and chelating groups. The Z-Sep sorbents can be selectively used when analysis of individual classes of drugs is required, for example, their use is more beneficial for analysis of chloramphenicol and avermectins. The use of an HPLC column with the RP-Amide water-compatible stationary phase resulted in better retention of more polar analytes and improved peak shapes across all nine compounds of interest while using MS-compatible mobile phases.
Materials
References
  1. Geis-Asteggiante, L.; Lehotay, S.J.; Lightfield, A. R.; Dutko, T.; Ng, C.; Bluhm, L. Ruggedness testing and validation of a practical analytical method for >100 veterinary drug residues in bovine muscle by ultrahigh performance liquid chromatography–tandem mass spectrometry. J.Chromatogr A. 2012, 1258, 43-54.
  2. Kaufmann, A.; Butcher, P.; Maden, K.; Widmer, M. Quantitative multiresidue method for a bout 100 veterinary drugs in different meat matrices by sub 2-μm particulate high-performance liquid chromatography couple to time of flight mass spectrometry. J.Chromatogr A. 2008, 1194, 66-79.
  3. Kinsella, B.; Lehotay, S.; Mastovska, K.; Lightfield, A.; Furey, A.; Danaher, M. Quantitative multiresidue method for about 100 veterinary drugs in different meat matrices by sub 2-µm particulate high-performance liquid chromatography couple to time of flight mass spectrometry. Anal. Chim. Acta. 2009, 637, 196-207.
  4. Martos, P.; Jayasundara, F.; Dolbeer, J.; Jin, W.; Spilsbury, L.; Mitchell, M.; Varilla, C.; Shurmer, B. Multiclass, Multiresidue Drug Analysis, Including Aminoglycosides, in Animal Tissue Using Liquid Chromatography Couple to Tandem Mass Spectrometry. J. Agric. Food. Chem. 2010, 58, 5932-5944.
  5. Mastovska, K.; Lightfield, A. Streamlining methodology for the multiresidue analysis of beta-lactam antibiotics in bovine kidney using liquid chromatography-tandem mass spectrometry. J.Chromatogr A. 2008, 1202, 118-123.
  6. Samanidou, V.; Nisyriou, S. Multi-residue methods for confirmatory determination of antibiotics in milk. J. Sep. Sci. 2008, 31, 2068-2090.
  7. Turnipseed, S.; Anderson, W.; Karbiwnyk, C.; Madson, M.; Miller, K. Multi-class, multi-residue liquid chromatography/tandem mass spectrometry screening and confirmation methods for drug residues in milk. Rapid Commun. Mass Spectrom. 2008, 22, 1467-1480.
  8. Whelan, A.; Kinsella, B.; Furey, A.; Moloney, M.; Cantwell, H.; Lehotay, S.; Danaher, M. Determination of anthelmintic drug residues in milk using ultra high performance liquid chromatography-tandem mass spectrometry with rapid polarity switching. J.Chromatogr A. 2010, 1217, 4612-4622
Related Links