GC-MS Tuning Solutions, Internal and Surrogate Standards
By: Jim Walbridge and Kathy Kiefer, Reporter US Vol 28
Because of its sensitivity and specificity, GC-MS is a widely used technique in environmental and forensic chemistry. Some important applications within these markets include many EPA methods, drugs of abuse monitoring and fire and explosives residue investigations. To get the most out of your GC-MS analysis, it is important to choose the proper internal standards, surrogates and tuning solutions.
back to top
Tuning Solutions
Tuning solutions are utilized to “tune” or optimize MS detector parameters. This maximizes the accuracy of mass and ion abundance measurements. Some tuning solutions also contain compounds used to assess injection port inertness and GC column performance. Our tuning solutions are formulated as specified in particular EPA methods and are important tools for the qualification of GC-MS systems.
back to top
Internal Standards
GC-MS protocols typically utilize the internal standard method of quantification. Internal standards gauge the method’s ability to extract target analytes from the sample matrix, and are used to monitor and adjust instrument fluctuations. Pentafluorobenzene, 1,4-difluorobenzene, 1,4-dichlorobenzene-d4 and chlorobenzene-d5 are examples of internal standards.
back to top
Surrogate Standards
Surrogate standards are used to monitor the entire analytical process from sample preparation to instrument performance. Toluene-d8, 4-bromofluorobenzene and dibromofluorobenzene are examples of surrogate standards.
The table below lists a sample of the many GC-MS standards and tuning solutions available in our catalog. A Certificate of Analysis is supplied with each product. The single and multi-component solutions are certified for identity, purity and concentration. Each component is guaranteed to be within +/-0.5% of the stated concentration.
Learn more about our complete offering of GC-MS standards.