Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Supelco SLB-IL60 Ionic Liquid GC Columns: Better High Temperature Stability
By: Leonard M. Sidisky, Katherine K. Stenerson, Michael D. Buchanan, Reporter US Volume 33.2
The SLB®-IL60 gas chromatography (GC) column is based on an ionic liquid stationary phase platform and displays desirable features that existing non-ionic liquid columns do not. This is the third of several Reporter articles which explore various aspects of this column[1,2].
Benefits of higher temperature when performing GC include decreased analysis times, elevated bake-out to remove large non-target compounds, and analysis of additional higher boiling compounds. The SLB-IL60 column is stable to 300 °C for both programmed and isothermal use. This is 20-40 °C higher than the programmed limits and 20-50 °C higher than the isothermal limits of traditional polyethylene glycol (PEG) columns.
To illustrate the superior stability of the SLB-IL60 column, it was compared directly to five popular commercially available PEG columns, each from a different manufacturer. All columns were 30 m ~ 0.25 mm I.D., 0.25 µm dimensions, except the SLB-IL60 column, which has a 0.20 µm film thickness. Table 1 shows the maximum temperature limits for all columns tested. Complete specifications of SLB-IL60 columns are shown in Table 2.
Table 1. Maximum Temperature Limits*
Column
Isothermal °C
Programmed °C
PEG 1
280
280
PEG 2
260
270
PEG 3
250
260
PEG 4
250
260
PEG 5
280
300
SLB-IL60
300
300
* Obtained from paperwork included with commercial columns.
Table 2. SLB-IL60 Column Specifications
Application Modified (deactivated) version of SLB-IL59 provides better inertness. Selectivity more polar than PEG/wax phases, resulting in unique elution patterns. Higher maximum temperature than PEG/wax columns (300 °C compared to 270-280 °C). Excellent alternative to existing PEG/wax columns. Also a good GCxGC column choice. Launched in 2012.
USP Code None
Phase Non-bonded; 1,12-Di(tripropylphosphonium)dodecane bis(trifluoromethylsulfonyl)imide
Temp. Limits 35 °C to 300 °C (isothermal or programmed)
Thermal Stress Test
Each column was subjected to an abbreviated thermal stress test, which was developed to determine high temperature (300 °C) stability. The sequence for this test is shown in Table 3. During this test, columns were subjected to a total of 15 high-temperature runs (oven programmed from 50 °C to 300 °C with a 20 minute hold). Over the duration of the test, each column was exposed to 300 °C for a total of 300 minutes. A polar column test mix and a rapeseed oil FAME mix were run on each column before and after thermal stress.
  • The polar column test mix contains several analyte types and can be used to measure key attributes of polar columns. The normal alkanes (pentadecane, hexadecane, heptadecane, octadecane, and eicosane) are used to measure column efficiency. The alcohol (1-octanol) and ketone (2-octanone) are used to measure the presence of hydrogen-bonding sites (exposed silanols). The acid/base pair (2,6-dimethylphenol/2,6-dimethylaniline) are used to measure the acid/base characteristic of the phase surface.
  • The rapeseed oil FAME mix is based on a simple vegetable oil that contains a series of saturated and unsaturated fatty acids ranging from C14 through C24 in carbon number. The elution location of C18:3n6 (an unsaturated FAME) relative to C18:0, C20:0, and C22:0 (saturated FAMEs) is used to calculate Equivalent Chain Length (ECL) values. These are similar to retention indices, except saturated FAMEs are the markers instead of saturated alkanes and that the analytes being measured are unsaturated FAMEs. ECL values are a key indicator of a column’s polarity/selectivity for FAME applications.
Before, during, and after the thermal stress test, several indicators of chromatographic performance (FID bleed, inertness, selectivity, polarity, and retention factor) were monitored for each column.
Table 3. Thermal Stress Test Sequence
  1. Rapeseed oil FAME mix: 200 °C isothermal
  2. Polar column test mix: 155 °C isothermal for PEG columns; 130 °C isothermal for SLB-IL60 column
  3. Bleed run to column’s programmed temperature limit
  4. Five ‘300 °C’ runs: 50 °C (2 min), 15 °C/min to 300 °C (20 min)
  5. Rapeseed oil FAME mix: 200 °C isothermal
  6. Five ‘300 °C’ runs: 50 °C (2 min), 15 °C/min to 300 °C (20 min)
  7. Rapeseed oil FAME mix: 200 °C isothermal
  8. Five ‘300 °C’ runs: 50 °C (2 min), 15 °C/min to 300 °C (20 min)
  9. Rapeseed oil FAME mix: 200 °C isothermal
  10. Polar column test mix: 155 °C isothermal for PEG columns; 130 °C isothermal for SLB-IL60 column
  11. Bleed run to column’s programmed temperature limit
FID Bleed
FID bleed levels from each column operated to its programmed temperature limit were measured before and after thermal stress. The data is displayed in Figure 1 and show that exposing any of the PEG columns to 300 °C for as short as 300 minutes triggers increased FID bleed, even when the column is subsequently operated within its programmed temperature limit. Better high temperature stability is exhibited by the SLB-IL60 column, in that it maintains a lower FID bleed level before, during, and after this thermal stress.
Figure 1. FID Bleed Levels (pA) Before/After Thermal Stress Test
The observed FID bleed levels from the 1st ‘300 °C’ and the 15th ‘300 °C’ run for all columns are displayed in Figure 2. All five PEG columns produced similar FID bleed levels, with the PEG 4 and PEG 2 columns ranking as the best overall performing PEG columns for this test. As shown, the SLB-IL60 column performed significantly better for this performance indicator than any PEG column. Most impressive is that the bleed level actually decreased for the SLB-IL60 column, even after exposed to 300 °C for 300 minutes. Figure 3 shows chromatograms obtained from the PEG 4 and SLB-IL60 columns, visually depicting the different amounts of FID bleed between the best performing PEG column and the SLB-IL60 column.
Figure 2. FID Bleed Levels (pA) During Thermal Stress Test
Figure 3. FID Bleed Chromatograms During Thermal Stress Test
CONDITIONS: column 1: PEG 4, 30 m × 0.25 mm I.D., 0.25 µm; column 2: SLB-IL60, 30 m x 0.25 mm I.D., df 0.20 µm (29505-U); oven: 50 °C (2 min), 15 °C/min to 300 °C (20 min); inj. temp.: 250 °C; carrier gas: helium, 1 mL/min; detector: FID, 300 °C; injection: 1 µL, splitless; liner: 4 mm I.D., split/splitless type, single taper wool packed, FocusLiner™ design; sample: methylene chloride
Inertness
A common procedure for measuring the inertness of a column is to compare the peak height of 1-octanol relative to the peak height of n-hexadecane under isothermal oven conditions. The alcohol will interact with any exposed silanols, whereas the alkane will not. Hexadecane (16 carbon atoms) generates a greater FID response (and peak height) than octanol (8 carbons atoms), so 60–70% octanol relative peak height is typical for PEG-like columns with good inertness. Octanol relative peak height values calculated from the polar column test mix runs that were performed before and after the thermal stress test are shown in Table 4. These are the same runs described in the Selectivity section. No substantial change in octanol relative peak height was noted for any of the PEG columns or for the SLB-IL60 column. This indicates the thermal stress test did not cause any loss of inertness for any column tested.
Table 4. Octanol Relative Peak Heights (%) Before/After Thermal Stress Test
Column Before After   Column Before After
PEG 1 70 60   PEG 4 61 64
PEG 2 60 67   PEG 5 69 67
PEG 3 62 67   SLB-IL60 69 67
Selectivity
Before and after the thermal stress test, a 9-component polar column test mix was analyzed on each column. All five PEG columns produced almost identical chromatography, with the PEG 3 column performing slightly better than the other PEG columns. Figure 4 shows chromatograms obtained from the PEG 3 and SLB-IL60 columns. The elution order of 2,6-dimethylaniline and 2,6-dimethylphenol (peaks 7 and 8) is reversed on the SLB-IL60 column compared to the PEG columns.
Comparing the before and after chromatograms reveals a selectivity change on all PEG columns, most notably the decrease in the relative retention of 2,6-dimethylaniline (peak 7) and 2,6-dimethylphenol (peak 8), and the increase in the relative retention of n-eicosane (peak 9). This indicates the PEG columns became less polar, and therefore less selective for polar analytes, and more selective for non-polar analytes. Conversely, no change in selectivity was observed for the SLB-IL60 column. This indicates the SLB-IL60 column can be exposed to 300 °C without changing its phase chemistry, or altering its relative amounts of possible analyte-phase interactions.
Figure 4. Selectivity Before/After Thermal Stress Test
CONDITIONS: column 1: PEG 3, 30 m × 0.25 mm I.D., 0.25 µm; column 2: SLB-IL60, 30 m × 0.25 mm I.D., 0.20 µm (29505-U); oven: 155 °C (25 min) for PEG 3; 130 °C (25 min) for SLB-IL60; inj. temp.: 250 °C; carrier gas: helium, 20 cm/sec for PEG 3; helium, 25 cm/sec for SLB-IL60; detector: FID, 250 °C; injection: 1 µL, 100:1 split; liner: 4 mm I.D., split/splitless type, single taper wool packed FocusLiner™ design; sample: polar column test mix (47302), 9 analytes, each at 500 µg/mL in methylene chloride
Polarity
A rapeseed oil FAME mix was analyzed on each column before, during, and after the thermal stress test. ECL values for C18:3n6 were calculated for each run and are shown in Figure 5. Decreasing ECL values indicate all PEG columns became less polar, experiencing a decrease in analyte-phase interactions responsible for retention of polarizable compounds (such as C18:3n6, an unsaturated FAME) relative to analyte-phase interactions responsible for retention of non-polar compounds (such as C18:0, a saturated FAME). The same phenomenon was not observed with the SLB-IL60 column, indicating this column can be exposed to 300 °C without causing a change in polarity.
Figure 5. C18:3n6 ECL Values Before, During, and After Thermal Stress Test
Retention Factor (k)
The rapeseed oil FAME mix chromatograms were also used to calculate retention factors, based on the C24:0 FAME peak. As shown in Figure 6, all of the PEG columns exhibited an increase in retention factor. This is another indicator the thermal stress test caused the PEG columns to become less polar, developing an increase in analyte-phase interactions responsible for the retention of non-polar compounds (such as C24:0, a saturated FAME). As expected, no change in retention factor was observed with the SLB-IL60 column, indicating this column is not affected by 300 °C oven programmed runs.
Figure 6. C24:0 Retention Factors Before, During, and After Thermal Stress Test
Conclusion
Columns based on polyethylene glycol phase chemistry are widely used for a variety of applications (such as solvents and FAMEs), but are limited to use below 260-280 °C oven temperature. It may be desirable to use higher temperature to decrease analysis time, perform an elevated bake-out for removal of large non-target compounds, or analyze higher boiling compounds. However, these PEG columns cannot be exposed to a 300 °C oven temperature, even for brief periods, without adversely affecting chromatographic performance, based on indicators such as FID bleed, inertness, selectivity, polarity, and retention factor. The SLB-IL60 column is similar in selectivity to PEG columns, but can be used to 300 °C without degradation of chromatographic performance. Because the SLB-IL60 column has better high temperature stability, it can expand the use of a polar column for applications that PEG columns cannot perform.
Featured Products
Description Cat. No.
SLB-IL60 GC Columns
15 m x 0.10 mm I.D., 0.08 µm df 29503-U
20 m x 0.18 mm I.D., 0.14 µm df 29504-U
30 m x 0.25 mm I.D., 0.20 µm df 29505-U
60 m x 0.25 mm I.D., 0.20 µm df 29506-U
30 m x 0.32 mm I.D., 0.26 µm df 29508-U
60 m x 0.32 mm I.D., 0.26 µm df 29509-U
References
  1. Sidisky, L.M.; Stenerson, K.K.; Buchanan, M.D. Supleco SLB-IL60 Ionic Liquid GC Coumns: Unique Selectivity; Supelco Reporter 32.2: 29–31.
  2. Sidisky, L.M.; Stenerson, K.K.; Buchanan, M.D. Supleco SLB-IL60 Ionic Liquid GC Coumns: Improved Resolution; Supelco Reporter 33.1: 21–22.
Trademarks
  • SLB is a registered trademark of Sigma-Aldrich Co. LLC
Related Links