[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Cross-Coupling](https://www.sigmaaldrich.com/US/en/applications/chemistry-and-synthesis/synthetic-methods/cross-coupling)KitAlysis™ High-Throughput Screening Platform
# KitAlysis™ High-Throughput Screening Platform
In the high-pressure environment of discovery chemistry, time is everything. While general catalytic methods exist, substrate dependency makes it difficult to predict optimal conditions in practice. A thoughtful screening process is the best way to cover a large amount of chemical space quickly while using small amounts of often precious intermediates.
KitAlysis High-Throughput Screening Kits provide the optimal solution to quickly and efficiently identify or optimize suitable catalytic reaction conditions.
Features include:
- Microscale format requires only ~100 mg total of each substrate to run 24 unique chemical reactions (3–4 mg per reaction)
- Reaction array (catalysts, solvents, bases, and temperature) is designed and validated by practicing medicinal/process chemists in some of the largest pharma companies in the world
- All components are included. End-user supplies only substrate and commonly used lab essentials (pipettes and syringes)
- KitAlysis Labware eliminates the need to set up reactions in a glove box
KitAlysis technology showcases the most widely used catalytic methods of synthetic chemists in both academia and industry, translated into an off-the-shelf screening system. Now, any bench chemist can rapidly run 24 unique microscale reactions in parallel with conditions tailored to ensure the best possible chance of success.
## Head-to-head Comparison
Head-to-head comparison of running test reaction (Scheme 1) under standard glove box conditions and using the KitAlysis™ Benchtop Inertion Box.
### Cost Analysis
__KitAlysis™____Inhouse Cost__
__24 Reactions____1 Reaction____3 Reactions____24 Reactions__
Material Cost-$751$225$9162
Labour Cost3$98$49$147$1,176
KitAlysis Cost$245---
Total$343$124$372$2,092
Saving$1,749---
1. Minimal typical cost of purchasing new catalyst.
2. Average cost of the 12 catalyst in the kit based upon the smallest SKU size available, excludes anhydrous solvents and bases.
3. Salary cost based on chemist salary http://cen.acs.org/articles/93/ i44/ American-Chemical-Societys-Members-Earn .html. Average time to plan, set-up, and analyze (single reaction = 1 hour, 24 reactions with KitAlysis = 2 hours)
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Related Articles__
- [Jørgensen’s Organocatalysts](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/jorgensen-organocatalysts)
- [MCAT- 53™ Catalyst for Ruthenium Formation](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/mcat-53-green-ruthenium-catalyst)
- [Role of High Purity Metal Salts in Chemical Synthesis](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/role-of-high-purity-metal-salts-in-chemical-synthesis)
- [Reductive amination with 2-picoline-borane complex](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/2-picoline-borane)
- [2nd Generation Buchwald Precatalysts](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/2nd-generation-buchwald-precatalysts)
- [MeOBIPHEP](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/MeOBIPHEP)
- [An Introduction to A-Phos, its Applications and Advantages](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/a-phos)
- [Trialkylphosphine for MBH Reactions](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/air-stable-trialkylphosphine)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Buchwald catalysts and ligands](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/buchwald-catalysts-and-ligands)
- [Environmental Accounting Supplier Toolkit](https://www.sigmaaldrich.com/US/en/life-science/ssbi/sustainability-toolkits/environmental-accounting-supplier-toolkit)
- [Renewable Electricity Toolkit](https://www.sigmaaldrich.com/US/en/life-science/ssbi/sustainability-toolkits/renewable-electricity-toolkit)
- [Crown Ethers and Cryptands for Advanced Applications](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/crown-ethers-cryptands-advanced-applications)
- [Purification of Extracellular Vesicles with Eshmuno® Q Resin and Natrix® Q Membrane](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/membrane-chromatography/purification-extracellular-vesicles)
- [Sterilizing filters for final sterile filtration](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/pharma-and-biopharma-manufacturing/final-sterile-filtration-and-filling/sterilizing-grade-membrane-filters)
- [Single-use assemblies for final sterile filtration](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/pharma-and-biopharma-manufacturing/mobius-single-use-assemblies/single-use-filtration-assemblies)
- [Membrane Prefilters Improve Sterilizing Filter Capacity and Process Economics in Bioprocessing](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/prefilters-bioburden-control/membrane-prefiltration-improves-sterilizing-filter-capacity-and-process-economics)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.