Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
DMPU-HF: Novel Nucleophilic Fluorinating Reagent
Introduction
An HF-based nucleophilic fluorination reagent is ideal for cost and atom economy. The most commonly used HF-based reagents are HF-organic base complexes like Olah’s reagent (pyridine-HF) and triethylamine-HF. However, these organic bases reduce the acidity of the system and may interfere with metal catalysts. For example, pyridine may bond strongly with many transition metals.
Our new HF-based reagent DMPU-HF (802794) overcomes these drawbacks. DMPU is non-basic, non-nucleophilic, and relatively weakly coordinating (toward cationic metal). Thus, DMPU-HF has high acidity and is less likely to interfere with metal catalysts. At the same time, DMPU is an excellent hydrogen-bonding acceptor that can form a stable complex with HF. Sigma-Aldrich is proud to offer the novel fluorinating reagent DMPU-HF (802794) in collaboration with the Hammond and Xu groups.
Advantages
  • Compatible with many metal catalysts1
  • High reactivity in acid-catalyzed reactions due to higher acidity compared to pyridine-HF and triethylamine-HF2
  • Bench-top stable
Representative Scheme
Special thanks to Professor Bo Xu and Professor GB Hammond for contributing this technology spotlight.