[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/US/en) Products Cart0 USEN Products ProductsApplicationsServicesResourcesSupport [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/US/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/US/en)[Applications](https://www.sigmaaldrich.com/US/en/applications)[Chemistry & Synthesis](https://www.sigmaaldrich.com/US/en/applications/chemistry-and-synthesis)[Synthetic Methods](https://www.sigmaaldrich.com/US/en/applications/chemistry-and-synthesis/synthetic-methods)Photocatalysis # Photocatalysis ![Photoreactor in organic synthesis lab for photoredox catalysis.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/applications/chemistry-and-synthesis/photoreactor-in-organic-synthesis-lab-400x248.jpg "Photoreactor in organic synthesis lab.") Photoredox catalysis, or so-called visible light photoredox catalysis, has emerged as a powerful tool in organic synthesis, building upon the foundation set by early pioneers in the areas of radical chemistry and photochemistry. Photoredox chemistry forges new bonds via open shell pathways and facilitates the rapid assembly of complex products en route to new areas of chemical space.1-7 In the presence of visible light, photocatalysts can provide access to entirely new, previously inaccessible bond formations through a wide array of synthetic transformations including but not limited to cross-coupling, C–H Functionalization, alkene and arene functionalization, and trifluoromethylation. * * * ## Related Products Slide 1 of 21 1 of 5 [![SynLED Parallel Photoreactor](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/255/907/0ab1eb8b-d6c2-4d0f-bb74-8d520ccc217a/640/0ab1eb8b-d6c2-4d0f-bb74-8d520ccc217a.jpg) \ Sigma-Aldrich \ Z742680 \ SynLED Parallel Photoreactor](https://www.sigmaaldrich.com/US/en/product/aldrich/z742680) Quick View [![Penn PhD Photoreactor M2](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/222/484/e7392e0b-9e00-49c7-a143-96ac09875cda/640/e7392e0b-9e00-49c7-a143-96ac09875cda.jpg) \ Sigma-Aldrich \ Z744035 \ Penn PhD Photoreactor M2](https://www.sigmaaldrich.com/US/en/product/sial/z744035) Quick View [![Photo KitAlysis™ Starter Kit](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/images/268/285/a4592e7c-edb6-4ef6-9c2d-1b60595220a2/640/a4592e7c-edb6-4ef6-9c2d-1b60595220a2.jpg) \ Sigma-Aldrich \ Z742612 \ Photo KitAlysis™ Starter Kit](https://www.sigmaaldrich.com/US/en/product/aldrich/z742612) Quick View [![9-Mesityl-1,3,6,8-tetramethoxy-10-phenylacridin-10-ium tetrafluoroborate 95%](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/346/971/3b31e99c-7108-4e71-8900-436919defeab/640/3b31e99c-7108-4e71-8900-436919defeab.png) \ Sigma-Aldrich \ 900693 \ 9-Mesityl-1,3,6,8-tetramethoxy-10-phenylacridin-10-ium tetrafluoroborate](https://www.sigmaaldrich.com/US/en/product/aldrich/900693) Quick View [![2,7-Dibromo-9-mesityl-10-methylacridinium tetrafluoroborate](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/308/435/bfade09b-6e50-4e0b-84b4-2c4090f3847d/640/bfade09b-6e50-4e0b-84b4-2c4090f3847d.png) \ Sigma-Aldrich \ 909033 \ 2,7-Dibromo-9-mesityl-10-methylacridinium tetrafluoroborate](https://www.sigmaaldrich.com/US/en/product/aldrich/909033) Quick View [![2,7-Dibromo-10-methylacridone ≥95%](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/127/310/391743e1-d5f0-4485-a5fc-039c537c7467/640/391743e1-d5f0-4485-a5fc-039c537c7467.png) \ Sigma-Aldrich \ 909319 \ 2,7-Dibromo-10-methylacridone](https://www.sigmaaldrich.com/US/en/product/aldrich/909319) Quick View [![2,7-Difluoro-9-mesityl-10-methylacridinium tetrafluoroborate](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/186/851/ddb72be7-ce8b-4f8d-9711-e134f14e60f5/640/ddb72be7-ce8b-4f8d-9711-e134f14e60f5.png) \ Sigma-Aldrich \ 909254 \ 2,7-Difluoro-9-mesityl-10-methylacridinium tetrafluoroborate](https://www.sigmaaldrich.com/US/en/product/aldrich/909254) Quick View [![2,7-Dimethoxy-9-mesityl-10-methylacridinium tetrafluoroborate](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/341/614/9ad9fc29-5274-4fc8-b11d-d58ec9cca41e/640/9ad9fc29-5274-4fc8-b11d-d58ec9cca41e.png) \ Sigma-Aldrich \ 909327 \ 2,7-Dimethoxy-9-mesityl-10-methylacridinium tetrafluoroborate](https://www.sigmaaldrich.com/US/en/product/aldrich/909327) Quick View [![10-(3,5-Dimethoxyphenyl)-9-mesityl-1,3,6,8-tetramethoxyacridin-10-ium tetrafluoroborate ≥93%](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/135/201/072e2043-4e54-476a-a8bc-5f7d273c9b0f/640/072e2043-4e54-476a-a8bc-5f7d273c9b0f.png) \ Sigma-Aldrich \ 900694 \ 10-(3,5-Dimethoxyphenyl)-9-mesityl-1,3,6,8-tetramethoxyacridin-10-ium tetrafluoroborate](https://www.sigmaaldrich.com/US/en/product/aldrich/900694) Quick View [![2,7-Dimethyl-9-mesityl-10-methylacridinium tetrafluoroborate](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/408/833/15f40532-5803-4c0b-b6ef-64ffc8e0d8e9/640/15f40532-5803-4c0b-b6ef-64ffc8e0d8e9.png) \ Sigma-Aldrich \ 909181 \ 2,7-Dimethyl-9-mesityl-10-methylacridinium tetrafluoroborate](https://www.sigmaaldrich.com/US/en/product/aldrich/909181) Quick View [![9-Mesityl-3,6-di-tert-butyl-10-phenylacridinium tetrafluoroborate ≥95%](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/193/970/56bd172e-aaa6-433c-8fbc-0d302049a47d/640/56bd172e-aaa6-433c-8fbc-0d302049a47d.png) \ Sigma-Aldrich \ 900421 \ 9-Mesityl-3,6-di-*tert*-butyl-10-phenylacridinium tetrafluoroborate](https://www.sigmaaldrich.com/US/en/product/aldrich/900421) Quick View [![Ir\[p-F(t-Bu)-ppy\]3](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/189/186/7badaac3-82af-4109-aab5-dea3a3aa916d/640/7badaac3-82af-4109-aab5-dea3a3aa916d.png) \ Sigma-Aldrich \ 900539 \ Ir\[*p*-F(t-Bu)-ppy\]3](https://www.sigmaaldrich.com/US/en/product/aldrich/900539) Quick View [![Ir(p-CF3-ppy)3 ≥95%](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/258/573/0173414a-1c46-4e6f-aeb4-b670009ddb89/640/0173414a-1c46-4e6f-aeb4-b670009ddb89.png) \ Sigma-Aldrich \ 904694 \ Ir(p-CF3-ppy)3](https://www.sigmaaldrich.com/US/en/product/aldrich/904694) Quick View [![Ir\[dF(t-Bu)-ppy\]3](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/254/294/d0fb19e5-05b2-4c1b-990b-a99fa60b3e73/640/d0fb19e5-05b2-4c1b-990b-a99fa60b3e73.png) \ Sigma-Aldrich \ 900570 \ Ir\[dF(t-Bu)-ppy\]3](https://www.sigmaaldrich.com/US/en/product/aldrich/900570) Quick View [![\[Ir(dFCF3ppy)2-(5,5’-dCF3bpy)\]PF6 ≥95%](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/422/901/e00f3148-fb86-4f94-9e79-21d064c3f327/640/e00f3148-fb86-4f94-9e79-21d064c3f327.png) \ Sigma-Aldrich \ 908568 \ \[Ir(dFCF3ppy)2-(5,5′-dCF3bpy)\]PF6](https://www.sigmaaldrich.com/US/en/product/aldrich/908568) Quick View [![\[Ir(dFOMeppy)2-(5,5′-dCF3bpy)\]PF6 ≥95%](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/401/909/9c746f9e-e2f3-41bc-94d2-6ef5c913fcc5/640/9c746f9e-e2f3-41bc-94d2-6ef5c913fcc5.png) \ Sigma-Aldrich \ 911607 \ \[Ir(dFOMeppy)2-(5,5′-dCF3bpy)\]PF6](https://www.sigmaaldrich.com/US/en/product/aldrich/911607) Quick View [![3DPAFIPN ≥95%](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/982/079/c27119ec-e454-4d20-9299-c722421949cc/640/c27119ec-e454-4d20-9299-c722421949cc.png) \ Sigma-Aldrich \ 908177 \ 3DPAFIPN](https://www.sigmaaldrich.com/US/en/product/aldrich/908177) Quick View [![3DPA2FBN ≥95%](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/409/053/fc1390c3-8253-4554-9954-f46c4831b149/640/fc1390c3-8253-4554-9954-f46c4831b149.png) \ Sigma-Aldrich \ 908185 \ 3DPA2FBN](https://www.sigmaaldrich.com/US/en/product/aldrich/908185) Quick View [![Potassium 5-bromo-1H-indole-1-carbodithioate ≥95%](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/228/701/1a692328-c86e-468d-a3b9-e44b9ae5f4a6/640/1a692328-c86e-468d-a3b9-e44b9ae5f4a6.png) \ Sigma-Aldrich \ 907502 \ Potassium 5-bromo-1*H*-indole-1-carbodithioate](https://www.sigmaaldrich.com/US/en/product/aldrich/907502) Quick View [![2,4,6-Tri(p-tolyl)pyrylium tetrafluoroborate salt ≥95%](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/200/002/98572c1b-e7ed-4aa1-a461-6fe10e91c236/640/98572c1b-e7ed-4aa1-a461-6fe10e91c236.png) \ Sigma-Aldrich \ 900685 \ 2,4,6-Tri(*p*-tolyl)pyrylium tetrafluoroborate salt](https://www.sigmaaldrich.com/US/en/product/aldrich/900685) Quick View [![Phenox O-PC™ A0202 New Iridium, ≥97%](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/structures/366/338/9ef4f784-93b8-4a63-917a-4bcf24c30f4c/640/9ef4f784-93b8-4a63-917a-4bcf24c30f4c.png) \ Sigma-Aldrich \ 901111 \ Phenox O-PC™ A0202](https://www.sigmaaldrich.com/US/en/product/aldrich/901111) Quick View * * * ## Featured Categories [![Three brown bottles of solvents.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/analytical-chromatography/reagentplus-solvent-grade-products.jpg)](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/solvents) [Solvents](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/solvents) Your Solvent Source: Find the right fit with Supelco®, Sigma-Aldrich®, & SAFC® brands, covering analytical, lab, & biopharmaceutical uses. Order online. [Shop Products](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/solvents) [![Graphics of furan, thiophene, and purine.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/building-blocks/Organic-Building-Blocks.jpg)](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/building-blocks/heterocyclic-building-blocks) [Heterocyclic Building Blocks](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/building-blocks/heterocyclic-building-blocks) We offer a comprehensive portfolio of heterocyclic building blocks, one of the largest and most diverse families of molecular fragments used in organic synthesis. [Shop Products](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/building-blocks/heterocyclic-building-blocks) [![Graphic of building blocks.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/applications/chemistry-and-synthesis/building-blocks.jpg)](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/building-blocks/organic-building-blocks) [Organic Building Blocks](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/building-blocks/organic-building-blocks) Find the basic components needed to drive your research forward in our portfolio of organic building blocks. Alkenesm alkanes, alkynes, arenes, allenes & more! [Shop Products](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/building-blocks/organic-building-blocks) [![Photoreactor in organic synthesis lab for photoredox catalysis.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/categories/catalysts/penn-phd-photoreactor-m2.jpg)](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/photocatalysts) [Photocatalysts](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/photocatalysts) Photocatalysis utilizes visible light to activate a chemical reaction. Our extensive portfolio of catalysts and photoreactors enable consistent reactions in photoredox catalysis. [Shop Products](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/photocatalysts) * * * Overview Related Articles & Protocols Support The powerful nature of photocatalysis arises, in part, from the ability to activate readily accessible, simple starting materials via single electron transfer pathways to provide access to reactive open shell species under mild reaction conditions. Upon formation, these distinct open shell species can be engaged in a wide variety of radical trapping/quenching events to ultimately deliver high-value products.  [![Document Search](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/ecommerce/document-search.png)](https://www.sigmaaldrich.com/documents-search) [Looking for More Specific Information?](https://www.sigmaaldrich.com/documents-search) Visit our document search for data sheets, certificates and technical documentation. [Find Documents](https://www.sigmaaldrich.com/documents-search) ## Related Articles - [Visible-light Photoredox Catalysis](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/photocatalysis/visible-light-photoredox-catalysis) The application of radical chemistry towards organic synthesis is well-developed and wide-reaching, though often hampered by a dependence on toxic radical initiators. - [Micro Photochemical Reactor](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/photocatalysis/micro-photochemical-reactor) Photocatalysis in organic synthesis has grown with innovative methods using visible light, contributing to new research. - [Photoredox Iridium Catalyst for SET Cross-Coupling](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/photocatalysis/photoredox-iridium) Effective Csp2- and Csp-hybridized coupling reactions established for catalysis, expanding to multi-step Csp3-coupling. - [Nicewicz Photoredox Catalysts for Anti-Markovnikov Alkene Hydrofunctionalization](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/photocatalysis/nicewicz-photoredox) Explore Nicewicz Photoredox catalysts—photoexcitable acridinium salts for efficient, metal-free anti-Markovnikov hydrofunctionalizations. - [See All (10)](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search) ## Related Protocols - [Step-by-Step Guide for KitAlysis™ High-Throughput Photocatalysis Reaction Screening Kit](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/chemistry-and-synthesis/photocatalysis/kitalysis-photocatalysis-setup) Materials and procedure Step-by-Step Guide for Photocatalysis High-Throughput Reaction Screening Kit ### Find More Articles and Protocols Enter Keywords Search ## How Can We Help In case of any questions, please submit a [customer support request](https://www.sigmaaldrich.com/US/en/support/customer-support) or talk to our customer service team: Email [custserv@sial.com](mailto:custserv@sial.com) or call +1 (800) 244-1173 ## Additional Support - [Chromatogram Search](https://www.sigmaaldrich.com/chromatogram-search) Use the Chromatogram Search to identify unknown compounds in your sample. - [Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps) Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science. - [Customer Support Request](https://www.sigmaaldrich.com/US/en/support/customer-support) Customer support including help with orders, products, accounts, and website technical issues. - [FAQ](https://maestro.my.site.com/knowledgeportal/s/) Explore our Frequently Asked Questions for answers to commonly asked questions about our products and services. * * * __Related Applications__ [Reaction Design & Optimization](https://www.sigmaaldrich.com/US/en/applications/chemistry-and-synthesis/reaction-design-and-optimization) Chemical reaction design and optimization in organic synthesis aims to precisely alter reaction parameters to achieve optimum results in finding pathways to target molecules. [C–H Functionalization](https://www.sigmaaldrich.com/US/en/applications/chemistry-and-synthesis/synthetic-methods/ch-functionalization) C–H functionalization is the reliable and predictable conversion of a C–H into a C–C, C–N, C–O, or C–X bond in a selective and controlled fashion. [Fluorination](https://www.sigmaaldrich.com/US/en/applications/chemistry-and-synthesis/synthetic-methods/fluorination) Fluorination is the introduction of fluorine to a compound via electrophilic fluorination, nucleophilic fluorination, difluoromethylation, trifluoromethylation, or perfluoroalkylation. [Cross-Coupling](https://www.sigmaaldrich.com/US/en/applications/chemistry-and-synthesis/synthetic-methods/cross-coupling) Cross-coupling is a common reaction in organic chemistry for the creation of C-C, C-N, and C-O bonds with the aid of a metal catalyst. * * * ### References 1\. Prier CK, Rankic DA, MacMillan DWC. 2013. Visible Light Photoredox Catalysis with Transition Metal Complexes: Applications in Organic Synthesis. Chem. Rev.. 113(7):5322-5363. [https://doi.org/10.1021/cr300503r](https://doi.org/10.1021/cr300503r) 2\. Fukuzumi S, Ohkubo K. Organic synthetic transformations using organic dyes as photoredox catalysts. Org. Biomol. Chem.. 12(32):6059-6071. [https://doi.org/10.1039/c4ob00843j](https://doi.org/10.1039/c4ob00843j) 3\. Shaw MH, Twilton J, MacMillan DWC. 2016. Photoredox Catalysis in Organic Chemistry. J. Org. Chem.. 81(16):6898-6926. [https://doi.org/10.1021/acs.joc.6b01449](https://doi.org/10.1021/acs.joc.6b01449) 4\. Romero NA, Nicewicz DA. 2016. Organic Photoredox Catalysis. Chem. Rev.. 116(17):10075-10166. [https://doi.org/10.1021/acs.chemrev.6b00057](https://doi.org/10.1021/acs.chemrev.6b00057) 5\. Skubi KL, Blum TR, Yoon TP. 2016. Dual Catalysis Strategies in Photochemical Synthesis. Chem. Rev.. 116(17):10035-10074. [https://doi.org/10.1021/acs.chemrev.6b00018](https://doi.org/10.1021/acs.chemrev.6b00018) 6\. Poplata S, Tröster A, Zou Y, Bach T. 2016. Recent Advances in the Synthesis of Cyclobutanes by Olefin \[2+2] Photocycloaddition Reactions. Chem. Rev.. 116(17):9748-9815. [https://doi.org/10.1021/acs.chemrev.5b00723](https://doi.org/10.1021/acs.chemrev.5b00723) 7\. Kärkäs MD, Porco JA, Stephenson CRJ. 2016. Photochemical Approaches to Complex Chemotypes: Applications in Natural Product Synthesis. Chem. Rev.. 116(17):9683-9747. [https://doi.org/10.1021/acs.chemrev.5b00760](https://doi.org/10.1021/acs.chemrev.5b00760) Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars) Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions) Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices) Social Media [![LinkedIn icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/linkedin.svg)](https://www.linkedin.com/company/merck-life-science) [![X icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/x-icon.svg)](https://x.com/Merck_lifesci) [![Facebook Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/facebook.svg)](https://www.facebook.com/Merck.lifescience) [![Instagram Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/instagram.svg)](https://www.instagram.com/mercklifescience/) [![YouTube Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/youtube.svg)](https://www.youtube.com/@MerckLifeScience/featured) MilliporeSigma __Research. Development. Production.__ We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production. [![Vibrant M string](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/vibrant-m-string-1-rrvcrp.svg)](https://www.sigmaaldrich.com/US/en) [Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco) © 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies. Reproduction of any materials from the site is strictly forbidden without permission. [Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)