[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Cross-Coupling](https://www.sigmaaldrich.com/US/en/applications/chemistry-and-synthesis/synthetic-methods/cross-coupling)Willis Pyridinates for Palladium-Catalyzed Cross-Coupling Reactions
# Willis Pyridinates for Palladium-Catalyzed Cross-Coupling Reactions
## Introduction
The Suzuki-Miyaura cross-coupling reaction is one of the most used transformations in the pharmaceutical industry to form carbon-carbon bonds.1,2 However, limitations to the application of this reaction in discovery chemistry persist, including the notorious low reaction efficiency, poor stability, and difficulty in preparing pyridine-2-boronates and boronic acids for cross-coupling.
__Read more about__
- [Representative Scheme and Examples](https://www.sigmaaldrich.com#representative-scheme-examples)
- [Advantages](https://www.sigmaaldrich.com#advantages)
- [References](https://www.sigmaaldrich.com#references)
[Related Materials](https://www.sigmaaldrich.com#related-materials)
Professor Willis and industrial partners at Pfizer have shown that by replacing boronates with pyridine-2-sulfinates as nucleophilic coupling partners, a palladium-catalyzed desulfinylative cross-coupling process of exceptional scope can be realized.3 Both aryl bromides and aryl chlorides can be employed as coupling partners to access challenging and medicinally relevant linked pyridine-heterocycle building blocks.
## [](https://www.sigmaaldrich.com)Representative Scheme and Examples
## [](https://www.sigmaaldrich.com)Advantages
- Bench-stable, solid reagents
- Efficient nucleophilic cross-coupling partners
- Convenient removal during aqueous work-up
Special thanks to Tim Markovic and [Prof. Michael Willis](https://www.sigmaaldrich.com/US/en/collections/professor-product-portal/willis) for contributing this Technology Spotlight!
## Related Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
### References
1\.
Miyaura N, Yamada K, Suzuki A. 1979. A new stereospecific cross-coupling by the palladium-catalyzed reaction of 1-alkenylboranes with 1-alkenyl or 1-alkynyl halides. Tetrahedron Letters. 20(36):3437-3440. [https://doi.org/10.1016/s0040-4039(01)95429-2](https://doi.org/10.1016/s0040-4039%2801%2995429-2)
2\.
2016\. [https://doi.org/10.1039/9781782622086](https://doi.org/10.1039/9781782622086)
3\.
Markovic T, Rocke BN, Blakemore DC, Mascitti V, Willis MC. Pyridine sulfinates as general nucleophilic coupling partners in palladium-catalyzed cross-coupling reactions with aryl halides. Chem. Sci.. 8(6):4437-4442. [https://doi.org/10.1039/c7sc00675f](https://doi.org/10.1039/c7sc00675f)
__Related Articles__
- [Jørgensen’s Organocatalysts](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/jorgensen-organocatalysts)
- [MCAT- 53™ Catalyst for Ruthenium Formation](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/mcat-53-green-ruthenium-catalyst)
- [Role of High Purity Metal Salts in Chemical Synthesis](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/role-of-high-purity-metal-salts-in-chemical-synthesis)
- [Reductive amination with 2-picoline-borane complex](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/2-picoline-borane)
- [2nd Generation Buchwald Precatalysts](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/2nd-generation-buchwald-precatalysts)
- [MeOBIPHEP](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/MeOBIPHEP)
- [An Introduction to A-Phos, its Applications and Advantages](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/a-phos)
- [Trialkylphosphine for MBH Reactions](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/air-stable-trialkylphosphine)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [C-C Bond Forming Reagents](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/chemical-synthesis/c-c-bond-forming-reagents)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.