[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/US/en) Products Cart0 USEN Products ProductsApplicationsServicesResourcesSupport [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/US/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/US/en)[Belysa® Immunoassay Curve Fitting Software](https://www.sigmaaldrich.com/US/en/services/software-and-digital-platforms/belysa-immunoassay-curve-fitting-software)Monitor Immunoassay Method Reproducibility with Belysa® Software # Monitor Immunoassay Method Reproducibility with Belysa® Software __Section Overview__ - [What Is Belysa® Immunoassay Curve Fitting Software?](https://www.sigmaaldrich.com#software) - [Evaluating a Single Plate](https://www.sigmaaldrich.com#single-plate) - [Over Multiple Plates From a Single Lot](https://www.sigmaaldrich.com#multiple-plates) - [Over Multiple Lots](https://www.sigmaaldrich.com#multiple-lots) - [Summary of Monitoring Method Reproducibility](https://www.sigmaaldrich.com#summary) - [References](https://www.sigmaaldrich.com#references) - [Highlighted Belysa® Publications](https://www.sigmaaldrich.com#publications) Belysa® software offers a user-friendly way to monitor immunoassay method reproducibility for ELISA readers, as well as Luminex® and SMCxPRO® instruments. Immunoassay users are often well aware of the potential for variability to influence the outcome of their experiments. At the individual plate level, between assays, and between lots of assays, there are numerous considerations when evaluating the method that is being applied to a study. An excellent paper by Khan et al. (2015) described the challenges that RUO (Research Use Only) assay users may face, and suggested strategies that can be used to evaluate them1. Belysa® software was designed to help immunoassay users address several key points raised in this, and other, papers. A key challenge for many scientists is the ability to see variability within their method. An individual plate may be easy to evaluate for replicate coefficient of variation percentage (%CV) or parameters such as the expected recovery of the assay. However, as the number of assays performed increases, or even as the volume of data increases with multiplex experiments, it can become repetitive and laborious to perform the appropriate checks. As projects extend to the point where multiple lots of the same kit are required, it is important to be aware of any lot-to-lot differences, which is normally done by comparing standard curves. To this end, we have designed the Belysa® Immunoassay Curve Fitting Software, a tool that allows you to easily monitor your method and ensure it is reproducible over time. Read on to see how this software can be used: 1. At the single assay level 2. Over multiple plates 3. Over multiple lots ## [](https://www.sigmaaldrich.com)What Is Belysa® Immunoassay Curve Fitting Software? [Belysa® Immunoassay Curve Fitting Software](https://www.sigmaaldrich.com/US/en/services/software-and-digital-platforms/belysa-immunoassay-curve-fitting-software) is a user-friendly tool that allows you to evaluate your assay data and compare standard curves. With Belysa__®__ software, bench researchers gain the ability to easily and affordably evaluate method reproducibility and consistency between immunoassays plate to plate and over the course of longitudinal studies. This software allows the user to check and flag technical parameters before comparing curves between runs to ensure the method was consistent over the course of a study. Discover how to gain more insights into your immunoassay methods in the [Belysa® Application Note](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/307/929/belysa-app-note-ms.pdf). ## [](https://www.sigmaaldrich.com)Evaluating a Single Plate The Belysa__®__ software allows the user to quickly evaluate a single plate of an immunoassay in a number of ways. First, the software shows the distribution of the samples against the standard curve and plot the lower limit of quantitation (LLOQ) (__Figure 1__). This will be done for all biomarkers within an acquisition file. From this, we can immediately tell that the assay had a suitable dynamic range for the samples tested. Further, the screen contains an optimization wizard which will select the optimal curve to fit the standard points as provided metrics, such as the curve equation and limit of detection (LOD) of the assay. ![Importing .CSV file allows a user to visualize their curve and the placement of samples, standards, and quality controls upon it. If previously annotated, Belysa® software will apply those values (e.g., Standard Curve value or dilution factor) however there is also the option to define them in a plate map. Once the data is calculated, standard curves may be fitted automatically or manually, and the user may assess parameters that have not met their criteria (e.g., % CV). Optimizations will be immediately reflected in the displayed data and in the curve statistic.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/protein-biology/immunohistochemistry/belysa-immunoassay-curve-fitting-software.jpg "Belysa® screen demonstrating the assay standard curve, samples, QC's, and standards") __Figure 1.__Belysa® Immunoassay Curve Fitting Software demonstrating the samples distributed on the standard curve and the position of the LLOQ (QC = quality control). The user can then move to the “raw data” tab. Within this data; are the details such as the variance between sample replicates or the expected recovery of the standard curve points. In the software, the user can set the Replicate %CV or % Recovery they accept, and flags will be applied against any data point that is outside the criteria (__Figure 2__). By using this tool, a scientist can quickly ensure that the standard curve and samples performed in a fashion that can be reproduced by another scientist. ![Raw data evaluation presents a challenge, especially within a large, multiplexed immunoassay or over the course of numerous plates. Belysa® software employs a flag system to alert the user to areas that have not met their own criteria and the accepted %CV of a sample or standard. A second alert will indicate to the user where the data is below certain automatically calculated parameters such as the Limit of Detection (LOD) or Limit of Quantitation (LLOQ). This allows you to quickly evaluate your data and ensure that only high-quality data is made available to generate your results.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/protein-biology/immunohistochemistry/belysa-single-96-well-immunoassay.jpg "Raw data display with flags directing the user’s attention to specific data points") __Figure 2.__The raw data generated by a single 96-well immunoassay. The Belysa® tool assigns flags according to the lab’s specifications. ## [](https://www.sigmaaldrich.com)Over Multiple Plates From a Single Lot The multi-plate experiment from a single lot is a strategy used for both fresh and archived samples where kits from a single batch have been purchased. The advantage of this approach is that there will be no variability from the assay lot, however, the lab may experience inter-assay variance simply by having multiple users conduct the assays. Differences in technique, such as pipetting or reagent preparation, can affect the Optical Density (OD) of the standard curve. It is important to compare the standard curves of each assay to ensure that the slopes of each plate’s standard curves are similar. If the slopes are similar, the samples should also report similarly. The Belysa® software contains a mathematical parallelism tool that allows for the importation of standard curves. Each curve will have a slope ratio calculated against the first curve added to the tool. A ratio of 1 indicates a set of perfectly parallel curves (__Figure 3__), although often it is not hard to spot a deviation by eye (__Figure 4__). | | | | |-------|-----|-------------------| | Curve | Lot | MCP-1 Parallelism | | P2 | 2 | 0.99 | | P3 | 1 | 0.99 | | P4 | 2 | 1.00 | | P5 | 3 | 1.00 | | P6 | 3 | 0.98 | ![Six kits from three lots of the Conferma™ MCP-1 ELISA were independently run. The standard curves were compared for similarity by assessing their mathematical parallelism as expressed by their slope ratios. Using plate 1 as a reference all of the other curves were imported into the Belysa® Relative Potency experiment. A slope ratio of 1 indicates perfect parallelism, and therefore that the method has been replicated between experimental runs. This tool confirms that both the assay manufacture and laboratory protocol were similar each time.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/protein-biology/immunohistochemistry/six-plates-of-monocyte-chemoattractant-protein-1.jpg "Six MCP-1 standard curves compared for mathematical parallelism") __Figure 3.__Six plates of [monocyte chemoattractant protein-1 (MCP-1) ELISA](https://www.sigmaaldrich.com/product/mm/ezmcp1) from three lots, run by two separate operators, with Plate 1 from Lot 1 used as the reference curve. A slope ratio of 1 indicates that the linear portions of the curve are mathematically parallel. The process to import the tool is extremely simple. Within minutes, a scientist can import all their curves and quickly determine that they or their team have faithfully executed their protocol multiple times. | | | | |-------|-----|------------------| | Curve | Lot | IL-6 Parallelism | | P2 | 1 | 0.866 | | P3 | 2 | 1.035 | | P4 | 3 | 1.001 | | P5 | 2 | 0.996 | | P6 | 3 | 1.001 | ![By importing multiple curves into the relative potency experiment a user can visualize their curves simultaneously, as well as assess their parallelism. If there has been a technical deviation in the executed method it can be easily identified through a change in the calculated slope ratio, the shape of the curve, or parameters such as the LLOQ of the assay (displayed as a dotted line).](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/protein-biology/immunohistochemistry/six-plates-of-interleukin-6-il-6-elisa.jpg "Belysa® software flagging a technical error") __Figure 4.__Six plates of [interleukin-6 (IL-6) ELISA](https://www.sigmaaldrich.com/product/mm/ezil6) from three lots, run by two separate operators, with Plate 1 from Lot 1 used as the reference curve. A slope ratio of 1 indicates that the linear portions of the curve are mathematically parallel. P2 was identified as a user deviation, later confirmed by the user. ## [](https://www.sigmaaldrich.com)Over Multiple Lots Perhaps the most frustrating and expensive source of variation is that between different lots of assays. Once a study enters its second year, the laboratory will likely need to purchase kits that are constructed from different lots of components. At this stage, a method is required to confirm the kits are performing similarly. One preferred strategy is to perform a sample bridge between the new lot and the prior lot to ensure the samples report similarly. The Belysa__®__ Parallelism Tool can aid and enhance this kind of work over a longitudinal study. As each new lot of kits arrives, the curves can be imported from previous batches so the slope ratios can be compared against a reference curve from the original batch to ensure consistent lot performance (__Figure 5__). ![A user executing their method reproducibly with a kit that has met strong manufacturing QC criteria should be able to demonstrate similar curves lot to lot. In this example, a scientist uses three lots of the Conferma™ IL-6 ELISA (Cat. No. EZIL6-98K) on different days before comparing the standard curves in the relative potency tool of Belysa® software. The slope ratios fell with +/- 0.1 indicating excellent similarity and therefore method reproducibility.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/protein-biology/immunohistochemistry/single-plate-from-three-lots-of-il-6-elisa.jpg "Three lots of an IL-6 ELISA evaluated for similarity") __Figure 5.__A single plate from three lots of [IL-6 ELISA](https://www.sigmaaldrich.com/product/mm/ezil6) run by the same scientist. Slope ratios were within the acceptance criteria of 1 +/- 0.1. ## [](https://www.sigmaaldrich.com)Summary of Monitoring Method Reproducibility The Belysa__®__ software is a tool designed by our software and immunoassay experts. Each of the applications was proposed to enhance the capability of immunoassay users to evaluate their method for consistency of application, starting with that of a single plate and working up to the evaluation of multiple kit lots over years of study. With an extremely user-friendly interface, these checks become rapid and easy to apply, ensuring that any method deviations are caught early and therefore may potentially be ameliorated. Discover more information about this software and schedule a demonstration by filling out our online contact form on our Belysa® Landing Page. [Request Information](https://www.sigmaaldrich.com/US/en/services/software-and-digital-platforms/belysa-immunoassay-curve-fitting-software) ### References 1\. Khan MU, Bowsher RR, Cameron M, Devanarayan V, Keller S, King L, Lee J, Morimoto A, Rhyne P, Stephen L, et al. 2015. Recommendations for adaptation and validation of commercial kits for biomarker quantification in drug development. Bioanalysis. 7(2):229-242. [https://doi.org/10.4155/bio.14.274.](https://doi.org/10.4155/bio.14.274.) ## [](https://www.sigmaaldrich.com)Highlighted Belysa® Publications - Carlson KN, Pavan‐Guimaraes J, Verhagen JC, Chlebeck P, Verhoven B, Jennings H, Najmabadi F, Liu Y, Burlingham W, Capitini CM, et al. 2021. IL‐10 and TGF‐β cytokines decrease immune activation during normothermic ex vivo machine perfusion of the rat liver. Liver Transplantation. [https://doi.org/10.1002/lt.26206](https://doi.org/10.1002/lt.26206). - Bender DE, Schaettler MO, Sheehan KC, Johanns TM, Dunn GP. 2021. Cytokine Profiling in Plasma from Patients with Brain Tumors Versus Healthy Individuals using 2 Different Multiplex Immunoassay Platforms. Biomarker Insights. [https://doi.org/10.1177/11772719211006666](https://doi.org/10.1177/11772719211006666). - Abreu D, Stone SI, Pearson TS, Bucelli RC, Simpson AN, Hurst S, Brown CM, Kries K, Onwumere C, Gu H, et al. 2021. A phase 1b/2a clinical trial of dantrolene sodium in patients with Wolfram syndrome. JCI Insight. [https://doi.org/10.1172/jci.insight.145188](https://doi.org/10.1172/jci.insight.145188). - Scheinman SB, Zaldua S, Dada A, Krochmaliuk K, Dye K, Marottoli FM, Thatcher GRJ, Tai LM. 2021. Systemic candesartan treatment modulates behavior, synaptic protein levels, and neuroinflammation in female mice that express human APOE4. Frontiers in Neuroscience. 15. [https://doi.org/10.3389/fnins.2021.628403](https://doi.org/10.3389/fnins.2021.628403). - Steinbach S, Jalili-Firoozinezhad S, Srinivasan S, Melo MB, Middleton S, Konold T, Coad M, Hammond PT, Irvine DJ, Vordermeier M, et al. 2021. Temporal dynamics of intradermal cytokine response to tuberculin in *Mycobacterium bovis* BCG-vaccinated cattle using sampling microneedles. Scientific Reports. 11(1). [https://doi.org/10.1038/s41598-021-86398-6](https://doi.org/10.1038/s41598-021-86398-6). - Zarkoob H, Allué-Guardia A, Chen Y-C, Jung O, Garcia-Vilanova A, Song MJ, Park J-G, Oladunni F, Miller J, Tung Y-T, et al. 2021. Modeling SARS-CoV-2 and influenza infections and antiviral treatments in human lung epithelial tissue equivalents. [https://doi.org/10.1101/2021.05.11.443693](https://doi.org/10.1101/2021.05.11.443693). - Oladunni FS, Park J-G, Pino PA, Gonzalez O, Akhter A, Allué-Guardia A, Olmo-Fontánez A, Gautam S, Garcia-Vilanova A, Ye C, et al. 2020. Lethality of SARS-CoV-2 infection in K18 human angiotensin-converting enzyme 2 transgenic mice. Nature Communications. 11(1). [https://doi.org/10.1038/s41467-020-19891-7](https://doi.org/10.1038/s41467-020-19891-7). ### Did you use Belysa® software in your research? __How to cite Belysa® Immunoassay Curve Fitting Software in your publication:__ - If you are based in the United States or Canada, state MilliporeSigma as the source of your assay. - If you are based anywhere in the world outside of the United States or Canada, state Merck KGaA, Darmstadt, Germany, as the source of your assay. - Include your use of Belysa® Immunoassay Curve Fitting Software and the catalog number (Cat. No. [40-122](https://www.sigmaaldrich.com/US/en/product/mm/40122)). - If you used our SMC® ultrasensitive assays, Conferma™ ELISAs, or MILLIPLEX® multiplex immunoassays, include the full kit name and catalog number. For MILLIPLEX® kits, also list the analytes you used from the kit. - Be sure to include the species of samples you used and how you diluted your samples. __Related Articles__ - [Conferma® ELISAs: Pursuing a “Pharmagrade” Immunoassay](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/elisa/conferma-sandwich-elisas) - [Luminex® Multiplexing Platforms Comparison](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/milliplex/luminex-multiplexing-platforms-comparison) - [Multiplex Detection with the xMAP® INTELLIFLEX® System](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/milliplex/multiplex-detection-with-the-xmap-intelliflex-system) - [SMCxPRO® Immunoassay Platform](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/smc-technologies/smcxpro-immunoassay-platform-for-ultrasensitive-biomarker-detection) - [Immunology Multiplex Assays | MILLIPLEX® Assays](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/milliplex/immunology-multiplex-assays) - [Multiplex Cancer Assays for Cancer Research| MILLIPLEX® Assays](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/milliplex/multiplex-cancer-assays-for-cancer-biomarker-detection) - [Drug Development Research | SMC® Technology](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/smc-technologies/how-ultrasensitive-biomarker-detection-platforms-accelerate-drug) - [Quality in MILLIPLEX® Multiplex Kits](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/milliplex/quality-in-milliplex-multiplex-kits) - [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search) Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register An unknown error has occured.