Purification of Histidine-Tagged Recombinant Proteins Using Ni Sepharose 6 Fast Flow
Extracted from Affinity Chromatography Vol. 2: Tagged Proteins, GE Healthcare, 2016
Ni Sepharose 6 Fast Flow consists of 90 µm beads of highly cross-linked agarose, to which a chelating ligand has been immobilized and subsequently charged with Ni2+ ions. The ligand density of Ni Sepharose 6 Fast Flow ensures high binding capacity, and the chromatography medium shows negligible leakage of Ni2+ ions. The high flow rate property of the Sepharose 6 Fast Flow matrix makes it well-suited for scaling-up but also for gravity-flow purposes. In addition, the medium is compatible with a wide range of additives commonly used in the purification of histidine-tagged proteins. See Appendix 1 (Characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products) for the main characteristics of Ni Sepharose 6 Fast Flow.
Ni Sepharose 6 Fast Flow
Fig 3.5. Ni Sepharose 6 Fast Flow is designed for scaling up purification of histidine-tagged proteins.
Ni Sepharose 6 Fast Flow is supplied preswollen in 20% ethanol, in pack sizes of 5, 25, 100, and 500 ml1, as well as in convenient prepacked formats as described later in this chapter.
1 Larger quantities are available. Contact your local representative for more information.
Column packing
See instructions supplied with the product, or refer to Appendix 6 (Column packing and preparation) for general guidelines for column packing.
Sample preparation
This sample preparation procedure is applicable for all formats containing Ni Sepharose 6 Fast Flow. See Cell lysis earlier in this chapter for a general description.
Adjust the sample to the composition and pH of the binding buffer by adding buffer, NaCl, imidazole, and additives from concentrated stock solutions; by diluting the sample with binding buffer; or by buffer exchange. To prevent the binding of host cell proteins with exposed histidine, it is essential to include imidazole at a low concentration in the sample and binding buffer (see Chapter 4, Optimizing purification of histidine-tagged proteins).
Pass the sample through a 0.22 µm or a 0.45 µm filter and/or centrifuge it immediately before sample application. Filtration is not necessary when using HisTrap FF crude, His GraviTrap, or His MultiTrap FF. If the sample is too viscous, dilute it with binding buffer to prevent it from clogging; increase lysis treatment (sonication, homogenization); or add DNase/RNase to reduce the size of nucleic acid fragments.
Buffer preparation
Binding buffer: 20 mM sodium phosphate, 0.5 M NaCl, 20 to 40 mM imidazole, pH 7.4. (The optimal imidazole concentration is protein dependent; 20 to 40 mM is suitable for many proteins.)
Elution buffer: 20 mM sodium phosphate, 0.5 M NaCl, 500 mM imidazole, pH 7.4.
Water and chemicals used for buffer preparation should be of high purity. Filter buffers through a 0.45 µm filter before use. Use high-purity imidazole, as this will give a very low or no absorbance at 280 nm.
The optimal concentration of imidazole needed in the sample and buffer to obtain the best purity and yield differs from protein to protein. In the binding buffer, 20 to 40 mM imidazole is suitable for many proteins; 500 mM imidazole in the elution buffer ensures complete elution of the target protein.
As an alternative to elution with imidazole, lower the pH to approximately pH 4.5. (Metal ions will be stripped off the medium below pH 4.0.)
Purification
  1. If the column contains 20% ethanol, wash it with 5 column volumes of distilled water. Use a flow velocity of 50 to 100 cm/h.
  2. Equilibrate the column with 5 to 10 column volumes of binding buffer at a flow velocity of 150 cm/h.
  3. Apply the pretreated sample.
  4. Wash with binding buffer until the absorbance reaches the baseline.
  5. Elute with elution buffer using a step or linear gradient.
    1. For step elution, 5 column volumes of elution buffer are usually sufficient.
    2. For linear gradient elution, a shallow gradient, over 20 column volumes, may separate proteins with similar binding strengths.
  6. After elution, regenerate the column by washing it with 5 to 10 column volumes of binding buffer. The column is now ready for a new purification.
The column does not need to be stripped and recharged between each purification if the same protein is going to be purified. Reuse of any purification column depends on the nature of the sample and should only be performed with identical tagged proteins to prevent cross-contamination. For more information on this topic and on cleaning and storage, refer to Appendix 1 (Characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products).
Use the elution buffer as blank when measuring absorbance manually. If imidazole needs to be removed from the protein, use a desalting column (see Chapter 11, Desalting/buffer exchange and concentration).  Low-quality imidazole will give a significant background absorbance at 280 nm.
Ni Sepharose 6 Fast Flow is compatible with reducing agents. However, we recommend removal of any weakly bound Ni2+ ions before applying buffer/sample that includes reducing agents. This can be accomplished by performing a blank run without reducing agents (see below). Do not store Ni Sepharose 6 Fast Flow with buffers that include reducing agents.
Leakage of Ni2+ from Ni Sepharose 6 Fast Flow is low under all normal conditions. For very critical applications, leakage during purification can be even further diminished by performing a blank run (as described below) before loading sample.
Blank run:
  1. Use binding buffer and elution buffer without reducing agents.
  2. Wash the column with 5 column volumes of distilled water (to remove the 20% ethanol).
  3. Wash with 5 column volumes of elution buffer.
  4. Equilibrate with 10 column volumes of binding buffer.