Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Alkaline Phosphatase, Calf Intestinal (CIP)
(CIP)
From Calf Intestine
(EC 3.1.3.1)
Product No. P4978
Store at –20 °C
Product Description
CIP is commonly used to remove 5’-phosphate groups from DNA, RNA and both ribo and deoxy-ribonucleoside triphosphates. Removal of 5’-phosphates is very useful in preventing self-ligation of cleaved DNA vectors. This property greatly reduces background (plasmids without insert) in cloning procedures.
Product Summary
Activity: approx. 10,000 units/ml (see label for lot specific activity)
DNase: None detected
Nickase: None detected
RNase: None detected
Unit Definition: One unit will hydrolyze 1.0 µmole of 4-nitrophenyl phosphate per minute at pH 9.8 at 37 °C (diethanolamine).
10X CIP Buffer (C 3225)
1 M NaCl
0.5 M Tris-HCl pH 7.9 at 25 °C
0.1 M MgCl2
0.01 M dithiothreitol
Storage buffer
10 mM Tris-HCl, pH 8.2
50 mM KCl
1 mM MgCl2
0.1 mM ZnCl2
50 % glycerol
Procedure: Dephosphorylation of DNA
  1. Dissolve DNA in 1X CIP Buffer (0.5µg DNA/10 µl).
  2. For 5’ overhang DNA add 0.1 units/pmol CIP; for 3’ overhang or blunt end DNA add 1 unit/pmol.
  3. Incubate 60 minutes at 37 °C.
  4. Extract with phenol/chloroform2 (P3803 or P2069) or gel purify the DNA.*
  5. Recover the DNA by alcohol precipitation.2
    *Note: Phenol extraction or gel purification makes heat inactivation unnecessary.
Heat Inactivation
Greater than 95% of the activity can be inactivated by heating to 75 °C for 10 minutes in the presence of 5 mM EDTA.
Materials
Reference
  1. Moessner,E. et al., Z. Physiol. Chem. 361, 543.
  2. Sambrook, J. et al., Molecular Cloning: A Laboratory Manual, Cold Spring Harbor Laboratory (1989), p 5.72, 6.22-6.47 and E.3-E.13.
10/01
Related Links