Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Enzymatic Method for Determining Asparaginase Activity (Asparaginase Activity Assay)
This assay protocol is suitable for the colorimetric/fluorometric detection of Asparaginase activity in bacteria, cell and tissue culture supernatants, plant samples, biological samples, and food samples using the Asparaginase Activity Assay Kit (MAK007). Asparaginase activity is determined by a coupled enzyme assay, which results in a colorimetric (570 nm)/fluorometric (λex = 535/λem = 587 nm) product, proportional to the aspartate generated. One unit of Asparaginase is defined as the amount of enzyme that catalyzes the formation of 1.0 μmole of aspartate per minute at 25 °C.
Precautions and Disclaimer
This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices. 
Reagents
Asparaginase Assay Buffer                                 25 mL
    Catalog Number MAK007A
Fluorescent Peroxidase Substrate, in DMSO       0.2 mL
    Catalog Number MAK007B
Substrate Mix                                                       1 vl
    Catalog Number MAK007C
Aspartate Enzyme Mix                                          1 vl
    Catalog Number MAK007D
Conversion Mix                                                     1 vl
    Catalog Number MAK007E
Asparaginase Assay Positive Control                     1 vl
    Catalog Number MAK007F
Aspartate Standard, 100 mM                             0.1 mL
    Catalog Number MAK007G
Reagents and Equipment Required but Not Provided
96 well flat-bottom plate – It is recommended to use black plates with clear bottoms (Catalog Number M5686 or equivalent) for fluorescence assays and clear plates (Catalog Number M4436 or equivalent) for colorimetric assays.
Fluorescence or spectrophotometric multiwell plate reader
Preparation Instructions
Briefly centrifuge vials before opening. Use ultrapure water for the preparation of reagents. To maintain reagent integrity, avoid repeated freeze/thaw cycles.
Asparaginase Assay Buffer – Allow buffer to come to room temperature before use.
Fluorescent Peroxidase Substrate – Allow reagent to come to room temperature before use. Mix well by pipetting, then aliquot and store, protected from light and moisture, at –20 °C. Upon thawing, the Fluorescent Peroxidase Substrate is ready-to-use in the colorimetric assay.
For the fluorescence assay, dilute an aliquot of the Fluorescent Peroxidase Substrate 5 to 10-fold with Asparaginase Assay Buffer, just prior to use. This will reduce the background of the fluorescence assay.
Substrate Mix – Reconstitute in 0.5 mL of water. Mix well by pipetting, then aliquot and store at –20 °C. Use within two months of reconstitution.
Aspartate Enzyme Mix and Conversion Mix – Reconstitute each in 220 μL of Asparaginase Assay Buffer. Mix well by pipetting, then aliquot and store at –20 °C. Use within two months of reconstitution.
Asparaginase Assay Positive Control – Reconstitute in 100 μL of Asparaginase Assay Buffer. Mix well by pipetting, then aliquot and store at –20 °C. Use within two months of reconstitution.
Storage/Stability
The kit is shipped on wet ice. Storage at –20 °C, protected from light, is recommended.
Procedure
All samples and standards should be run in duplicate.
Aspartate Standards for Colorimetric Detection
Dilute 10 μL of the 100 mM (100 nmole/μL) Aspartate Standard Solution with 990 μL of Asparaginase Assay Buffer to prepare a 1 mM (1 nmole/μL) standard solution. Add 0, 2, 4, 6, 8, and 10 μL of the 1 mM Aspartate standard solution into a 96 well plate, generating 0 (blank), 2, 4, 6, 8, and 10 nmole/well standards. Add Asparaginase Assay Buffer to each well to bring the volume to 50 μL.
Aspartate Standards for Fluorometric Detection
Dilute 10 μL of the 100 mM (100 nmole/μL) Aspartate Standard Solution with 990 μL of Asparaginase Assay Buffer to prepare a 1 mM (1 nmole/μL) standard solution. Dilute 10 μL of the 1 mM standard solution with 90 μL of Asparaginase Assay Buffer to generate a 0.1 mM (0.1 nmole/μL) standard solution. Add 0, 2, 4, 6, 8, and 10 μL of the 0.1 mM Asparate standard solution into a 96 well plate, generating 0 (blank), 0.2, 0.4, 0.6, 0.8, and 1.0 nmole/well standards. Add Asparaginase Assay Buffer to each well to bring the volume to 50 μL.
Sample Preparation
Both the colorimetric and fluorometric assays require 50 μL of sample for each reaction (well).
Tissue or cells should be rapidly homogenized with 4 volumes of Asparaginase Assay Buffer. Centrifuge at 15,000 x g for 10 minutes to remove insoluble materials. Bring samples to a final volume of 50 μL with Asparaginase Assay Buffer.
Serum samples can be directly added to wells. Add 1–50 μL samples into wells of a 96 well plate. Bring samples to final volume of 50 μL with Asparaginase Assay Buffer.
For unknown samples, it is suggested to test several sample volumes to make sure the readings are within the standard curve range.
For the positive control (optional), add 5 μL of the Asparaginase Assay Positive Control to wells. Adjust well volume to 50 μL with water.
Aspartate, oxaloacetate, and pyruvate in the samples will generate a background signal. To remove the effect of background, a sample blank may be set up by omitting the Aspartate Enzyme Mix. The blank readings can then be subtracted from the sample readings.
Assay Reaction
1. Set up the Reaction Mixes according to the scheme in Table 1. Prepare enough Reaction Mixes for the number of samples, positive controls, and standards to be performed.
Table 1.
Reaction Mixes
Reagent Sample Blank Samples and Standards
Asparaginase Assay Buffer 40 μl 42 μl
Substrate Mix 4 μl 4 μl
Aspartate Enzyme Mix 2 μl
Conversion Mix 2 μl 2 μl
Fluorescent Peroxidase Substrate 2 μl 2 μl
2. Add 50 μL of the appropriate Reaction Mix to each well. Mix well using a horizontal shaker or by pipetting.
3. After 2–3 minutes, take the initial measurement (Tinitial). For colorimetric assays, measure the absorbance at 570 nm (A570)initial. For fluorometric assays, measure fluorescence intensity (FLUinitial,
λex = 535/λem = 587 nm).
4. Incubate the plate at 25 °C taking measurements every 5 minutes. Protect the plate from light during the incubation.
5. Continue taking measurements until the value of the most active sample is greater than the value of the highest standard. At this time the most active sample is near or exceeds the end of the linear range of the standard curve.
6. The final measurement for calculating the enzyme activity would be the penultimate reading or the value before the most active sample is near or exceeds the end of the linear range of the standard curve. The time of the penultimate reading is Tfinal.
7. Calculate the change in measurement from Tinitial to Tfinal.
ΔA570 = (A570)final – (A570)initial
or
ΔFLU = (FLUfinal) – (FLUinitial)
Note: It is essential the initial and final measurements fall within the linear range of the reaction.
Results
Calculations
Correct for the background by subtracting the value obtained for the 0 (blank) standard from all readings. Plot the aspartate standard curve.
Note: A new standard curve must be set up each time the assay is run.
Compare the Δmeasurement value (ΔA570 or ΔFLU) of each sample to the standard curve to determine the amount of aspartate generated (B) between Tinitial and Tfinal.
The Asparaginase activity of a sample may be determined by the following equation:
Asparaginase Activity = B x Sample Dilution Factor
                                    (Tfinal – Tinitial) x V
B = Amount (nmole) of aspartate generated between Tinitial and Tfinal
Tinitial = Time of first reading in minutes.
Tfinal = Time of second reading in minutes.
V = sample volume (mL) added to well.
Asparaginase activity is reported as nmole/min/mL = milliunit/mL, where one unit of Asparaginase is defined as the amount of enzyme that catalyzes the formation of 1.0 mmole of aspartate per minute at 25 °C.
MAK007-Colorimetric-Assay-Kit
MAK007-Fluorometric-Assay-Kit