Indirect Co-culture of Embryonic Stem Cells with Embryonic Fibroblasts
Indirect Co-culture of Embryonic Stem Cells with Embryonic Fibroblasts
A. Day 1
  1. Coat T-75 flasks with 10 mL of 0.1% gelatin in DPBS and incubate for at least 30 minutes at 37°C.
  2. Thaw PMEF vial (s) quickly in a 37°C water bath, transfer to 15 mL tube already containing 10 mL warm medium. Gently invert tube, and pellet cells at 4°C, ~1000 rpm for approximately 4–5 minutes.
  3. Remove supernatant, resuspend cells, remove gelatin from plates/flasks, and aliquote feeder cell suspension per densities recommended below.
  4. Remove excess gelatin from flask prior to seeding.
  5. Seed flask with MEF feeder cell suspension: approximately 1.5 × 105 cells per mL MEF should result in 95% confluence within 24 hours.
  6. Incubate at 37°C overnight.
B. Day 2
  1. Coat T-75 flasks with 10 mL of 0.1% gelatin in DPBS and incubate for at least 30 minutes at 37°C.
  2. Thaw PMEF vial (s) quickly in a 37°C water bath, transfer to 15 mL tube already containing 10 mL warm medium. Gently invert tube, and pellet cells at 4°C, ~1000 rpm for approximately 4–5 minutes.
  3. Remove supernatant, resuspend cells, remove gelatin from plates/flasks, and aliquote feeder cell suspension per densities recommended below.
  4. Remove excess gelatin from flask prior to seeding.
  5. Seed flask with MEF feeder cell suspension: approximately 1.5 × 105 cells per mL MEF should result in 95% confluence within 24 hours.
  6. Incubate at 37°C overnight.
C. Day 3
  1. Feed ESC on MEF feeder layer with fresh ESC media.
D. Day 4-8
  1. Feed ESC on MEF feeder layer with fresh ESC media or pass cells, at a 1:2 ratio, if required. (After thawing ESC, 2–3 passages are preferred before seeding onto a Millicell®-24 or Millicell® 96-well Cell Culture Insert Plate. Both cell types are lifted at once and passed on to a new T-75 containing inactivated MEF.) ESC colonies grown on Millicell®-96 Cell Culture Insert Plate 1.0 PET membrane, stained for alkaline phosphatase activity, after culturing via indirect co-culture with ESC in apical well, at a 200 cell per well seeding density, and MEF in the single-well feeder tray.
E. Day 9
  1. Feed ESC on MEF feeder layer with fresh ESC media.
  2. Coat Millicell®-24 or Millicell®-96 single-well feeder tray with approximately 5–10 mL of 25 µg/mL fibronectin in DPBS and incubate for 45 minutes at room temperature.
  3. Remove excess fibronectin from single-well tray.
  4. Thaw MEF using protocol from Day 1, section A.
  5. Seed fibronectin coated single-well tray with MEF cell suspension: approximately 1.67 × 106 MEF cells per single-well tray will result in 95% confluence within 24 hours.
  6. Cover with lid and incubate single-well tray at 37°C overnight.
F. Day 10
  1. Lift ESC and MEF feeder cells from T-75 flasks:
  2. Wash flasks 2X with 10 mL of pre-warmed DPBS (incubate 1–2 minutes per wash).
  3. Remove DPBS and add 3 mL TrypLE and incubate at room temp for 2–3 minutes.
  4. Monitor detachment of cells with an inverted microscope and add ESC media to inactivate TrypLE.
  5. Mix well and wash flask wall to remove all cells from flask.
  6. Separate ESC from MEF feeder cells:
  7. Transfer ESC/MEF cell suspension to a new T-75 flask and incubate at 37°C for 45 minutes.
  8. Remove non-adherent cells (ESC) and transfer to another new T-75 flask and incubate at 37°C for 45 minutes.
  9. Remove non-adherent cells (ESC) again and seed cell culture filter plate wells with ESC suspension.
  10. Seed apical wells of the cell culture plates with ESC. Seed approximately 200–500 cells per well in 100 µL ESC media for Millicell®-96 plates and approximately 1000–1500 cells per well in 400 µL ESC media for Millicell®-24 plates.
  11. Remove media from the MEF seeded single-well trays and replace with approximately 28 32 mL ESC media.
  12. Combine ESC seeded cell culture filter plates to MEF seeded single-well trays.
  13. Incubate assembly at 37°C overnight.
G. Day 12 and Day 14
  1. Feed ESC and MEF indirect co-culture with ESC media.
H. Day 16
  1. Analyze alkaline phosphatase activity to demonstrate that ESC is undifferentiated with an alkaline phosphatase detection kit.
Note: This protocol is designed to grow undifferentiated embryonic stem cells in an indirect co culture with the fibroblast feeder layer. Although it is targeted for use with Millicell®-24 and Millicell®-96 plates, this protocol can be used with Millicell® single-well inserts as well.
Materials and Reagents
  • Millicell®-24 Cell Culture Insert Plates — Millipore cat. nos. PSHT010R5, PSRP010R5
  • Millicell®-96 Cell Culture Insert Plates — Millipore cat. no. PSRP004R5
  • Primary mouse embryo fibroblasts (DMEF) — Millipore cat. no. PMEF-CFL
  • 129/S6 Murine embryonic stem cells (ESC) — Millipore cat. no. SCR012
ESC Media:
  • Knock out DMEM — Millipore cat. no. SLM-220-B
  • 20% ES qualified Serum — Millipore cat. no. ES-009-B
  • 1% Glutamax-1
  • 1% PenStrep — Millipore cat. no. TMS-ABZ-C
  • 1% Non Essential Amino Acids — Millipore cat. no. TMS-001-C
  • 0.1% ESGRO® (LIF) — Millipore cat. no. ESG1106
  • 0.1% 2-mercaptoethanol — Millipore cat. no. ES-007-E
MEF Media:
  • DMEM — Millipore cat. no. SLM-022-B
  • 10% Fetal Bovine Serum — Millipore cat. no. ES-009-B
  • 1% Glutamax-1
  • 1% PenStrep — Millipore cat. no. TMS-ABZ-C
  • 1% Non Essential Amino Acids — Millipore cat. no. TMS-001-C
  • Gelatin 2% Solution — Millipore cat. no. SF008
  • Mitomycin C powder — Sigma cat. no. M4287
  • DPBS — Millipore cat. no. BSS-1005-B
  • TrypLE™ Select (1X), liquid — Invitrogen cat. no. 12563
  • Tissue culture flasks and tubes
  • Fibronectin Solution, 1mg/mL — Millipore cat. no. FC010
Other:
  • Alkaline phosphatase detection kit — Millipore cat. no. SCR004
Materials