Experienced User Protocol
GenElute™ Blood Genomic DNA Kit (NA2010, NA2020)
For experienced users, an abbreviated protocol is listed below.  For data, images, troubleshooting and other information, please see the full length protocol page.
1. Release DNA
· Enzymatic digestion.
2. Prepare Column
· Add solution and spin.
3. Bind DNA
· Add ethanol and spin.
4. Wash Column
· Wash and spin twice.
5. Elute DNA
· Spin 1 minute.
Storage and Stability
Store the kit at room temperature. If any kit reagent forms a precipitate, warm at 55–65 °C until the precipitate dissolves and allow to cool to room temperature before use.
Precautions and Disclaimer
The GenElute Blood Genomic DNA Kit is for laboratory use only, not for drug, household, or other uses.
The Lysis Solution C contains a chaotropic salt, which is an irritant. The Column Preparation Solution is an irritant. Avoid contact with skin. Wear gloves, safety glasses, and suitable protective clothing when handling these solutions or any reagent provided with the kit. Please consult the Safety Data Sheet (SDS) for information regarding hazards and safe handling practices.
Related Links