Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Enzymatic Method for Determining Glucose-6-Phosphate (Glucose-6-Phosphate Assay)
Introduction
This assay protocol is suitable for the colorimetric detection of Glucose-6-Phosphate (G6P) in cell and tissue culture supernatants, urine, plasma, serum, and other biological samples using the Glucose-6-Phosphate Assay Kit (MAK014).  G6P concentration is determined by a coupled enzyme assay, which results in a colorimetric (450 nm) product, proportional to the G6P present. This kit has a linear range of detection between 0.2–1.0 nmole.
Reagents
Glucose-6-Phosphate Assay Buffer                    25 mL
    Catalog Number MAK014A
Glucose-6-Phosphate Enzyme Mix                    1 vL
    Catalog Number MAK014B
Glucose-6-Phosphate Substrate Mix                  1 vL
    Catalog Number MAK014C
Glucose-6-Phosphate Standard 10 µmole            1 vL
    Catalog Number MAK014D
Reagents and Equipment Required but Not Provided
96 well flat-bottom plate – It is recommended to use clear plates (Catalog Number M4436 or equivalent) for colorimetric assays.
Spectrophotometric multiwell plate reader
10 kDa Molecular Weight Cut-Off (MWCO) Spin Filter (Catalog Number Z706345 or equivalent)
Preparation Instructions
Briefly centrifuge vials before opening. Use ultrapure water for the preparation of reagents. To maintain reagent integrity, avoid repeated freeze/thaw cycles.
Glucose-6-Phosphate Assay Buffer – Allow buffer to come to room temperature before use.
Glucose-6-Phosphate Enzyme Mix – Reconstitute in 220 µL of water. Mix well by pipetting, then aliquot and store at –20 °C. Use within 2 months of reconstitution.
Glucose-6-Phosphate Substrate Mix – Reconstitute in 220 µL of Glucose-6-Phosphate Assay Buffer. Mix well by pipetting, then aliquot and store at 4 °C. Use within 2 months of reconstitution.
Glucose-6-Phosphate Standard – Reconstitute in 100 µL of water to generate a 100 mM Standard Solution. Mix well by pipetting, then aliquot and store at –20 °C. Use within 2 months of reconstitution. 
Storage / Stability
The kit is shipped on wet ice. Storage at –20 °C, protected from light, is recommended.
Procedure
All samples and standards should be run in duplicate.
Glucose-6-Phosphate Standards for Colorimetric Detection
Dilute 10 µL of the 100 mM G6P Standard Solution with 990 µL of water to generate a 1 mM standard solution. Add 0, 2, 4, 6, 8, 10 µL of the 1 mM standard solution into a 96 well plate, generating 0 (blank), 2, 4, 6, 8, and 10 nmole/well standards. Add Glucose-6-Phosphate Assay Buffer to each well to bring the volume to 50 µL.
Sample Preparation
Liquid or solution samples may be assayed directly. If enzymes are present in the samples, samples should be deproteinized with a 10 kDa MWCO spin filter.
Tissue (10–100 mg) or cells (1–5 x 106) can be homogenized in 2–3 volumes of ice cold PBS or other buffer (pH 6.5–8). Centrifuge the samples at 13,000 x g for 10 minutes to remove insoluble material. Samples should be deproteinized with a 10 kDa MWCO spin filter.
Add 1–50 µL samples into duplicate wells of a 96 well plate. Bring samples to a final volume of 50 µL with Glucose-6-Phosphate Assay Buffer.
For unknown samples, it is suggested to test several sample volumes to make sure the readings are within the standard curve range.
Assay Reaction
  1. Set up the appropriate Reaction Mixes according to the scheme in Table 1. 50 µL of the Master Reaction Mix is required for each reaction (well).
    Note: NADH or NADPH in samples will generate background readings. To remove the effect of NADH or NADPH background, a blank sample may be set up for each sample by omitting the Glucose-6-Phosphate Enzyme Mix. The blank readings can then be subtracted from the sample readings.
    Table 1.
    Reaction Mixes
Reagent
Blank Sample
Samples & Standards
G6P Assay Buffer
46 µL 46 µL
G6P Enzyme Mix
-
2 µL
G6P Substrate Mix
2 µL
2 µL
  1. Add 50 µL of the appropriate Reaction Mix to each of the wells. Mix well using a horizontal shaker or by pipetting, and incubate the reaction for 30 minutes at room temperature. Protect the plate from light during the incubation.
  2. Measure the absorbance at 450 nm (A450).
Results
Calculations
The background for the assay is the value obtained for the 0 (blank) G6P standard. Correct for the background by subtracting the blank value from all readings. Background values can be significant and must be subtracted from all readings.
Use the values obtained from the appropriate G6P standards to plot a standard curve. The amount of G6P present in the samples may be determined from the standard curve.
Note: A new standard curve must be set up each time the assay is run.
Concentration of G6P
        Sa/Sv = C
Sa = Amount of G6P in unknown sample (nmole) from standard curve
Sv = Sample volume (µL) added into the wells
C = Concentration of G6P in sample
Sample Calculation
G6P Amount (Sa) = 5.84 nmole
Assay volume (Sv) = 50 µL
Concentration of G6P in sample
5.84 nmole/50 µL = 0.1169 nmole/µL
0.1169 nmole/µL x 260.14 ng/nmole=30.41 ng/µL
Materials
Precautions and Disclaimer
This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices. 
Related Links