A Recommended Standard Method for Isolating Mononuclear Cells
Extracted from Isolation of Mononuclear Cells, Methodology and applications (PDF)
GE Healthcare, 2010
Cell separation using Ficoll-Paque products can be carried out over a wide range of blood sample volumes. With its high yield, this method can be adapted to the processing of very small amounts of blood, such as may be obtained from children. For maximum reproducibility of separation it is recommended that a standardized procedure be used. The following procedure has been evaluated in our laboratories with Ficoll-Paque PLUS and is recommended for separation of normal blood samples. Simple changes can easily be made to suit a particular centrifugation system. The same procedure is recommended when separating cells using Ficoll-Paque PREMIUM, Ficoll-Paque PREMIUM 1.084, and Ficoll-Paque PREMIUM 1.073.
To standardize the technique, blood volume and diameter of the centrifuge tube should be chosen first. These factors determine the height of the blood sample in the tube and consequently the centrifugation time. Increasing the height of the blood sample in the tube increases red cell contamination. The separation is, however, not appreciably affected by changing the diameter of the tube. Hence a larger volume can be separated with the same degree of purification in a tube of larger diameter if the height of the blood sample in the tube and the separation time are kept constant.
The yield and degree of purity of the mononuclear cells depend to a considerable extent on the efficiency of red cell removal.
When erythrocytes in whole blood are aggregated, some mononuclear cells are trapped in the clumps and therefore sediment with the erythrocytes. This tendency to trap mononuclear cells is reduced by diluting the blood. Dilution gives a better yield of mononuclear cells and reduces the size of the red cell clumps. Aggregation of erythrocytes is enhanced at higher temperatures (37°C), which consequently decreases the yield of mononuclear cells. At lower temperatures (4°C); however, the rate of aggregation is decreased but the time of separation is increased, which also decreases the yield of mononuclear cells. A compromise temperature of 18ºC to 20°C gives optimal results.
Equipment and solutions required but not provided
  1. Sterile balanced salt solution or other standard salt solutions (see Preparation of reagents).
  2. Centrifuge with swing-out rotor (brake should be off).
  3. Sterile centrifuge tubes and pipettes.
  4. Sterile needles and syringes.
  5. Red blood cell lysis solution of choice (if isolating granulocytes).
Preparation of reagents
Diluent and washing solution
The balanced salt solution for dilution of the blood and cell washing can be made according to the instruction below. Other diluents and washing fluids such as isotonic Ca2+/Mg2+ free phosphate buffered saline (e.g. Dulbecco’s PBS), salt solutions (e.g., Hank’s) or cell culture media (e.g., RPMI 1640) may also be used.
Balanced salt solution
To prepare the balanced salt solution, mix one volume stock solution A with nine volumes stock solution B and sterilize. At least 20 ml for each sample should be processed. Other sterile standard salt solutions may also be used.
Solution A   Concentration (g/L)
Anhydrous d-glucose 5.5 × 10-3 M (0.1%) 1.0
CaCl2.2H2O 5.0 × 10-5 M 0.0074
MgCl2.6H20 9.8 × 10-4 M 0.1992
KCl 5.4 × 10-3 M 0.4026
TRIS 0.145 M 17.565
Dissolve in approximately 950 ml of distilled water and add concentrated HCl until the pH is 7.6 before adjusting the volume to 1L.
Solution B   Concentration (g/L)
NaCl 0.14 M 8.19
To prepare the balanced salt solution, mix 1 volume of solution A with 9 volumes of solution B. Prepare the solution freshly each week. Other standard salt solutions may be used.
Ficoll-Paque product
Warm the Ficoll-Paque density gradient media to 18°C to 20°C before use. For samples larger than 3 ml, see Notes.
Preparation of the sample
Fresh blood should be used to ensure high viability of isolated mononuclear cells. Prepare the sample at 18ºC to 20°C.
  1. To a 10 ml centrifuge tube add 2 ml of defibrinated- or anticoagulant-treated blood and an equal volume of balanced salt solution (final volume 4 ml).
  2. Mix the blood and buffer by inverting the tube several times or by drawing the mixture in and out of a pipette.
Procedure for isolation of mononuclear cells
  1. Invert the Ficoll-Paque media bottle several times to ensure thorough mixing.
    For withdrawal of Ficoll-Paque media by syringe: Snap-off the polypropylene cap and insert the syringe needle through the septum (Figure 1).
    For withdrawal of Ficoll-Paque media by pipette: Remove the snap-off polypropylene cap. Lift the aluminum ring. Pull off the metal seal. Remove the silver ring.
    Remove the rubber closure. Using aseptic techniques, withdraw the required volume of Ficoll-Paque media (Figure 2).
  2. Add Ficoll-Paque media (3 mL) to the centrifuge tube.
  3. Carefully layer the diluted blood sample (4 mL) onto the Ficoll-Paque media solution (Figure 3).
    When layering the sample do not mix the Ficoll-Paque media solution and the diluted blood sample.
  4. Centrifuge at 400 g for 30 to 40 min at 18ºC to 20°C (brake should be turned off).
  5. Draw off the upper layer containing plasma and platelets using a sterile pipette, leaving the mononuclear cell layer undisturbed at the interface (Figure 4 and Figure 5). The upper layer, which contains the plasma, may be saved for later use.
  6. Transfer the layer of mononuclear cells to a sterile centrifuge tube using a sterile pipette.
Polypropylene cap and silver ring removal
Figure 1. Polypropylene cap and silver ring removal
Withdrawal of Ficoll-Paque media
Figure 2. Withdrawal of Ficoll-Paque media
Blood sample layered onto Ficoll-Paque media
Figure 3. Blood sample layered onto Ficoll-Paque media
Prior to removal of the upper layer.
Figure 4. Prior to removal of the
upper layer.
After the upper layer (Plasma) removed.
Figure 5. After the upper layer (Plasma) removed.
Washing the cell isolate
  1. Estimate the volume of the transferred mononuclear cells. Add at least 3 volumes (~ 6 mL) of balanced salt solution to the mononuclear cells in the centrifuge tube.
  2. Suspend the cells by gently drawing them in and out of a pipette.
  3. Centrifuge at 400 to 500 × g for 10 to 15 min at 18°C to 20°C.
Note: A centrifugation at high speed increases the mononuclear cell recovery. However, if it is important to also get rid of platelets a lower centrifugation speed is recommended (60 to 100 × g).
  1. Remove the supernatant.
  2. Resuspend the mononuclear cells in 6 to 8 ml balanced salt solution.
  3. Centrifuge at 400 to 500 × g (or 60 to 100 × g for removal of platelets) for 10 min at 18°C to 20°C.
  4. Remove the supernatant.
  5. Resuspend the cell pellet in media appropriate for the application.
Procedure for isolation of granulocytes
  1. Perform density gradient centrifugation using Ficoll-Paque media as described above in Procedure for isolation of mononuclear cells, steps 1 to 6.
  2. Draw off the upper layer of Ficoll-Paque media using a sterile pipette, leaving the white cell layer of granulocytes above the red blood cell layer undisturbed.
  3. Collect the thin white cell layer of granulocytes with a pipette and transfer to a sterile centrifuge tube.
  4. Resuspend the cells in at least five volumes of balanced salt solution and centrifuge at 400 × g for 15 min.
  5. Lyse remaining red blood cells with any red blood cell lysis solution of choice.
  6. Centrifuge the granulocytes at 400 to 500 × g for 10 to 15 min at 18°C to 20°C.
  7. Remove the supernatant.
  8. Resuspend the granulocytes in 6 to 8 ml balanced salt solution.
  9. Centrifuge at 400 to 500 × g for 10 min at 18°C to 20°C.
  10. Remove the supernatant.
  11. Resuspend the cell pellet in media appropriate for the application.
Heparin, EDTA, citrate, acid citrate dextrose (ACD), and citrate phosphate dextrose (CPD) may be used as anticoagulants for the blood sample. Defibrinated blood requires no anticoagulant. Defibrination, however, results in a lower mononuclear cell yield and may cause increased contamination by red cells3. It also causes selective loss of monocytes.
Bøyum has found that a slightly purer mononuclear cell preparation is obtained using EDTA instead of heparin as anticoagulant3. It has also been noted in the purification of mononuclear cells from sources other than peripheral blood that addition of heparin may cause gelling of cell suspensions4. Citrate-stabilized blood may result in better quality RNA and DNA than other anticoagulants, and produce a higher yield of mononuclear cells. Heparin affects T-cell proliferation and binds to many proteins. EDTA is good for DNA based assays, but influences Mg2+ concentration and causes problems for cytogenetic analysis5. It has also been shown that the RNA yield is higher in EDTA-blood than in heparin-blood6.
Blood volume: Larger volumes of blood may be processed with the same efficiency of separation by using centrifuge tubes of increased diameter while maintaining approximately the same heights of Ficoll-Paque media (2.4 cm) and blood sample (3.0 cm) as in the standard method described above. Increasing the tube diameter does not affect the separation time required.
Blood sample storage: Blood samples should be free of clots and processed as soon as possible after collection to ensure optimal results. Delays in processing the blood can result in loss of viability, lower cell recoveries and more contaminating granulocytes and/or erythrocytes. Indeed, storage for 24 h at room temperature has been reported to result in reduced lymphocyte yield, altered expression of surface markers, and reduced response to mitogenic stimulation 5,7,8. Dead cell removal: Dead cells are removed during cell isolation using Ficoll-Paque PLUS9,10,58.
Density and temperature: The temperature at which density gradient separations are carried out are naturally affected by room temperature, centrifuge temperature, temperature of the density gradient media, and temperature of the liquid sample. There can, at times, also be confusion regarding what is meant by room temperature (e.g., in Europe it can be 20ºC and in North America > 20ºC). It is known that the densities of Ficoll-Paque products decrease with increase in temperature. For instance Ficoll-Paque PREMIUM (1.077 g/ml) exhibits a density of 1.0772, 1.0767, and 1.0758 at 20ºC, 22ºC, and 25ºC, respectively.
Pathological blood samples: The standard method described above has been developed for the purification of mononuclear cells from peripheral blood of normal, healthy, human donors. Different results may be obtained with samples taken from donors with infections or other pathological conditions, such as cancer (see Further applications of Ficoll-Paque PLUS and Ficoll-Paque PREMIUM, page 12).
Platelet removal: If it is important to remove all platelets from the mononuclear cell fraction a second centrifugation in a 4% to 20% sucrose gradient layered over Ficoll-Paque PLUS can be applied. This procedure will effectively remove any platelet contamination11. Platelets will remain at the top of the sucrose gradient and mononuclear cells will sediment through the sucrose gradient to the top of the Ficoll-Paque PLUS layer. Alternatively, the platelets may be removed by aggregation with adenosine-5’-diphoshate (ADP) before separating the mononuclear cells12.
Troubleshooting inadequate performance
If used according to the recommended standard procedure, Ficoll-Paque PLUS and Ficoll-Paque PREMIUM may be expected to give trouble-free isolation of human peripheral blood mononuclear cells with results as shown on page 2. As mentioned earlier, Ficoll-Paque PREMIUM 1.073 and Ficoll-Paque PREMIUM 1.084 will lead to isolation of cell preparations having slightly different density subsets of mononuclear cells. However, deviations in certain experimental parameters may lead to poor results and the troubleshooting chart given here is intended to assist in the rapid identification and correction of the problem causing reduced performance.
Deviation in Performance Likely Source of Problem Comments
Increased red blood cell and contamination of the lymphocytes. A. Temperature too low. The densities of all Ficoll-Paque products are greater at low temperature (see Notes, Density and temperature) and red blood cells are aggregated less well, so granulocytes and red blood cells are prevented from entering the Ficoll-Paque media layer. Raise the temperature to 18ºC to 20°C.
  B. Centrifugation speed too slow and/ or centrifugation time too short. Adequate time and g-force must be used to ensurecomplete sedimentation of non-lymphoid cells.
Low yield and viabilityof mononuclear cells. Temperature too high. Ficoll-Paque products are less dense at high  temperature and some lymphocytes may penetrate into the Ficoll-Paque media layer. Cell viability may also be affected. Reduce thetemperature to 18ºC to 20°C.
Low yield of mononuclear cells with normal viability. Blood not diluted 1:1 with balanced salt solution; unusually highhematocrit. The high cell density results in large numbers oflymphocytes being trapped by red blood cell aggregates. Dilute the blood sample further.
Low yield of mononuclear cells with increased granulocyte contamination. Vibration of the centrifuge rotor, leading to stirring of the gradient. Vibration may cause broadening of the mononuclear cell band and mixing with the underlying cells. Check that the rotor is properly balanced. Choose rotor speed to avoidnatural resonant frequencies.
Low yield of mononuclear cells, low viability, and contamination by other cell types. Sample contains cells withabnormal densities; densitiesdifferent from those in normalhuman blood. May be encountered with pathological blood samples,non-human blood samples, old blood samples, or samples from sources other than peripheral blood. Percoll™ PLUS, a medium for density gradient centrifugation, may be moresuitable than Ficoll-Paque products for such separations.
  1. Isolation of mononuclear cells and gr anulocytes from human blood. (Paper IV). Bøyum, A., Scand. J., Clin. Lab. Invest. 21 Suppl, 97, 77–89 (1968).
  2. Isolation of leucocytes from human blood – further observations. (Paper II). Bøyum, A., Scand. J. Clin. Lab. Invest. 21 Suppl, 97, 31–50 (1968).
  3. Isolation of lymphocytes, granulocytes and macrophages. Bøyum, A., Scand. J., Immunol. 5 Suppl, 5, 9–15 (1976).
  4. Gel formation with leucocytes and heparin. Almeida, A.P., Beaven, M.A., Life Sci, 26, 549–555 (1980).
  5. Biological sample collection and processing for molecular epidemiological studies. Holland, N.T., Smith, M.T., Eskenazi, B., Bastaki, M., Mutation Research, 543, 217-234 (2003).
  6. Collection and storage of Human Blood Cells for mRNA Expression Profiling: A 15-Month Stability Study. Marteau, J., Mohr, S., le Pfister, M., Visvikis-Siest, S., Clinical Chemistry, 51, 1250-1252 (2005).
  7. Altered lymphocyte markers and blastogenic responses associated with 24 hour delay in processing of blood samples. Kaplan, J., Nolan, D., Ree, A. J., Immunol. Methods, 50, 187–191 (1982).
  8. Stability of hematological analytes depends on the hematology analyser used: A stability study with Bayer Advia 120, Beckman Coulter LH 750 and Sysmex XE 2100. Imeri, F., Herklotz, R., Risch, L., Arbetsleitner, C., Zerlauth, M., Risch, G.M., Huber, A.R., Clinica Chimica Acta, 397, 68-71 (2008).
  9. How to optimize multiparameter flow cytometry for leukaemia/lymphoma diagnosis. Paiette, E., Best Practice & Research Clinical Haematology, 16, 671-683 (2003).
  10. Dendritic Cells Cross-Dressed with Peptide MHC Class I Complexes Prime CD8 T Cells. Dolan, B.P., Gibbs Jr, K.D., Ostrand-Rosenberg, S. J., Immunol., 177, 6018-6024 (2006).
  11. Purification of lymphocytes and platelets by gradient centrifugation. Perper, R.J., Zee, T.W., Mickelson, M.M., J. Lab. Clin. Med., 72, 842–848 (1968).
  12. Platelet aggregation technique used in the preparation of lymphocyte suspensions. Vives, J., Parra, M., Castillo, R., Tissue Antigens, 1, 276–278 (1971).
  13. Quantitation of Fc receptors and surface immunoglobulin is affected by cell isolation procedures using Plasmagel and Ficoll-Hypaque. Alexander, E.L., Titus, J.A., Sega, D.M., J. Immunol. Methods, 22, 263–272 (1978).
  14. Analysis of the lymphocyte distribution during Isopaque-Ficoll isolation of mononuclear cells from human peripheral blood. Hokland, P., Heron, I., J. Immunol. Methods, 32, 31–39 (1980).
  15. The Isopaque-Ficoll method re-evaluated: Selective loss of autologous rosette-forming lymphocytes during isolation of mononuclear cells from human peripheral blood. Hokland, P., Heron, I., Scand. J. Immunol, 11,353–356 (1980).
  16. Lymphocyte imunnophenotyping by flow cytometry in normal adults Comparison of fresh whole blood lysis technique, Ficoll-Paque separation and cryopreservation. Romeu, M.A., Mestre, M., González, L., Valls, A., Verdaguer, J., Corominas, M., Bas, J., Massip, E., Buendia, E., J. Immunol. Methods, 154, 7-10 (1992).
  17. Expression of adhesion molecules on T lymphocytes in young children and infants – a comparative study using whole blood lysis or density gradient separation. Lin, S.-J, Chao, H.-C, Yan, D.-C, Huang, Y.-J., Clin. Lab. Haem, 24, 353-359 (2002).
  18. Reactivity in mixed cultures of mononuclear leucocytes separated on Ficoll-Hypaque. Bain, B., Pshyk, K. Proceedings 7th Leucocyte Culture Conference, (Ed. Daguillard, F.), Academic Press, New York, 29–37 (1973).
  19. Membrane Surface Nanostructures and Adhesion Property of Tlymphocytes Exploited by AFM. Wu, Y., Lu, H., Cai, J., He, X., Hu, Y., Zhao, H., Wang, X., Nanoscale Res. Lett., 4, 942-947 (2009).
  20. A role for interleukin-12/23 in the maturation of human natural killer and CD56 T cells in vivo. Guia, S., Cognet, C., de Beaucoudrey, L., Tessmer, M.S., Jouanguy, E., Berger, C., Filipe-Santos, O., Feinberg, J., Camcioglu, Y., Levy, J., Jumaah, S.A., Al-Hajjar, S., Stephan, J., Fieschi, C., Abel, L., Brossay, L., Casanova, J., Vivier, E., J. Blood, 111, 5008-5016 (2008).
  21. Targeting NF- B activation via pharamacologic inhibition of IKK2-induced apoptosis of human acute myeloid leukemia cells. Frelin,C., Imbert, V., Griessinger, E., Peyron, A-C., Rochet, N., Philip, P., Dageville, C., Sirvent, A., Hummelsberger, M., Berard, E., Dreano, M., Sirvent, N., Peyron, J. Blood, 105, 804-911 (2005).
  22. Ficoll-separated mononuclear cells from sepsis patients are contaminated with granulocytes. Van den Akker, E.L.T., Baan, C.C., Van den Berg, B., Russcher, H., Joosten, K., Hokken-Koelega, A.C.S., Lamberts, S.W. J., Koper, J.W., Intensive Care Med, 34, 912-916 (2008).
  23. A novel technique for the enrichment of primary ovarian cancer cells. Chan, J.K., Hamilton, C.A., Anderson, E.M., Cheung, M.K., Baker, J, Husain, A., Teng, N.N., Kong, C.S., Negrin, R.S., Am. J. Obstet. Gynecol., 197, 507. e1—507.e5, (2007).
  24. Non-lymphoid cells obtained by the Bøyum technique and their significance in cancer patients. Kluin-Nelemans, J.C., van Helden, H.P.T., J. Clin. Lab. Immunol., 4, 99–102 (1980).
  25. Gradient separation of normal and malignant cells. II. Application to in vivo tumour diagnosis. Minami, R., Yokota, S., Teplitz, R.L., Acta. Cytol., 22, 584–588 (1978).
  26. Enhancement of human amniotic cell growth by Ficoll-Paque gradient fractionation. Chang, H-C., Jones, O.W., Bradshaw, C., et al., In Vitro., 17, 81–90 (1981).
  27. Optimization of immunomagnetic separation for cord blood-derived hematopoietic stem cells. Kekarainen, T., Mannelin, S., Laine, J., Jaatinen, T., BMC Cell Biology, 7 (2006).
  28. Prolonged ex vivo culture of human bone marrow mesenchymal stem cells influences their supportive activity toward NOD/SCID-repopulating cells and committed progenitor cells of B lymphoid and myeloid lineages. Briquet, A., Dubois, S., Bekaert, S., Dolhet, M., Beguin, Y., Gothot, A., Haematologica, 95(1), 47-56 (2010).
  29. The active translation of MHCII mRNA during dendritic cells maturation supplies new molecules to the cell surface pool. Malanga, D., Barba, P., Harris, P.E., Maffei, A., Del Pozzo, G., Cell. Immunol., 246, 75-80 (2007).
  30. Reduced number and function of peripheral dendritic cells in celiac disease. Ciccocioppo, R., Ricci, G., Rovati, B., Pesce, I., Mazzocchi, S., Piancatelli, D., Cagnoni, A., Millimaggi, D., Danova, M., Corazza, G.R., Clin. and Exp. Immunol., 146, 487-496 (2007).
  31. Anti-proteinase 3 antibodies (c-ANCA) prime CD14-dependent leukocyte activation. Hattar, K., van Burck, S., Bickenbach, A., Grandel, U., Maus, U., Lohmeyer, J., Csernok, E., Hartung, T., Seeger, W., Grimminger, F., Sibelius, U., J. Leukocytes Biol., 78, 992-1000 (2005).
  32. Modeling the initiation and progression of Human Acute Leukemia in Mice. Lubin, I., Faktorowich, Y., Lapidot, T., Gan, Y., Eshhar, Z., Gazit, E., Levite, M., Reisner, Y., Science. New Series, 252, 427-431 (1991).
  33. Noncanonical Wnt11 Signaling Is Sufficient to Induce Cardiomyogenic Differentiation in Unfractionated Bone Marrow Mononuclear Cells. Flaherty, M.P., Abdel-Latif, A.,Li, Q., Hunt, G., Ranjan, S., Ou, Q., Tang, X., Johnson, R.K., Bolli, R., Dawn, B., Circulation, 117, 2241-2252 (2008).
  34. Double-label expression studies of prostacyclin synthase, thromboxane synthase and COX isoforms in normal aortic endothelium. Kawka , D.W., Ouellet, M., Hétu, P., Singerm I.I., Riendeau, D., Biochimica et Biophysica Acta., 1771, 45–54 (2007).
  35. Growth of bone marrow stromal cells on small intestinal submucosa: an alternative cell source for tissue engineered bladder. Zhang, Y., Lin, H., Frimberger, D., Epstein, R.B., Kropp, B.P., BJU International, 96, 1120-1125 (2005).
  36. Expression of mRNA for multiple serotonin (5-HT) receptor types/subtypes by the peripheral blood mononuclear cells of rhesus macaques. Yang, G., Qiu, C., Zhao, H., Liu, Q., Shao, Y., J. Neuroimmunol., 178, 24-29 (2006).
  37. Functional Analysis of Neuron-like Cells Differentiated from Neural Stem Cells Derived from Bone Marrow Stroma Cells in vitro. Xu, R., Jiang, X., Guo, Z., Chen, J., Zou, Y., Ke, Y., Zhang, S., Li, Z., Cai, Y., Du, M., Qin, L., Tang, Y., Zeng, Y., Cell Mol. Neurobiol., 28, 545-558 (2008).
  38. The bovine lymphoid system: Binding and stimulation of peripheral blood lymphocytes by lectins. Pearson, T.W., Roelants, G.E., Lundin, L.B., et al., J. Immunol. Methods, 26, 271–282 (1979).
  39. Acid a-naphthyl acetate asterase: presence of activity in bovine and human T and B lymphocytes. Yang, T.J., Jantzen, P.A., Williams, L.F., J. Immunol., 38, 85–93 (1979).
  40. The effect of blue light exposure in an ocular melanoma animal model. Di Cesare, S., Maloney, S., Fernandes, B.F., Martins, C., Marshall, J., Antecka, E., Odashiro, A.N., Dawson, W.W., Buriner Jr, M.N., J. Exp. and Clin. Cancer Res., 28:48, 1-9 (2009).
  41. Comparative study of six methods for lymphocyte isolation from several mammalian sources and determination of their carbohydrate composition. Hueso, P., Rocha, M., (Article in Spanish) Rev. Esp. Fisiol., 34, 339–344 (1978).
  42. Effect of cryopreservation on IL-4, IFNc and IL-6 production of porcine peripheral blood lymphocytes. Li, X., Zhong, Z., Liang, S., Wang, X., Zhong, F., Cryobiology, 59, 322-326 (2009).
  43. A comparison of the methods used for the separation of fish lymphocytes. Blaxhall, P.C., J. Fish Biol, 18, 177–181 (1981).
  44. Separation of human peripheral blood monocytes on continuous density gradients of Polyvinylpyrrolidone-coated silica gel (Percoll). Brandslund, I., Møller-Rasmussen, J., Fisker, D., et al., J. Immunol. Methods, 48, 199–211 (1982).
  45. Efficient separation of human T lymphocytes from venous blood using PVP-coated colloidal silica particles (Percoll). Feucht, H.E., Hadam, M.R., Frank, F., et al., J. Immunol. Methods, 38, 43–51 (1980).
  46. An improved technique for the isolation of lymphocytes from small volumes of peripheral mouse blood. Mizobe, F., Martial, E., Colby‑Germinario, S., et al., J. Immunol. Methods, 48, 269–279 (1982).
  47. Quantitative and Qualitative Comparative Analysis of Gradient Separated Hematopoietic Cells from Cord Blood and Chemotherapy-Mobilized Peripheral Blood. Sato, J., Kawano, Y.F., Takaue, Y., Hirao, A., Makirnoto, A., Okarnoto, Y., Abe, T., Shimokawa, T., Iwai, A., Kurodaa, Y., Stem Cells, 13, 548-555 (1995).
  48. EC Guide to GMP (Good Manufacturing Practice), annex 1 “Manufacture of Sterile Medicinal Products”.
  49. United States Pharmacopeia. Recommendations for ancillary materials, chapter <1043>
  50. Lymphocyte differentiation in the rabbit thymus. Leene, W., et al., Ann. Immunol. (Paris), 127, 911-921 (1976).
  51. Separation of leucocytes: improved cell purity by fine adjustments of gradient medium density and osmolality. Bøyum, A., et al., Scand. J. Immunol., 34, 697-712 (1991).
  52. Isolation of bovine colostral lymphocytes: in vitro blastogenic responsiveness to concanavalin A and bovine rotavirus. Archambault, et al., Ann. Rech. Vet., 19, 169-174 (1988).
  53. Characterization and species distribution of high affinity GTP-coupled receptors for human rantes and monocyte chemoattractant protein. Van Riper et al., JEM, 177, 851-856 (1993).
  54. Cryopreserved mesenchymal stromal cell treatment is safe and feasible for severe dilated ischemic cardiomyopathy, Chin et al., Cytotherapy, 12, 31-37 (2010),
  55. Mesenchymal stem cells from multiple myeloma patients display distinct genomic profile as compared with those from normal donors, Garayoa et al., Leukemia, 23, 1515-1527 (2009).
  56. GMP-manufactured density gradient media for optimized mesenchymal stromal/stem cell isolation and expansion, Grisendi et al., Cytotherapy, 12, 466-477 (2010).
  57. Manufacturing of human placenta-derived mesenchymal stem cells for clinical trials. Brooke et al., British J. of Haematology, 144, 571-579 (2008).
  58. High gradient magnetic cell separation with MACS, Miltenyi et al., Cytometry, 11, 231-238 (1990).
  59. Reduced expression of ATP-binding cassette transporter G1 increases cholesterol accumulation in macrophages of patients with type 2 diabetes mellitus. Mauldin, J. P. et al., Circulation, 117, 2785-2792 (2008).
  60. Defined serum-free culturing conditions for neural tissue engineering of human cord blood stem cells. Hamad A. et al., Acta. Neurobiol. Exp., 69, 11-23 (2009).
  61. High purity and yield of natural Tregs from cord blood using a single step selection method, Figueroa-Tentori et al., J. Immunol. Methods, 339, 228-235 (2008).
Related Links