KiCqStart™ Universal SYBR® Green qPCR Protocol
Background
KiCqStart SYBR Green qPCR ReadyMix is a 2X concentrated, ready-to-use reaction cocktail that contains all components, except primers and template, for real-time quantitative PCR (qPCR). This unique combination of proprietary buffer, stabilizers, and KiCqStart Taq DNA polymerase delivers maximum PCR efficiency, sensitivity, specificity and robust fluorescent signal using fast, or conventional cycling protocols with SYBR Green qPCR.
Highly specific amplification is crucial to successful qPCR with SYBR Green I dye technology because this dye binds to and detects any dsDNA generated during amplification. KiCqStart Taq DNA polymerase contains a proprietary mixture of monoclonal antibodies that bind to the polymerase and keep it inactive prior to the initial PCR denaturation step (> 48 hours at room temperature). Activation of the enzyme is instantaneous at 95 °C. Replication of fragments up to 200 bp is complete in less than 20s at 60 °C.
For additional information, please see the qPCR Technical Guide or SYBR Green I-based qPCR technical animation.
Instrument Compatibility for KiCqStart® SYBR® Green qPCR ReadyMix™
Different real-time PCR systems employ different strategies for the normalization of fluorescent signals and correction of well-to-well optical variations. It is critical to match the appropriate qPCR reagent to your specific instrument. KiCqStart SYBR Green qPCR ReadyMix does not contain an internal reference dye. Please consult the following table to find the optimal kit for your instrument platform.
KiCqStart® SYBR® Green qPCR ReadyMix™ Compatible Instruments
KCQS00 Bio-Rad CFX384™, Bio-Rad CFX96™, Bio-Rad MiniOpticon™, Bio-Rad/MJ Chromo4™, Bio-Rad/MJ Opticon 2, Bio-Rad/MJ Opticon®, Cepheid SmartCycler®, Eppendorf Mastercycler® ep realplex, Eppendorf Mastercycler® ep realplex2 s, Illumina Eco qPCR, Qiagen/Corbett Rotor-Gene® 3000, Qiagen/Corbett Rotor-Gene® 6000, Qiagen/Corbett Rotor-Gene® Q, Roche LightCycler™ 480
KCQS01 Applied Biosystems 7500, Applied Biosystems 7500 Fast, Applied Biosystems ViiA 7, Stratagene Mx3000P®, Stratagene Mx3005P™, Stratagene Mx4000™
KCQS02 Applied Biosystems 5700, Applied Biosystems 7000, Applied Biosystems 7300, Applied Biosystems 7700, Applied Biosystems 7900, Applied Biosystems 7900 HT Fast, Applied Biosystems 7900HT, Applied Biosystems StepOnePlus™, Applied Biosystems StepOne™
KCQS03 BioRad iCycler iQ™, BioRad iQ™5, BioRad MyiQ™
Supplies
Assay Guidelines
  • Primer Design:
    • Predesigned primers: We offer a panel of predesigned primers that have been optimized for use with the KiCqStart SYBR Readymixes. These KiCqStart Primers can be ordered under product number KSPQ12012.
    • Custom Primers:
      • The design of highly specific primers is the single most important parameter for successful real-time PCR with SYBR Green I dye. The use of computer aided primer design programs is encouraged in order to minimize the potential for internal secondary structure and complementation at 3’-ends within each primer and the primer pair.  Custom primers can be ordered under product number OLIGO.
    • Amplicon Size: KiCqStart SYBR Green qPCR ReadyMix can readily amplify fragments between 400 and 500 bp; however, to take full advantage of fast cycling protocols, amplicon size should be limited to less than 150 bp.
    • Primer Concentration: Optimal results may require titration of primer concentration between 100 and 500 nM. A final concentration of 300 nM for each primer is effective for most reactions.
  • Assay Setup:
    • Preparation of a reaction cocktail is recommended to reduce pipetting errors and maximize assay precision. Assemble the reaction cocktail with all required components except sample template (genomic DNA or cDNA) and dispense equal aliquots into separate reaction tubes. Add the DNA template to each reaction as the final step. Dilution of template samples to allow at least a 5 µL addition will improve assay precision.
    • Suggested input quantities of template are: cDNA corresponding to 1 pg to 100 ng of total RNA; 100 pg to 100 ng genomic DNA
Reaction Setup
Reagent Volume for 20 μL reaction Final Concentration
KiCqStart SYBR Green qPCR ReadyMix (2X)
10.0 µL 1X
Forward Primer variable 100 – 500 nM (start with 300 nM)
Reverse Primer variable 100 – 500 nM (start with 300 nM)
Nuclease-free water (W1754) variable Add to q.s. to 20 µL
Template at least 5 μL variable
Final Volume (µL) 20 µL  
Final reaction volume may vary from 10 to 50 μL, scale all components proportionally.
After sealing each reaction, vortex gently to mix contents. Centrifuge briefly to collect components at the bottom of the reaction tube.
PCR Cycling Protocol
  Fast 2-Step Cycling Fast 3-Step Cycling Standard Cycling
Initial denaturation: 95 °C, 30 sec * 95 °C, 30 sec * 95 °C, 2-3 min *
PCR cycling (30-45 cycles): 95 °C, 3-5 sec 95 °C, 3-5 sec 95 °C, 10-15 sec
    55 to 65 ºC, 15 sec  
Collect data at end of extension step 60 °C, 20-30 sec 68 to 72 °C, 10 sec 60 °C, 30-60 sec
Melt Curve (dissociation stage) Refer to instrument instructions (optional)
* Full activation of KiCqStart Taq DNA polymerase occurs within 1 second at 95 ºC; however, optimal initial denaturation time is template dependent and will affect qPCR efficiency and sensitivity. Amplification of genomic DNA or supercoiled plasmid DNA targets may require 5 to 10 min at 95 ºC to fully denature and fragment the template. Short double-stranded DNA template (PCR product) or single-stranded DNA template, may require as little as 1sec at 95 ºC. Use 30 sec at 95 ºC as a general starting point.
Extension time is dependent upon amplicon length and minimal data collection time requirement for your qPCR instrument. Some primer sets may require a 3-step cycling protocol for optimal performance. Optimal annealing temperature and time or primer concentration may need to be empirically determined for any given primer set and real-time instrument.
Materials
SYBR is a registered trademark of Molecular Probes, Inc., KiCqStart is a trademark of Quanta BioSciences, TaqMan is a registered trademark of Roche Molecular Systems, Inc.  LightCycler is a registered Trademark of Roche. Applied Biosystems, StepOne, StepOnePlus, ViiA, and ROX are trademarks Life Technologies Corporation. Stratagene, MX3000P, MX3005P and MX4000 are trademarks of Agilent Technologies, Inc. Mastercycler is a trademark of Eppendorf. Rotor-Gene is a registered trademark of Qiagen GmbH.  SmartCycler is a trademark of Cepheid. CFX96, CFX384, iCycler iQ, iQ5, MyiQ, Opticon, MiniOpticon and Chromo4 are trademarks of Bio-Rad Laboratories. SYBR is a registered Trademark of Molecular Probes, Inc.
Refer to product technical bulletin for product licensing information.
Related Links