Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Method for PCR amplification of Bacterial DNA with low contamination using MTP™ Taq DNA Polymerase (D7442)
Background
MTP™ Taq DNA Polymerase is a recombinant thermostable enzyme from Thermus aquaticus expressed in E. coli and purified using a proprietary process to minimize levels of contaminating DNA. The enzyme has both 5'→3' DNA polymerase and exonuclease activities, is ~95 kDa by SDS-PAGE, and has no detectable endonuclease or 3'→5' exonuclease activities. Each lot of MTP Taq undergoes strict quality control testing to ensure the absence of detectable levels of contaminating DNA.
Contaminating DNA present in most other polymerase preparations often preclude or obscure the accurate interpretation of results, especially when targeting conserved sequences, e.g., bacterial 16S rRNA region.1 Through Sigma's proprietary DNA removal methods and strict quality control standards, we can ensure the absence of the most commonly found contaminant DNA. Each lot of MTP Taq is assayed using PCR and primers specific to (1) the conserved region of bacterial 16S rRNA, (2) the Taq expression vector, and (3) the human β-actin gene.
While MTP Taq ensures a high-quality, low contaminant DNA polymerase for reliable PCR amplification, DNA contaminants can be introduced into PCR through a number of other reagents.2 To further minimize the risk of contaminant DNA during PCR, we include 10x MTP Taq Buffer with each tube of MTP Taq DNA Polymerase. Each lot of 10x MTP Taq Buffer undergoes the same strict quality control testing as MTP Taq DNA polymerase to ensure the absence of contaminating DNA. To prevent false positive PCR results, only DNA-free reagents should be used in PCR reactions with MTP Taq DNA polymerase.
Figure 1.
Representative QC gel assaying for the presence of conserved 16S rRNA sequence. PCR products run in lanes 1-3 are from "no-template" reactions; PCR products in lanes 4-5, 6-7, and 8-9 are from reactions with 37 fg, 370 fg, and 3.7 pg of E. coli genomic DNA added, Catalog Number D4889.
Unit definition: One unit incorporates 10 nmol of total deoxyribonucleoside triphosphates into acid precipitable DNA in 30 minutes at 74 °C.
Reagents
  • MTP™ Taq DNA Polymerase (D7442):
    • MTP Taq DNA Polymerase (D7067)
    • 10X MTP Taq Buffer (M9943)
  • PCR grade water (W1754)
  • Primers diluted to working concentration (10µM working stocks are sufficient for most assays)
  • DNA to be amplified
  • Dedicated pipettes
  • Thermal cycler
  • Sterile filter pipette tips
  • Sterile 1.5 mL screw-top microcentrifuge tubes (such as CLS430909)
  • PCR tubes, select one of the following to match desired format:
  • dNTP mix, 10 mM each of dATP, dCTP, dGTP, and dTTP (D7295)
  • Thermal cycler
Procedure
Note: Every precaution should be taken to avoid contamination of reagents with unknown/unwanted DNA. This includes the following:
  • Use a "clean area" for the setup of PCR reactions.2 A clean area is a separate lab space (such as a hood) preferably in a separate lab, both of which are free from PCR products. The clean area should contain dedicated lab coats and pipettes (see below) and should be cleansed (either with 10% bleach or UV lamp) after each use. PCR products should never enter a clean area.
  • Use sterile, aerosol barrier pipette tips to minimize the risk of aerosol contamination. Change tips after each single use.
  • Change gloves frequently, especially after handling DNA.
Amplification Procedure
The optimal conditions for the concentration of MTP Taq DNA Polymerase, template DNA, primers, and MgCl2 will depend on the system being utilized. It may be necessary to determine the optimal conditions for each individual component. It is recommended that the enzyme and the MgCl2 be titrated to determine the optimal efficiency if the below protocol is shown to be less than satisfactory.
  1. Add the following reagents to a suitable PCR tube/plate in the following order:
    Volume
    Reagent
    Final Concentration
    w µL Water -
    5 µL 10x MTP Taq buffer 1x
    1 µL 10 mM dNTP mix 200 µM each dNTP
    x µL Forward Primer (typically 15-30 bases in length) 0.1–0.5 µM
    y µL Reverse Primer (typically 15-30 bases in length) 0.1–0.5 µM
    0.5 µL MTP Taq DNA Polymerase 0.05 units/µL
    z µL Template DNA (typically 10 ng) 200 pg/µL
    50 µL* Total reaction volume    
    *Maintain final concentrations when scaling reaction volumes. Preparing enzyme / nucleotide / buffer master mixes either with or without primers or template facilitates the setup of multiple reactions.
  2. Vortex gently to mix. Briefly centrifuge to collect reaction on the bottom of the tube.
  3. If using a thermal cycler without a heated lid, add 50 µL of mineral oil to the top of each tube to prevent evaporation.
  4. Cycle using thermocycler of choice. The amplification parameters may require optimization for individual primers, templates, and thermal cyclers.
    Common cycling parameters are:
    Step Temperature Time Cycles
    Denature 94 ºC 1 minute 1
    Denature 94 ºC 1 minute 30
    Anneal 5 ºC below primer Tm
    15 - 60 seconds
    Extend 72 ºC 30 - 65 seconds/kb
    Cool 4 ºC ≥ 0 second 1
  5. Amplified DNA can be evaluated by standard methods (e.g. agarose gel electrophoresis).3
  6. If used, mineral oil may be removed by a single 50 µL chloroform extraction.
Materials
MTP is a trademark of Sigma-Aldrich Co. LLC.
Restorase is a registered trademark of Sigma-Aldrich Co. LLC.
TWEEN is a registered trademark of Croda International PLC.
Label Licence Statement
NOTICE TO PURCHASER: DISCLAIMER OF LICENCE
No license is conveyed with the purchase of this product under any of US Patents Nos.5,804,375, 5,994,056, 6,171,785, 6,214,979, 5,538,848, 5,723,591, 5,876,930, 6,030,787, and 6,258,569 and corresponding patents outside the United States, or any other patents or patent applications, relating to the 5’ Nuclease and dsDNA-Binding Dye Processes. For further information contact the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Related Links