REDExtract-N-Amp™ Plant PCR Kits Protocol
Catalog Numbers: XNAPS, XNAP, XNAPE, and XNAPR
Product Description
The REDExtract-N-Amp Plant PCR Kits contain all the reagents needed to rapidly extract and amplify genomic DNA from plant leaves. Briefly, the DNA is extracted from a piece of leaf tissue, a 0.5 to 0.7 cm disk cut with a standard paper punch, by incubation in Extraction Solution at 95 °C for 10 minutes. There is no need for freezing plant tissue in liquid nitrogen, mechanical disruption, organic extraction, column purification or precipitation of the DNA. After an equal volume of Dilution Solution is added to the extract to neutralize inhibitory substances, the extract is ready for PCR.
An aliquot of the diluted extract is then combined with the REDExtract-N-Amp PCR ReadyMix™ and userprovided PCR primers to amplify target DNA. The REDExtract-N-Amp PCR ReadyMix is a 2X reaction mix containing buffer, salts, dNTPs, and Taq polymerase. It is optimized specifically for use with the extraction reagents. It also contains JumpStart™ Taq antibody for hot start PCR to enhance specificity and REDTaq® dye to allow direct loading of the PCR product onto an agarose gel.
Reagents Provided
(100 preparations)
Extraction Solution E7526 1.2 ml 12 ml 12 ml 120 ml
Dilution Solution D5688 1.2 ml 12 ml 12 ml 120 ml
REDExtract-N-Amp PCR ReadyMix. This is a 2X PCR reaction mix containing buffer, salts, dNTPs, Taq polymerase and JumpStart Taq antibody. R4775 0.15 ml 1.2 ml 5 x 1.2 ml 12 ml
Collection Tubes, 2 ml T5449
Not included 2 x 50 each 2 x 50 each Not included
Reagents and Equipment Required But Not Provided
Precautions and Disclaimer
The REDExtract-N-Amp Plant PCR Kits are for laboratory use only. Not for drug, household or other uses. Consult the SDS for information regarding hazards and safe handling practices.
The Extraction Solution, Dilution Solution and REDExtract-N-Amp PCR ReadyMix can be stored at 2-8 °C for short-term; –20 °C for long-term. Do not store in a "frost-free" freezer.
All steps are carried out at room temperature unless otherwise noted.
A.    DNA extraction
1.    Rinse the paper punch and forceps in 70% ethanol prior to use and between handling different samples.
2.    Punch a 0.5 to 0.7 cm disk of leaf tissue into a 2 ml collection tube using a standard one-hole paper punch. If
       frozen plant tissue is used, keep the leaves on ice while punching disks.
3.    Add 100 µL of Extraction Solution to the collection tube. Close the tube and vortex briefly. Make sure the disk
       is covered by the Extraction Solution.
4.    Incubate at 95 °C for 10 minutes. Note that leaf tissues usually do not appear to be degraded after this
5.    Add 100 µL of Dilution Solution and vortex to mix.
6.    Store the diluted leaf extract at 2-8 °C. It is not necessary to remove the leaf disk before storage.
B.    PCR amplification
The REDExtract-N-Amp PCR ReadyMix contains JumpStart Taq antibody for specific hot start amplification. Therefore, PCR reactions can be assembled at room temperature without premature Taq DNA polymerase activity.
Typical final primer concentrations are ~0.4 µM each. The optimal primer concentration and cycling parameters will depend on the system being used.
1.    Add the following reagents to a thin-walled PCR microcentrifuge tube:
Reagent Volume
Water, PCR reagent x µL
REDExtract-N-Amp PCR ReadyMix 10 µL
Forward primer y µL
Reverse primer y µL
Leaf disk extract 4 µL*
Total volume 20 µL
*Note: The REDExtract-N-Amp PCR ReadyMix is formulated to compensate for components in the Extraction and Dilution Solutions. If less than 4 μL of leaf disk extract is added to the PCR reaction volume, use a 50:50 mixture of Extraction:Dilution Solutions to bring the volume of leaf disk extract up to 4 μL.
2.    Mix gently and briefly centrifuge to collect all components at the bottom of the tube.
3.    For thermal cyclers without a heated lid, add 20 μL of mineral oil to the top of each tube to prevent evaporation.
4.    The amplification parameters should be optimized for individual primers, template, and thermal cycler.
Common cycling parameters:
Step Temperature Time Cycles
Initial Denaturation 94 °C 3 min. 1
Denaturation 94 °C 0.5-1 min 30-35
Annealing 45 to 68 °C 0.5-1 min
Extension 72 °C 1-2 min
(~1 kb/min)
Final Extension 72 °C 10 min. 1
Hold 4 °C Indefinitely  
5.    The amplified DNA can be loaded directly onto an agarose gel after the PCR is completed. It is not necessary to
       add a separate loading buffer/tracking dye.
       Note: PCR products can be purified, if desired, for downstream applications, such as sequencing, with the
       GenElute™ PCR Clean-Up Kit, Catalog No. NA1020.
Related Products
Troubleshooting Guide
Problem Cause Solution
Little or no PCR
product is
PCR reaction may be inhibited due to contaminants in the
plant extract.
Dilute the extract with a 50:50 mix of Extraction and Dilution solutions. To test for inhibition, include a DNA control and/or spike a known amount of template (100-500 copies) into the PCR mixture along with the plant extract.
A PCR component may be missing or
Run a positive control to insure components are functioning. A checklist
is also recommended when assembling reactions.
There may be too few cycles performed. Increase the number of cycles (5-10 additional cycles at a time).
The annealing
temperature may be too high.
Decrease the annealing temperature in 2-4 °C increments.
The primers may not be designed optimally. Confirm the accuracy of the sequence information. If the primers are less than 22 nucleotides long, try to lengthen the primer to 25-30 nucleotides. If the primer has a GC content of less than 45%, try to redesign the primer with a GC content of 45-60%.
The denaturation
temperature may be too high or too low.
Optimize the denaturation temperature by increasing or decreasing the temperature in 1 °C increments.
The denaturation time may be too long or too short. Optimize the denaturation time by increasing or decreasing the time in 10 second increments.
The extension time may be too short. Increase the extension time in 1 minute increments, especially for long templates.
Target template is
In most cases, inherently difficult targets are due to unusually high GC content and/or secondary structure. Betaine has been reported to help amplification of high GC content templates at a concentration
of 1.0-1.7 M.
Multiple products
JumpStart Taq antibody is not working correctly.
Do not use DMSO or formamide with REDExtract-N-Amp PCR ReadyMix. It can interfere with the enzyme-antibody complex.Other cosolvents, solutes (e.g., salts) and extremes in pH or other reaction conditions may reduce the affinity of the JumpStart Taq antibody for Taq polymerase and thereby compromise its effectiveness.
Touchdown PCR may be needed. “Touchdown” PCR significantly improves the specificity of many PCR reactions in various pplications. Touchdown PCR uses an annealing/extension temperature thatis higher than the Tm of the primers during the initial PCR cycles. The annealing/extension temperature is then reduced to the primer Tm for the remaining PCR cycles. The change can be performed in a single step or in increments over several cycles.
Contamination Reagents are
Sigma recommends that a reagent blank without DNA template be included as a control in every PCR run to determine if the reagents used in extraction or PCR are contaminated with a template from a previous reaction.
  1. Dieffenbach, C.W. and Dveksler, G.S. (Eds.) PCR Primer: A Laboratory Manual, 2nd ed., Cold Spring Harbor Laboratory Press, New York, (2003). Catalog Number Z701270
  2. Don, R.H. et al. ‘Touchdown' PCR to circumvent spurious priming during gene amplification. Nucleic Acids Res., 19, 4008 (1991).
  3. Erlich, H.A. (Ed.) PCR Technology: Principles and Applications for DNA Amplification, Stockton Press, New York (1989).
  4. Griffin, H.G. and Griffin, A.M. (Eds.) PCR Technology: Current Innovations, CRC Press, Boca Raton, FL, 1994.
  5. Innis, M.A., et al. (Eds.) PCR Strategies, Academic Press, New York (1995).
  6. Innis, M., et al. (Eds.) PCR Protocols: A Guide to Methods and Applications, Academic Press, San Diego, California (1990).
  7. McPherson, M.J. et al. (Eds.) PCR 2: A Practical Approach, IRL Press, New York (1995).
  8. Newton, C.R. (Ed.) PCR: Essential Data, John Wiley & Sons, New York (1995).
  9. Roux, K.H. Optimization and troubleshooting in PCR. PCR Methods Appl., 4, 5185-5194 (1995).
  10. Saiki, R., PCR Technology: Principles and Applications for DNA Amplification, Stockton, New York (1989).
Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224 and 5,618,711. The purchase of this product includes a limited, nontransferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser’s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser's activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
GenElute, JumpStart, ReadyMix and REDExtract-N-Amp are trademarks, and REDTaq is a registered trademark, of Sigma-Aldrich Co. LLC.
JC,RC,PHC 01/13-1
Related Links