Applications with REDTaq™
by Brian Ward and Keming Song
Sigma-Aldrich Corporation, St. Louis, MO, USA
Since its introduction, REDTaq has been well received by the scientific community. The original product idea was to add convenience to PCR by formulating a reaction that also contained loading buffer and dye. The end product would allow the researcher to see which samples contained enzyme and would remove one step in post-PCR product analysis, but would not hinder any conceivable post-PCR uses for the amplified DNA. The product name, REDTaq, is meant to reflect simplicity of use to the customer; the enzyme would work like Taq polymerase in every other way, but have color.
Everyday Taq Taq for use with Genomic DNA or
complex templates
without Hot Start Antibody With Hot Start Antibody Containing MgCl2 Separate MgCl2
Containing MgCl2 With a tube of dNTPs Without a tube of dNTPs Without a tube of dNTPs
REDTaq® DNA Polymerase
JumpStart™REDTaq®DNA Polymerase
REDTaq® SuperPak™ DNA Polymerase D6063) REDTaq® Genomic DNA Polymerase, with MgCl2 (D8312) REDTaq® Genomic DNA Polymerase, without MgCl2 (D2812)
Readymixes- premixed PCR mastermix
with buffer and dNTPs
High Fidelity Proofreading Taq
without Hot Start Antibody With Hot Start Antibody without Hot Start Antibody with Hot Start Antibody
high [Taq] low [Taq]
ReadyMix™PCR Reaction Mix
JumpStart™ REDTaq® ReadyMix™
REDAccuTaq® LA DNA Polymerase
JumpStart™ REDAccuTaq® LA DNA Polymerase
Results and Discussion
REDTaq in downstream applications
From the myriad of chromophores that could have potentially fulfilled the above objectives, one was selected to provide an amplification system that performed as well as our standard Taq DNA polymerase. Included in the screening were PCR and post-PCR concerns including PCR toxicity, PCR product purification methods, restriction digests, ligation, and transformation. One of the most stringent tests of the screening was PCR yield. As can be seen in Figure 1, the PCR yields with the newly formulated product are comparable to those of Taq. Additionally, PCR products from a REDTaq reaction mixture can be used in automated sequencing without causing fluorescent artifacts or hindering the sequencing reaction or length of read. An unforeseen benefit of REDTaq is that one can easily see if a reaction has been thoroughly mixed. All in all, REDTaq performs as we first envisioned. Aside from the added conveniences of visualization and direct loading, the additives have no effect on the reaction.
Compatibility of REDTaq in long and hot start PCR
Since the initial formulation of REDTaq, we have found the formulation to be completely inert when stored as a PCR master mix. This has allowed the recent release of REDTaq ReadyMixes. Slight optimizations of the initial formulation have been shown compatible with Taq blends (AccuTaq™) and Taq directed antibodies (JumpStart™). In Figure 2 the compatibility of REDTaq with hot start antibody is demonstrated by showing the effect of REDTaq with and without antibody when template amounts are low.
Figure 1. Relative PCR product yields as a function of target sequence
length using REDTaq and Taq DNA polymerases.
Product yields were
quantified as acid insoluble counts from [32P]dCTP containing PCRs. Yields are
normalized to those of Taq DNA polymerase. Lambda phage DNA was used
as a template, and reactions were performed in duplicate. Product quality
was independently verified by agarose gel electrophosesis of parallelnonradioactive
Figure 2. REDTaq with and without hot start antibody. PCR reactions
with wheat total genome DNA: 400 bp target amplicon. Reactions were
assembled then allowed to incubate at room temperature for two hours
prior to amplification. Lane 1 contains markers. Lanes 2-3 show duplicate
PCR using 0.4 ng total template with REDTaq DNA polymerase. Lanes 4-5
show yields for the same reaction using JumpStart REDTaq DNA polymerase,
in duplicate.
Optimizing REDTaq PCR
As REDTaq is essentially identical in performance with Taq, REDTaq and Taq PCR optimization are operationally identical. However, REDTaq is different from Taq in its magnesium requirements. Although methods of optimizing magnesium concentration for REDTaq and Taq will be unchanged, one should be aware that magnesium concentrations optimized for Taq may not be optimal for REDTaq, even if all other parameters (template, primers cycling conditions etc.) are the same. PCR optimization can usually be effected by changing one or more of the following (in order of importance): template quality, primer design, cycling parameters and magnesium concentrations. There are many excellent reviews covering PCR optimization,1,2 and most good technical libraries will include recent books covering general PCR techniques.3,4 Most troublesome PCR can be made to work by changing one or more of the parameters described previously. However, this is far from a general method for PCR optimization. If template quality is sufficient, then the problem is often due to the complexities involved in PCR annealing, which is a function of the melting temperature (Tm). Tm is the temperature at which the annealing and dissociation rates of the primer: template complex are equal.
Dissociation is unimolecular and therefore independent of concentration, while annealing is bimolecular and dependent on PCR product and primer concentrations. To put it simply, Tm values change during PCR. One technique, which allows a certain amount of error in Tm calculation, is ‘Touchdown’ PCR. 5 In this method, the annealing temperature is set five to ten degrees above the calculated Tm. Annealing temperatures of subsequent cycles are slowly dropped over the course of cycling. Touchdown PCR forces the initial amplification steps to occur at the most stringent annealing conditions possible, with subsequent, less stringent amplification relying on the mass action of the correct amplicon to outcompete any side products. This strategy is useful in that it is a ubiquitous means to get amplification. It is worth trying after a positive control experiment has demonstrated there are no PCR reagent problems.
Figure 3. Amplification of large fragments from human genomic
and lambda DNAs.
Lanes 1-3, JumpStart REDAccuTaq LA amplification
of 5, 10 and 20 kb fragments from human genomic DNA; lane 4, 20 kb
lambda DNA amplification. Lanes 5-8 are the same as 1-4 except using
a competitor’s long and accurate enzyme blend. Markers in lane 9 at 10,
8, 6, 5, 4, 3, 2.5, 2, 1.5, and 1.0 kb (1 kb ladder). Reactions were
assembled and then incubated at room temperature for two hours
before being subjected to identical cycling conditions.
REDTaq DNA polymerase with its optimized reaction buffer performs identically and can directly replace Taq DNA polymerase in most applications. The exception to this is with troublesome PCR. The difference becomes a concern when using non-standard magnesium concentrations.
  1. Linz, U., Protocol optimization and the need for standardization of the polymerase chain reaction. Meth. Mol. Cell. Biol., 2, 98-102 (1991).
  2. Roux, K. H., Optimizing and troubleshooting in PCR. PCR Methods Appl., 4, S185-94 (1995).
  3. Innis M. A and Gelfand, D. H, PCR protocols: a guide to methods and applications, pp. 3-20, (Academic Press, New York, 1990).
  4. Powell, S. J., PCR: essential data, pp. 72-86 (Wiley & Sons, New York, 1995).
  5. Don, R. H. et al., ‘Touchdown’ PCR to circumvent spurious priming during gene amplification. Nucl. Acid Res., 19, 4008 (1991). REDTaq , JumpStart, AccuTaq and JumpStart REDAccuTaq are trademarks of the Sigma-Aldrich Corporation.
Related Links