[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Cloning & Expression](https://www.sigmaaldrich.com/US/en/applications/genomics/cloning-and-expression)Restriction Enzyme Digest Protocol
# Restriction Enzyme Digest Protocol
Our restriction enzyme collection has been optimized for digestion using five unique buffers. When digesting DNA using a single enzyme, use the buffer supplied with the enzyme.
## Protocol for DNA Digestion with a Single Restriction Enzyme
1. Add components to a clean tube in the order shown:
1 µL DNA (concentration 1 µg/µL)
2 µL 10x buffer
1 µL restriction enzyme
16 µL sterile water
2. Incubate the reaction at digestion temperature (usually 37 °C) for 1 hour.
3. Stop the digestion by heat inactivation (65 °C for 15 minutes) or addition of 10 mM final concentration EDTA.
4. The digested DNA is ready for use in research applications.
When using two restriction enzymes at once, first check the enzyme activities in each buffer, using the table on the Restriction Enzyme Buffer Reference. If they both have 100% activity in the same buffer, you can proceed with your double digestion protocol using that buffer. Alternatively, the optimal buffer can be determined from the chart of common double digestions. In some cases, sequential digestion is recommended due to buffer incompatibility (composition or temperature).
## Protocol for DNA Digestion with Two Restriction Enzymes
1. Add components to a clean tube in the order shown:
1 µL DNA (concentration 1 µg/µL)
2 µL 10x buffer
1 µL each restriction enzyme
15 µL sterile water
2. Incubate the reaction at digestion temperature (usually 37 °C) for 1 hour.
3. Stop the digestion by heat inactivation (65°C for 15 minutes) or addition of 10 mM final concentration EDTA.
4. The digested DNA is ready for use in research applications.
## Protocol for Sequential DNA Digestion
1. Add components to a clean tube in the order shown:
1 µL DNA (concentration 1 µg/µL)
2 µL 10x buffer
1 µL restriction enzyme
16 µL sterile water
2. Incubate the reaction at digestion temperature (usually 37 °C) for 1 hour.
3. Stop the digestion by heat inactivation (65 °C for 15 minutes) or addition of 10 mM final concentration EDTA.
4. Recover the DNA using a purification kit: re-suspend and dilute the DNA to 1 µg/µL.
5. Prepare second digestion according to step 1. Continue through step 3.
6. The digested DNA is ready for use in research applications.
__Related Articles__
- [Introduction to Cell Transfection](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/transfection-and-gene-editing/transfection-reagents)
- [Reverse Transfection of Plasmid DNA](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/transfection-and-gene-editing/xtg-reverse-transfect-pdna)
- [3xFLAG® System Expression Vectors for Ultra-Sens](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/3x-flag)
- [Blue-White Screening & Protocols for Colony Selection](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/blue-white-screening)
- [Bacterial Transformation Protocols](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/competent-cells)
- [Genotypes, Phenotypes and Markers](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/definitions-and-genotypes-of-e-coli)
- [Restriction Enzyme Cloning Glossary](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/estriction-enzyme-cloning-manual-glossary)
- [How Transfection Works](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/how-transfection-works)
__Related Product Categories__
- [Detection Substrates & Enzymes](https://www.sigmaaldrich.com/US/en/products/protein-biology/proteins-and-enzymes/detection-substrates-and-enzymes)
- [Biological Buffers](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/biochemicals/biological-buffers)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.