5-Bromo-2′-deoxy-uridine Labeling and Detection Kit II Protocol & Troubleshooting
Product No. 11299964001
Storage of samples after fixation
Storage of samples after fixation with ethanol fixative is possible for up to 2 weeks at +2 to +8 °C.
Reduction of high background
Using BrdU Labeling and Detection Kit II note the following:
a) Anti BrdU-POD will bind nonspecifically to some plastic material. Use sterile flat bottomed microplates that are anti-adsorbant (e.g., Maxisorp from NUNC or Costar). Always use flat bottomed sterile microplates. You can do a further blocking step with PBS/1% BSA before adding the antibody. Non-specific binding of BrdU to microplates can also be overcome by using poly-lysine to change the surface of the wells.
b) Another possible reason for high background: The anti-BrdU is not washed away by the washing steps. In this case, it may help to increase the number of washing steps (1-3- min, 4-5 times).
In addition to these points: Some cell lines show a nonspecific binding of the antibody at a cell concentration of more than 2x104 per ml. This may be due to the extracellular matrix. Reducing the cell number will in this case help to reduce background levels.
Related Links