Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Anti-Digoxigenin-POD (poly), Fab fragments Protocol & Troubleshooting
Product No. 11633716001
Protocol
Substrates for POD:
- BM Blue POD substrate, soluble and precipitating
- DAB substrate
Troubleshooting
DIG Application Manual for Nonradioactive In Situ Hybridization 4th Edition, page 32, Correction for 50× Denhardt’s
50× Denhardt’s:
1% Ficoll (type 400), 1% polyvinylpyrrolidone, and 1% bovine serum albumin.